Thermo Scientific metal coating and thickness gauges
Use this comparison chart to see which system is right for you
Still have questions?
Visit the Noncontact Coating Weight Gauges for Metallic and Coil Coating Lines page, or
Contact an application specialist          Join our mailing list
  RM310EC-LE RM310EC RM310EH RM315EC RM300EL PROSIS
Applications
Zinc (below 300 gsm per side or G200)
Hot dip galvanization lines
Electrolytic galvanization lines
Galvanized (below 500 gsm per side  or G400)
Thick coatings for crash barriers
Zinc alloys (except Galvanneal)
Zn/Al, Zn/Ni, Zn/Al/Mg, Al/Si
✓*        
Galvanneal (iron content in %)          
Tin        
Aluminized coatings          
Chrome          
Paint        
Passivation coatings          
Oil film          
*total coating only
100 - 7000 gsm
LE
X-ray
25kV
X-ray
45kV
X-ray
Kr-85
isotope
Pm-147
isotope
Prosis
reflection
  XRF XRF XRF Beta backscatter Beta backscatter NIR spectroscopy
Applications
Zinc (below 300 gsm per side or G200)
Hot dip galvanization lines
Electrolytic galvanization lines
Galvanized (below 500 gsm per side or G400)
Thick coatings for crash barriers
Zinc alloys (except Galvanneal)
Zn/Al, Zn/Ni, Zn/Al/Mg, Al/Si
✓*      
Galvanneal (iron content in %)        
Tin        
Aluminized coatings ✓*        
Chrome          
Paint      
Passivation coatings          
Oil film          
*total coating only
100 - 7000 gsm
O-frame or single-beam scanners are available
  RM310EC-LE RM310EC RM310EH RM315EC RM300EL PROSIS
Applications
Zinc (below 300 gsm per side or G200)
Hot dip galvanization lines
Electrolytic galvanization lines
Galvanized (below 500 gsm per side  or G400)
Thick coatings for crash barriers
Zinc alloys (except Galvanneal)
Zn/Al, Zn/Ni, Zn/Al/Mg, Al/Si
✓*        
Galvanneal (iron content in %)          
Tin        
Aluminized coatings          
Chrome          
Paint        
Passivation coatings          
Oil film          
*total coating only
100 - 7000 gsm
LE
X-ray
25kV
X-ray
45kV
X-ray
Kr-85
isotope
Pm-147
isotope
Prosis
reflection
  XRF XRF XRF Beta backscatter Beta backscatter NIR spectroscopy
Applications
Zinc (below 300 gsm per side or G200)
Hot dip galvanization lines
Electrolytic galvanization lines
Galvanized (below 500 gsm per side or G400)
Thick coatings for crash barriers
Zinc alloys (except Galvanneal)
Zn/Al, Zn/Ni, Zn/Al/Mg, Al/Si
✓*      
Galvanneal (iron content in %)        
Tin        
Aluminized coatings ✓*        
Chrome          
Paint      
Passivation coatings          
Oil film          
*total coating only
100 - 7000 gsm
O-frame or single-beam scanners are available
Sensor/system compatibility chart
LE
X-ray
25kV
X-ray
45kV
X-ray
Kr-85
isotope
Pm-147
isotope
Prosis
reflection
  XRF XRF XRF Beta backscatter Beta backscatter NIR spectroscopy
RM310EC-LE          
RM310EC        
RM310EH        
RM 315 EC        
RM300EL        
PROSIS          
RM 115 (lab)      
100 - 7000 gsm