Inductively Coupled Plasma Mass Spectrometry (ICP-MS)
An advanced and comprehensive ICP-MS portfolio
Our innovative ICP-MS solutions are designed to meet the needs of all laboratories, from high-throughput and routine to cutting-edge research facilities. With advanced performance for reliable and accurate data generation and simple, intuitive workflows for ease-of-use, our ICP-MS instruments enable you to work smarter now and be ready for future challenges.
Popular ICP-MS products
Featured ICP-MS categories
Total elemental analysis at sub-ppt to ppm levels, this compact, benchtop system delivers powerful, highly sensitive trace detection, fast.
Advanced interference removal for high accuracy even in challenging analytical applications, plus unique ease-of-use. Includes the new solution for semiconductor analysis.
Class-leading elimination of interferences for ultra-sensitive, reliable multi-element analyses at trace levels, even without sample preparation.
Perform precise and accurate isotope ratio analysis in a wide variety of samples.
An intuitive, user-friendly platform software, designed to simplify workflows and maximize efficiency. Enable easy rotation of lab technicians with this common software solution.
ICP-MS technique selector
Features Total elemental analysis at sub-ppt to ppm levels, this compact, benchtop system quickly delivers powerful yet highly sensitive trace level detection. Advanced interference removal for incredible accuracy in challenging and semiconductor-based analytical applications, combined with unique ease-of-use. Class-leading elimination of interferences for ultrasensitive, reliable multi-element analyses at trace levels, even without sample preparation. Combines high resolution, high sensitivity and multicollection with the latest innovations in amplifier technology for multi-isotope analysis down to trace levels.
Applications High quality, high-throughput analysis for routine labs that require sensitive detection limits. Perfect for challenging samples. From routine to research level analyses; for labs seeking to meet current and future regulations, expand into new markets or make new scientific breakthroughs. Suitable for high-matrix samples. Magnetic sector ICP-MS systems especially suitable for semiconductor, geological and material science laboratories, organic and inorganic solutions or solid matrices. Multicollector ICP-MS is the ultimate tool for studying Earth and nuclear sciences as well as biochemistry, food, forensics and medical sciences that require high-precision isotope ratio measurements.
Size Benchtop Benchtop Floor-standing Floor-standing
Key Technology Single quadrupole + flatapole design QCell Triple quadrupole, including flatapole design QCell Double focusing magnetic sector field available with either discrete dynode detector or with additional single Faraday detector. Double-focusing magnetic sector field with moveable Faraday collectors; also available with multiple ion counting devices.
Peripheral Compatibility Full Integration of most LA, IC/LC and GC systems. Compatible with most LA, IC/LC and GC systems
Automation Easily paired with a range of lab automation systems for ultimate throughput.
Software Qtegra Intelligent Scientific Data Solution (ISDS) software ELEMENT Software Suite Multicollector Software Suite
Trace Element Analysis Using ICP-MS for Environmental Samples
Trace Element Analysis Using ICP-MS for Environmental Samples
This webcast series focuses on both routine regulatory monitoring (Part I) and solutions to improve productivity and overcome the challenges of high-matrix samples (Part II). In addition, we will discuss the use of triple quadrupole ICP-MS to meet the needs of environmental researchers and specialty environmental applications.
Featured video
SQ-ICP-MS, TQ-ICP-MS or HR-ICP-MS - an easy guide to finding your perfect solution!
Impurities are a prevalent issue in the semiconductor industry, particularly semiconductor wafer production. The iCAP TQs ICP-MS system from Thermo Scientific offers high performance ICP-MS for ultratrace quantification of trace elements in a variety of semiconductors.
Learn more