What is metabolomics?
What is metabolomics?
Metabolomics [1] is a newly emerging field of 'omics' research. The metabolome [2] is defined as the complete set of small molecule endogenous metabolites, intermediates and metabolism products found in an organism. It can provide an instantaneous snapshot of the entire physiology of a living being. With its potential to provide a comprehensive snapshot of the biochemistry of a biological system, metabolomics can be used for life science research in areas such as disease and biomarker discovery. Metabolomics can also be combined with genomics, transcriptomics and proteomics studies, which are also known as multi-omics, to provide comprehensive insights into biological processes.
Join the Community!
Metabolomics Analysis: Profiling
In general, metabolomics refers to metabolite profiling or the differential study of the metabolome between experimental and control groups when challenged with an external stimulus such as a drug treatment, a biochemical or environmental stress, or pathologies such as mutant/resistance-bred organisms. The stimulus could also be non-biological, such as food processing; as a consequence, metabolomics has huge potential across several application areas, including food and nutrition.
Mass spectrometry-based metabolomics
Because metabolomics aims to comprehensively identify and measure a large number of compounds in complex mixtures, its goals are a challenge for standard analytical chemistry. As a result, mass spectrometry has emerged as an alternative to NMR-based metabolomics, offers high selectivity and sensitivity, and has the potential to assess metabolites in both a qualitative and quantitative manner.
For scientists without specialized training in the field of mass spectrometry, using this approach can feel daunting. There are four fundamental areas one must master in order to be successful in metabolomics:
  1. Experimental design [3]. Separating the biological study into multiple analytical experiments and ensuring quality control for accuracy and precision of results are both paramount to successful experimental design.
  2. Sample preparation [4] is a must when using mass spectrometry-based techniques. There are multiple techniques based on sample type, and there are entire methods for extracting specific classes of metabolites.
  3. Analytical procedures. Small molecule compounds are chemically and structurally diverse, so the mass spectrometer must be combined with a separation technique to reduce sample complexity prior to MS analysis. Depending on the separation technique, specific metabolite classes may be discriminated against and not appear at their normal abundance.
  4. Data analysis. Huge amounts of complex raw data must be analyzed in a stringent manner to remove experimental artifacts. To accomplish this task, stringent statistical tools and software are required for accurate compound identification and quantitation.
Selecting the analytical technique
As endogenous metabolites are diverse in their physico-chemical properties, as well as varying in abundance, a true comprehensive metabolomics study will require orthogonal sample preparation and separation techniques. In reality, most experiments are not comprehensive and there is a bias towards certain classes of compounds.
The mass spectrometer can also introduce bias into the study. Utilizing positive or negative ion mode can affect which compounds can be ionized and detected.
Learn more about metabolomics analysis techniques
Liquid chromatography (LC)-MS offers the broadest coverage of compounds due to its ability to work with different column chemistries. Common LC-MS compounds include lipids, polyamines, alcohols etc.
Samples that are volatile and amenable to chemical derivatization are well suited for gas chromatography (GC)-MS. Also GC offers high resolving power and a low cost-per-sample.
Ion chromatography (IC)-MS is best suited to charged or very polar metabolites that are difficult to analyze by LC-MS, including metabolites such as sugar phosphates and amino acids. IC also offers high resolution, enabling separation of isomers for mass spectrometry analysis.
References
  •  [1] Nicholson J, Connelly J, Lindon J, et al. (2002) Metabonomics: a platform for studying drug toxicity and gene function. Nat Rev Drug Discov 1:153–161. PubMed
  •  [2] Wikipedia contributors, "Metabolome," Wikipedia, The Free Encyclopedia, https://en.wikipedia.org/w/index.php?title=Metabolome&oldid=731647663 (accessed September 12, 2016).
  •  [3] Dunn WB, Wilson ID, Nicholls AW et al. (2012) The importance of experimental design and QC samples in large-scale and MS-driven untargeted metabolomic studies of humans. Bioanalysis 4(18):2249–2264. PubMed
  •  [4] Raterink RJ, Lindenburg PW, Vreeken, RJ, et al. (2014) Recent developments in sample-pretreatment techniques for mass spectrometry-based metabolomics. TrAC Trends in Analytical Chemistry 61:157–167. ScienceDirect