Determine bacterial counts without delay
Detect microbial contamination or determine bacterial counts in clinical specimens with easy-to-use, easy-to-read dip slides. Dip slides are coated with two different types of media on a double-sided stick and come complete with a transport container and growth chamber all-in-one. Dip slides decrease the time between collection and inoculation which increases recovery and accuracy of results.
Make colony counting simple for aerobic bacteria, yeasts and fungi with the easily disposable Thermo Scientific™ Dip-Slides™. 
Make colony counting simple for aerobic bacteria, yeast, mold, coliform bacteria and Enterobacteriaceae with the easily disposable Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial.
Make colony counting for the bacterial assessment of urine simple with the easily disposable Thermo Scientific™ Oxoid™ Dip-Slides™ Clinical. 
Download the prepared media selection guide and use it to discover what we can offer to ensure maximum testing flexibility. Organized by organism type and by functionality, we have plates, bottles, bagged media, dip-slides, diluents and more. Take a look and find the right product to solve your next challenge.
Download the guide
Resources