Our LanthaScreen kinase activity assay toolbox of "mix and match" assay reagents is designed to enable rapid assay configuration. Our portfolio of phospho-specific antibodies, matched fluorescent peptide substrates and in-house produced kinases have been tested in >200 different kinase assays to date.
Phospho-specific Antibodies with Peptide Substrates
To facilitate in-house kinase assay configuration and screening, we have developed a selection of Tb-labeled phospho-specific antibodies, which may be accessed in small scale and screening quantities.  Matched and validated substrates are available for kinase assay configuration.
These antibodies may also be available with Europium labels, contact us at discoverysciences@invitrogen.com, for more information.
Tyrosine Kinase Assays
SKU Product Catalog Size
PV5692 LanthaScreen Eu-PY20 Antibody Kit 25 μg
PV5696 LanthaScreen Eu-PY20 Antibody Kit 1 mg
PV5693 Alexa Fluor 647-Poly GT
1 nmol
PV5836 Alexa Fluor 647-Poly GT 10 nmol
PV3552 LanthaScreen Tb-PY20 Antibody Kit 25 μg
PV3553 LanthaScreen Tb-PY20 Antibody Kit 1 mg
PV3610 Fluorescein-Poly GT, 30 μM 1 mL
PV3611 Fluorescein-Poly GAT, 30 μM 1 mL
Serine/Threonine Kinase Assays
SKU Product Catalog Size
PV3560 LanthaScreen Tb-pSer (PKC) Antibody Kit 25 μg
PV3561 LanthaScreen Tb-pSer (PKC) Antibody Kit 1 mg
PV3506 Fluorescein-PKC Substrate 1 mg
PV3562 LanthaScreen Tb-IKBa [pSer32] Antibody Kit 2 5μg
PV3563 LanthaScreen Tb-IKBa [pSer32] Antibody Kit 1 mg
PV3507 Fluorscein-IKK substrate 1 mg
PV3564 LanthaScreen Tb-pCrosstide Antibody Kit 25 μg
PV3565 LanthaScreen Tb-pCrosstide Antibody Kit 1 mg
PV3509 Fluorescein-Crosstide Substrate 1 mg
PV3542 LanthaScreen Tb-CREB pSer133 Antibody
25 μg
PV3508 Fluorescein-CREBtide Substrate, 1 mg/mL 1 mg
PV4899 LanthaScreen Tb-pERM (pLRRKtide) Antibody Kit 25 μg
PV4900 LanthaScreen Tb-pERM (pLRRKtide) Antibody Kit 1 mg
PV4901 Fluorescein-ERM (LRRKtide) Substrate 1 mg
PV5130 LanthaScreen Tb-anti-phospho-p53 [pSer15] Antibody Kit 25 μg
PV5131 LanthaScreen Tb-anti-phospho-p53 [pSer15] Antibody Kit 1 mg
PV5132 Fluorescein-p53 [Ser15] Substrate 1 mg
Phospho-specific Antibodies with Physiological Protein Substrates
As an extension to our peptide-based substrates for LanthaScreen® kinase activity assays, we now offer physiological protein substrates that are directly labeled with a green fluorescent protein (GFP) acceptor fluorophore. Pairing the GFP-labeled substrate with a corresponding terbium (Tb)-labeled phosphospecific antibody allows detection of the phosphorylated product.
Ordering Information
SKU Product Catalog Size
PV4451 LanthaScreen Tb-ATF2 [pSer71] Antibody Kit 25 μg
PV4452 LanthaScreen Tb-ATF2 [pSer71] Antibody Kit 1 mg
PV4445 GFP-ATF2 (19-96) 20 nmol
PV4453 LanthaScreen Tb-c-Jun [pSer73] Antibody Kit 25 μg
PV4454 LanthaScreen Tb-c-Jun [pSer73] Antibody Kit 1 mg
PV4446 GFP-c-jun (1-179) 20 nmol
PV4757 LanthaScreen Tb-4E-BP1 [pThr46] Antibody Kit 25 μg
PV4758 LanthaScreen Tb-4E-BP1 (pThr46) Antibody Kit 1 mg
PV4759 GFP-4E-BP1 20 nmol
PV4810 LanthaScreen Tb-eIF2a [pSer52] Antibody Kit 25 μg
PV4811 LanthaScreen Tb-eIF2a [pSer52] Antibody Kit 1 mg
PV4809 GFP-eIF2a 20 nmol
PV4813 LanthaScreen Tb-MAP2K1 [pSer217/221] Antibody Kit 25 μg
PV4814 LanthaScreen Tb-MAP2K1 [pSer217/221] Antibody Kit 1 mg
PV4812 Fluorescein-MEK1 20 nmol
PV4844 LanthScreen Tb-anti-STAT1 [pTyr701] Antibody Kit 25 μg
PV4845 LanthScreen Tb-anti-STAT1 [pTyr701] Antibody Kit 1 mg
PV5211 GFP-STAT1 20 nmol
PV4846 LanthaScreen Tb-STAT3 [pTyr705] Antibody Kit 25 μg
PV4847 LanthaScreen Tb-STAT3 [pTyr705] Antibody Kit 1 mg
PV5212 GFP-STAT3 20 nmol
Buffers and Other Supplemental Assay Reagents
To complete our offering of antibodies and substrates for the development of TR-FRET kinase assays, we offer a selection of validated kinase buffers and other assay reagents.
Ordering Information
SKU Product Catalog Size
PV3574 TR-FRET Dilution Buffer 100 mL
PV3189 5X Kinase Buffer A 4  mL
PV5125 5X Kinase Buffer Q 4 mL
PV5101 5X Kinase Buffer R 4 mL
PV5213 5X Kinase Buffer S 4 mL
PV4794 1X mTOR Assay Buffer 20 mL
P2825 Kinase Quench Buffer 1 mL
PV3227 10 mM ATP 500 μL
P2325 1M DTT 1 mL
More Information
Generic LanthaScreen reagents