Eu Kinase Binding Assay
On this page you will find:
  • A table listing the names and accession numbers of kinases validated in the assay, the reagents validated for each kinase, and links to detailed protocols
  • A summary of assay requirements
Assay Requirements
A LanthaScreen® Eu Kinase Binding Assay requires three primary components: a kinase protein, a kinase tracer, and a europium-labeled antibody.  Each of the three components is listed in the selection table with a link to the web page for each individual product.  Kinase proteins and antibodies have small and large sizes available in our web catalog.  Please inquire for bulk 500 µL sizes of our Kinase Tracers.  A plate reader capable of reading the TR-FRET signal produced by the assay is also required.  Plate reader information and support is available here.
  • Kinase protein—Kinases sold by Life Technologies and validated in the assay are listed in the table.
  • Kinase tracer—Six different kinase tracers enable over 300 validated assays.
  • Europium-labeled antibody—The optimal Life Technologies antibody for each kinase is listed.
  • Assay buffer—For most LanthaScreen® Eu Kinase Binding Assays the recommended buffer is 5X Kinase Buffer A.  The protocol for each assay indicates the optimal buffer.
The TR-FRET LanthaScreen® Eu Kinase Binding Assay can be applied to kinases of interest not included in the selection table.
No records were found matching your criteria
Kinase Accession # Recommended Antibody Recommended Tracer Detailed Protocol
ABL1 NP_005148.2 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 1710  
ABL1 E255K
NP_005148.2, E255K
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 1710  
ABL1 G250E
NP_005148.2, G250E
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 1710  
ABL1 H396P NP_005148.2 Eu-Anti-GST Kinase Tracer 1710  
ABL1 M351T NP_005148.2 Eu-Anti-GST Kinase Tracer 1710  
ABL1 Q252H NP_005148.2 Eu-Anti-GST Kinase Tracer 1710  
ABL1 Y253F
NP_005148.2, Y253F
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 1710  
ABL2 (ARG) NP_009298.1 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
ACVR1 (ALK2) NP_001096.1 Eu-Anti-GST Kinase Tracer 236  
ACVR1 (ALK2) R206H NP_001096.1 Eu-Anti-GST Kinase Tracer 236  
ACVR2A NP_001607.1 Eu-Anti-GST Kinase Tracer 178
ACVR2B NP_001097.2 Eu-Anti-GST Kinase Tracer 178
ALK NP_004295 Eu-Anti-GST Kinase Tracer 236  
ALK C1156Y NP_004295.2
Eu-Anti-GST Kinase Tracer 236  
ALK F1174L NP_004295.2 Eu-Anti-GST Kinase Tracer 236  
ALK L1196M NP_004295.2 Eu-Anti-GST Kinase Tracer 236  
ALK R1275Q NP_004295.2 Eu-Anti-GST Kinase Tracer 236  
AMPK (A1/B1/G2) NP_006242.5 Eu-Anti-His Kinase Tracer 236  
AMPK (A1/B1/G3) NP_006242.5 Eu-Anti-His Kinase Tracer 236  
AMPK (A1/B2/G1) NP_006242.5 Eu-Anti-His Kinase Tracer 236  
AMPK (A2/B2/G1) NP_006243.2 Eu-Anti-His Kinase Tracer 236  
AMPK (A2/B2/G2) NP_006243.2 Eu-Anti-His Kinase Tracer 236  
AMPK A1/B1/G1
PRKAA1:  NP_006242.4; PRKAB1: NP_006244.2; PRKAG1:  NP_002724.1
Eu-Anti-GST Kinase Tracer 236  
AMPK A2/B1/G1
PRKAA1:  NP_006243.2; PRKAB1: NP_006244.2; PRKAG1:  NP_002724.1
Eu-Anti-GST Kinase Tracer 236  
AURKA (AURORA A) NP_940839
Eu-Anti-His Kinase Tracer 236  
AURKB (AURORA B) NP_004208
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
AXL NP_068713
Eu-Anti-His Kinase Tracer 236  
AXL R499C NP_068713.2
Eu-Anti-GST Kinase Tracer 236  
BLK
NP_001706
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
BMPR1A (ALK3)
NP_004320.2 Eu-Anti-GST Kinase Tracer 178
BMPR1B (ALK6) NP_001194.1 Eu-Anti-GST Kinase Tracer 178
BMPR2
NP_001195.2 Eu-Anti-His Kinase Tracer 236  
BMX NP_001712 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
BRAF NP_004324.2 Eu-Anti-GST Kinase Tracer 178
BRAF V599E
NP_004324.1, V599E
Eu-Anti-GST Kinase Tracer 178  
BRSK1 (SAD1) NP_115806
Eu-Anti-GST Kinase Tracer 236  
BRSK2 NP_003948.2 Eu-Anti-GST Kinase Tracer 236  
BTK NP_000052 Eu-Anti-His Kinase Tracer 236  
CAMK1D (CAMKI DELTA) NP_705718 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CAMK2A (CAMKII ALPHA) NP_741960 Eu-Anti-His Kinase Tracer 236  
CAMK2B (CAMKII BETA) NP_742078 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CAMK2D (CAMKII DELTA) NP_742113 Eu-Anti-His Kinase Tracer 236  
CAMK2G (CaMKII gamma) NP_751909.1
Contains S36P SNP
Eu-Anti-GST Kinase Tracer 236  
CAMK4 (CAMKIV) NP_001735 Eu-anti- GST Kinase Tracer 236  
CAMKK1 (CAMKKA) NP_115670 Eu-Anti-His Kinase Tracer 236  
CAMKK2 (CAMKK BETA) NP_757380.1 Eu-anti-GST Kinase Tracer 236  
CASK NP_003679.2 Eu-anti-GST Kinase Tracer 236  
CDC42 BPA (MRCKA) NP_055641 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CDC42 BPB (MRCKB)
NP_006026.2 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CDC7/DBF4 NP_003494.1 Eu-anti-GST Kinase Tracer 236  
CDK1/cyclin A2
NP_001777.1 Eu-anti-GST Kinase Tracer 236  
CDK1/CYCLIN B
NP_001777; NP_114172
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CDK11 (Inactive) NP_055891.1 Eu-anti-GST Kinase Tracer 236  
CDK14 (PFTK1)/cyclin Y NP_036527.1 Eu-anti-GST Kinase Tracer 236  
CDK16 (PCTK1)/cyclin Y NP_055891.1 Eu-anti-GST Kinase Tracer 236  
CDK2/CYCLIN A
NP_001789; NP_001228
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CDK2/cyclin A1 NP_036527.1 Eu-anti-GST Kinase Tracer 236  
CDK2/cyclin E1 NP_006192.1 Eu-anti-GST Kinase Tracer 236  
CDK2/cyclin O NP_001789.2 Eu-anti-GST Kinase Tracer 236  
CDK3/cyclin E1
NP_001249.1 Eu-anti-GST Kinase Tracer 236  
CDK5 (Inactive) NP_004926.1 Eu-anti-GST Kinase Tracer 236  
CDK5/P25
NP_004926.1; NP_003876
Eu-anti-GST Kinase Tracer 236  
CDK7/CYCLIN H/MNAT1
NP_001790 (Q130R); NP_001230; NP_002422
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CDK8/CYCLIN C NP_001789.2 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CDK9 (Inactive) NP_001252.1 Eu-anti-GST Kinase Tracer 236  
CDK9/CYCLIN K
NP_001252; NP_003849
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CDK9/CYCLIN T1 NP_004926.1 Eu-Anti-His Kinase Tracer 236  
CHEK1 (CHK1) NP_001265 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CHEK2 (CHK2) NP_009125 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
CHUK (IKK ALPHA) NP_001269 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CLK1 NP_004062 Eu-anti-GST Kinase Tracer 236  
CLK2 NP_001252.1 Eu-anti-GST Kinase Tracer 236  
CLK4 NP_065717 Eu-anti-GST Kinase Tracer 236  
CSF1R (FMS) NP_005202 Eu-Anti-His Kinase Tracer 236  
CSK NP_004374 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
CSNK1D (CK1 DELTA) NP_620693
Eu-Anti-GST Kinase Tracer 199  
CSNK1E (CK1 EPSILON) NP_001885 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
CSNK1G1 (CK1 GAMMA 1) NP_071331 Eu-Anti-GST Kinase Tracer 222  
CSNK1G2 (CK1 GAMMA 2) NP_001310 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
CSNK1G3 (CK1 GAMMA 3)
NP_004375, R174G
Eu-Anti-GST Kinase Tracer 222  
DAPK2 NP_055141 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
DDR1 NP_001945 Eu-Anti-GST Kinase Tracer 178
DDR2
NP_006173, S642A
Eu-Anti-GST Kinase Tracer 178  
DDR2 N456S NP_006173.2 Eu-Anti-GST Kinase Tracer 236  
DDR2 T654M NP_006173.2 Eu-Anti-GST Kinase Tracer 178  
DMPK NP_004400 Eu-Anti-GST Kinase Tracer 236  
DYRK1A NP_001387 Eu-Anti-GST Kinase Tracer 236  
DYRK1B NP_004705 Eu-Anti-GST Kinase Tracer 236  
DYRK2 NP_006473.2 Eu-Anti-GST Kinase Tracer 236  
DYRK3 NP_003573 Eu-Anti-GST Kinase Tracer 236  
EGFR (ERBB1) NP_005219.2 Eu-Anti-GST Kinase Tracer 199  
EGFR (ERBB1) L858R
NP_005219.2. L858R
Eu-Anti-GST Kinase Tracer 199  
EGFR (ERBB1) L861Q
NP_005219.2, L861Q
Eu-Anti-GST Kinase Tracer 199  
EGFR (ErbB1) T790M NP_005219.2 Eu-Anti-GST Kinase Tracer 236  
EGFR (ErbB1) T790M L858R NP_005219.2 Eu-Anti-GST Kinase Tracer 199  
EGFR d746-750 NP_005219.2 Eu-Anti-GST Kinase Tracer 199  
EIF2AK2 NP_002750.1 Eu-Anti-GST Kinase Tracer 236  
EPHA1 NP_005223.2
Eu-Anti-GST Kinase Tracer 236  
EPHA2 NP_004422.2 Eu-Anti-GST Kinase Tracer 236  
EPHA3 NP_005224
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
EPHA4 NP_004429.1
Eu-Anti-GST Kinase Tracer 236  
EPHA5 NP_004430.1
Eu-Anti-GST Kinase Tracer 236  
EPHA6 NP_001073917.2 Eu-Anti-GST Kinase Tracer 236  
EPHA7 NP_004431.1 Eu-Anti-GST Kinase Tracer 236  
EPHA8 NP_065387 Eu-Anti-GST Kinase Tracer 236  
EPHB1 NP_004432 Eu-Anti-GST Kinase Tracer 236  
EPHB2 NP_004433 Eu-Anti-GST Kinase Tracer 236  
EPHB3 NP_004434 Eu-Anti-GST Kinase Tracer 178  
EPHB4 NP_004435 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 178  
ERBB4 (HER4) NP_005226 Eu-Anti-GST Kinase Tracer 178  
FER NP_005237
Eu-Anti-GST Kinase Tracer 236  
FES (FPS) NP_001996 Eu-Anti-His Kinase Tracer 236  
FGFR1 NP_000595 Eu-Anti-His Kinase Tracer 236  
FGFR1 V561M NP_075598.2 Eu-Anti-GST Kinase Tracer 236  
FGFR2
NP_075261
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
FGFR3
NP_000133 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
FGFR3 G697C NP_000133.1 Eu-Anti-GST Kinase Tracer 236  
FGFR3 K650E
NP_000133, K650E
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
FGFR3 K650M NP_000133.1 Eu-Anti-GST Kinase Tracer 236  
FGR NP_005239 Eu-Anti-His Kinase Tracer 236  
FLT1 (VEGFR1) NP_002010 Eu-Anti-GST Kinase Tracer 236  
FLT3 NP_004110 Eu-Anti-His Kinase Tracer 236  
FLT3 D835Y
NP_004110, D835Y
Eu-Anti-His Kinase Tracer 236  
FLT3 ITD
NP_004110.2 Eu-Anti-GST Kinase Tracer 236  
FLT4 (VEGFR3)
NP_891555
Eu-Anti-GST Kinase Tracer 236  
FRAP1 (MTOR) NP_004949.1 Eu-Anti-GST Kinase Tracer 314  
FRK (PTK5) NP_002022 Eu-Anti-GST Kinase Tracer 236  
FYN NP_694592 Eu-Anti-His Kinase Tracer 236  
FYN A
NP_002028.1 Eu-Anti-GST Kinase Tracer 236  
GRK1 NP_002920.1 Eu-Anti-GST Kinase Tracer 236  
GRK4 NP_892027 Eu-Anti-GST Kinase Tracer 236  
GRK5 NP_005299
Eu-Anti-GST Kinase Tracer 236  
GRK6 NP_001004106 Eu-Anti-GST Kinase Tracer 236  
GRK7 NP_631948 Eu-Anti-GST Kinase Tracer 236  
GSK3A (GSK3 alpha) NP_063937.2
Eu-Anti-GST Kinase Tracer 236  
GSK3B (GSK3 BETA) NP_002084 Eu-Anti-His Kinase Tracer 236  
HCK NP_002101 Eu-Anti-GST Kinase Tracer 236  
HIPK3 (YAK1) NP_005725.2 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
ICK NP_055735.1
Eu-Anti-GST Kinase Tracer 236  
IGF1R NP_000866 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
IKBKB (IKK BETA) NP_001547 Eu-Anti-GST Kinase Tracer 236  
IKBKE (IKK EPSILON) NP_054721 Eu-Anti-GST Kinase Tracer 236  
INSR NP_000199 Eu-Anti-GST Kinase Tracer 236  
IRAK1 NP_001560 Eu-Anti-GST Kinase Tracer 236  
IRAK4
NP_057207, AAH13316
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
ITK NP_005537 Eu-Anti-GST Kinase Tracer 236  
JAK2 NP_004963 Eu-Anti-GST Kinase Tracer 236  
JAK2 JH1 JH2 NP_004963 Eu-Anti-GST Kinase Tracer 236  
JAK3 NP_000206
Eu-Anti-GST Kinase Tracer 236  
KDR (VEGFR2) NP_002244 Eu-Anti-His Kinase Tracer 236  
KIT
NP_000213
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
KIT A829P NP_000213.1 Eu-Anti-GST Kinase Tracer 222  
KIT D816H NP_000213.1 Eu-Anti-GST Kinase Tracer 222  
KIT D816V NP_000213.1 Eu-Anti-GST Kinase Tracer 222  
KIT D820E NP_000213.1 Eu-Anti-GST Kinase Tracer 222  
KIT N822K NP_000213.1 Eu-Anti-GST Kinase Tracer 222  
KIT T670E NP_000213.1
Eu-Anti-GST Kinase Tracer 236  
KIT T670I
NP_000213, T670I
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
KIT V559D T670I NP_000213.1 Eu-Anti-GST Kinase Tracer 222  
KIT V654A
NP_000213, V654A
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
KIT Y823D NP_000213.1 Eu-Anti-GST Kinase Tracer 222  
LATS1 NP_004681.1 Eu-Anti-GST Kinase Tracer 236  
LATS2 NP_055387.2
Eu-Anti-GST Kinase Tracer 236  
LCK NP_005347 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
LIMK1 NP_002305
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
LIMK2 NP_005560
Eu-Anti-GST Kinase Tracer 178  
LRRK2 NP_940980.2 Eu-Anti-GST Kinase Tracer 236  
LRRK2 G2019S NP_940980.2 (G2019S) Eu-Anti-GST Kinase Tracer 236  
LRRK2 I2020T NP_940980.2 (I2020T) Eu-Anti-GST Kinase Tracer 236  
LRRK2 R1441C NP_940980.2 (R1441C) Eu-Anti-GST Kinase Tracer 236  
LTK (TYK1) NP_002335.2
Eu-Anti-GST Kinase Tracer 236  
LYN A NP_002341.1 Eu-anti-GST Kinase Tracer 236 Inquire Here
LYN B AAH59394 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MAP2K1 (Inactive) NP_002746 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MAP2K1 (MEK1) NP_002746 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MAP2K1 (MEK1) S218D S222D
NP_002746, S218D, S222D
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MAP2K2 (MEK2) NP_109587 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MAP2K3 (MEK3) NP_002747 Eu-Anti-GST Kinase Tracer 236  
MAP2K6 (MKK6) NP_002749 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MAP2K6 (MKK6) S207E T211E
NP_002749, S207E, T211E
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MAP3K10 (MLK2) NP_002437 Eu-Anti-GST Kinase Tracer 236  
MAP3K11 (MLK3) NP_002746
Eu-Anti-GST Kinase Tracer 236  
MAP3K14 (NIK) NP_003945.2 Eu-Anti-GST Kinase Tracer 236  
MAP3K2 (MEKK2) AAF63496.3 Eu-Anti-GST Kinsae Tracer 178  
MAP3K3 (MEKK3) NP_002392 Eu-Anti-GST Kinase Tracer 236  
MAP3K5 (ASK1) NP_005914
Eu-Anti-GST Kinase Tracer 236  
MAP3K7/MAP3K7IP1 (TAK1-TAB1)
NP_663306; NP_006107
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
MAP3K9 (MLK1)
NP_149132 Eu-Anti-GST Kinase Tracer 236  
MAP4K1 (HPK1) NP_009112.1 Eu-Anti-GST Kinase Tracer 222  
MAP4K2 (GCK) NP_004570.2 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MAP4K3 (GLK) NP_003609.2 Eu-Anti-GST Kinase Tracer 236  
MAP4K4 (HGK) NP_004825 Eu-Anti-GST Kinase Tracer 236  
MAP4K5 (KHS1)
NP_942089
Eu-Anti-GST Kinase Tracer 236  
MAPK10 (JNK3) NP_002744 Eu-Anti-GST Kinase Tracer 199  
MAPK12 (Inactive) NP_002960 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MAPK13 (Inactive) NP_002745 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MAPK14 (Inactive)
NP_620581
Eu-Anti-GST Kinase Tracer 199  
MAPK14 (P38 ALPHA) NP_620581 Eu-Anti-GST Kinase Tracer 199  
MAPK15 (ERK7) NP_620590.2 Eu-Anti-His Kinase Tracer 236  
MAPK8 (JNK1) NP_002741.1 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 199  
MAPK9 (Inactive) NP_002743 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MAPK9 (JNK2) NP_002743
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 199  
MAPKAPK5 (PRAK) NP_003659
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
MARK1 (MARK) NP_002960 Eu-Anti-GST Kinase Tracer 236  
MARK2 NP_059672.2 Eu-Anti-GST Kinase Tracer 236  
MARK4 NP_002745
Eu-Anti-GST Kinase Tracer 236  
MELK NP_055606.1
Eu-Anti-GST Kinase Tracer 236  
MERTK (CMER)
NP_620581 Eu-Anti-GST Kinase Tracer 236  
MERTK (cMER) A708S
NP_006334.2
Eu-Anti-GST Kinase Tracer 236  
MET (CMET) NP_000236.2
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
MET D1228H NP_000236.2 Eu-Anti-GST Kinase Tracer 236  
MET M1250T
NP_000236.2 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
MINK1 NP_002743
Eu-Anti-GST Kinase Tracer 236  
MKNK2 (MNK2) NP_060042.2
Eu-Anti-GST Kinase Tracer 236  
MLCK (MLCK2)
NP_872299, R175G
Eu-Anti-GST Kinase Tracer 236  
MST1R (RON)
NP_002438 Eu-Anti-GST Kinase Tracer 236  
MUSK NP_005583.1
Eu-Anti-GST Kinase Tracer 236  
MYLK (MLCK) NP_444253 Eu-Anti-GST Kinase Tracer 236  
MYLK2 (SKMLCK) NP_149109 Eu-Anti-GST Kinase Tracer 236  
MYO3B (MYO3 beta)
NP_620482.3 Eu-Anti-GST Kinase Tracer 236  
NEK2
NP_002488
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
NLK NP_057315.1 Eu-Anti-GST Kinase Tracer 178
NUAK1 (ARK5) NP_055655 Eu-Anti-His Kinase Tracer 236  
NUAK2 NP_112214.1 Eu-Anti-GST Kinase Tracer 222  
PAK4 NP_005875 Eu-Anti-GST Kinase Tracer 236  
PAK6 NP_064553 Eu-Anti-His Kinase Tracer 236  
PAK7 (KIAA1264) NP_065074 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
PDGFRA (PDGFR ALPHA) NP_006197 Eu-Anti-GST Kinase Tracer 236  
PDGFRA D842V
NP_006197, D842V
Eu-Anti-GST Kinase Tracer 236  
PDGFRA T674I
NP_006197, T674I
Eu-Anti-GST Kinase Tracer 236  
PDGFRA V561D
NP_006197, V561D
Eu-Anti-GST Kinase Tracer 236  
PDGFRB (PDGFR BETA) NP_002600 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
PDK1 NP_002604 Eu-Anti-His Kinase Tracer 236  
PHKG1 NP_006204 Eu-Anti-GST Kinase Tracer 236  
PHKG2 NP_000285 Eu-Anti-GST Kinase Tracer 236  
PIK3CA/PIK3R1 (P110 ALPHA/p85 Alpha)
PIK3CA: NP_006209.2; PIK3R1(p110a): NP_852664.1
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 1710
PIK3CD/PIK3R1 (P110 DELTA/P85 alpha)
NP_005017.2; NP_852664.1
Eu-Anti-GST Kinase Tracer 314 Inquire Here
PIK3CG (P110 GAMMA NP_002640.2
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 314  
PIM1 NP_002639 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
PIM2
NP_006866 Eu-Anti-GST Kinase Tracer 236  
PKN1 (PRK1) NP_998725 Eu-Anti-GST Kinase Tracer 236  
PLK4
NP_055079.3
Eu-Anti-GST Kinase Tracer 236  
PRKACA (PKA)
NP_002721.1 Eu-Anti-His Kinase Tracer 236  
PRKACB (PRKAC beta)
NP_002722.1
Eu-Anti-GST Kinase Tracer 236  
PRKACG (PRKAC gamma)
NP_002723.2 Eu-Anti-GST Kinase Tracer 236  
PRKCH (PKC ETA) NP_006246
Eu-Anti-His Kinase Tracer 236  
PRKCI (PKC IOTA) NP_002731 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
PRKCN (PKD3) NP_005804
Eu-Anti-GST Kinase Tracer 236  
PRKCQ (PKC THETA) NP_006248 Eu-Anti-His Kinase Tracer 236  
PRKD1 (PKC MU)
NP_002733 Eu-Anti-GST Kinase Tracer 236  
PRKD2 (PKD2) NP_057541 Eu-Anti-GST Kinase Tracer 236  
PRKG1 NP_006249 Eu-Anti-GST Kinase Tracer 236  
PRKG2 (PKG2) NP_006250 Eu-Anti-GST Kinase Tracer 236  
PRKX NP_005035 Eu-Anti-GST Kinase Tracer 236  
PTK2 (FAK) NP_722560
Eu-Anti-GST Kinase Tracer 236  
PTK2B (FAK2) NP_004094 Eu-Anti-GST Kinase Tracer 236  
PTK6 (BRK)
NP_005966 Eu-Anti-His Kinase Tracer 178  
RAF1 (CRAF) Y340D Y341D
NP_002871, Y340D, Y341D
Eu-Anti-GST Kinase Tracer 178  
RET NP_066124 Eu-Anti-GST Kinase Tracer 236  
RET G691S NP_066124.1 Eu-Anti-GST Kinase Tracer 236  
RET M918T NP_066124.1 Eu-Anti-GST Kinase Tracer 236  
RET V804L
NP_066124, V804L
Eu-Anti-GST Kinase Tracer 236  
RET V804M NP_066124.1 Eu-Anti-GST Kinase Tracer 236  
RET Y791F
NP_066124, Y791F
Eu-Anti-GST Kinase Tracer 236  
RIPK2
NP_003812 Eu-Anti-His Kinase Tracer 199  
RIPK3
NP_006862.2 Eu-Anti-GST Kinase Tracer 178
ROCK1 NP_005397 Eu-Anti-GST Kinase Tracer 236  
ROCK2 NP_004841 Eu-Anti-GST Kinase Tracer 236  
ROS1 NP_002935
Eu-Anti-GST Kinase Tracer 236  
RPS6KA1 (RSK1)
NP_002944 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
RPS6KA2 (RSK3) NP_066958 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
RPS6KA3 (Inactive)
NP_004577 Eu-Anti-His Kinase Tracer 236  
RPS6KA3 (RSK2) NP_004577 Eu-Anti-His Kinase Tracer 236  
RPS6KA4 (MSK2)
NP_003933
Eu-Anti-GST Kinase Tracer 236  
RPS6KA5 (MSK1) NP_004746.2 Eu-Anti-GST Kinase Tracer 236  
RPS6KA6 (RSK4) NP_055311 Eu-Anti-GST Kinase Tracer 236  
RPS6KB1 (P70S6K)
NP_003152, T412E
Eu-Anti-GST Kinase Tracer 236  
SGK (SGK1)
NP_005618, S589D
Eu-Anti-GST Kinase Tracer 236  
SGK3 (SGKL)
NP_037389, S487D
Eu-Anti-GST Kinase Tracer 236  
SIK1 NP_775490.2 Eu-Anti-GST Kinase Tracer 236  
SIK3 NP_079440.3 Eu-Anti-GST Kinase Tracer 236  
SLK NP_055535.1 Eu-Anti-GST Kinase Tracer 236  
SNF1LK2
NP_056006.1 Eu-Anti-GST Kinase Tracer 236  
SRC NP_005408 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
SRPK2 NP_004577 Eu-Anti-GST Kinase Tracer 236  
STK16 (PKL12)
NP_003682
Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
STK17A (DRAK1)
NP_004751 Eu-Anti-GST Kinase Tracer 236  
STK17B (DRAK2)
NP_004217.1 Eu-Anti-GST Kinase Tracer 222  
STK22D (TSSK1)
NP_114417 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236  
STK25 (YSK1) NP_006365
Eu-Anti-GST Kinase Tracer 236  
STK3 (MST2) NP_006272.2 Eu-Anti-GST Kinase Tracer 222  
STK32B (YANK2) NP_060871.1
Contains R198G SNP
Eu-Anti-GST Kinase Tracer 236  
STK32C (YANK3) NP_775846.2
Eu-Anti-GST Kinase Tracer 236  
STK33
NP_112168 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 222  
STK38 (NDR) NP_009202.1 Eu-Anti-GST Kinase Tracer 236  
STK38L (NDR2) NP_055815.1 Eu-Anti-GST Kinase Tracer 236  
STK39 (STLK3) NP_037365.2 Eu-Anti-His Kinase Tracer 236  
STK4 (MST1)
NP_006273 Eu-Anti-GST Kinase Tracer 222  
SYK NP_003168 Eu-Anti-GST Kinase Tracer 236  
TAOK1 NP_065842.1 Eu-Anti-GST Kinase Tracer 236  
TAOK2 (TAO1)
NP_004774 Eu-Anti-GST Kinase Tracer 236  
TAOK3 (JIK) NP_057365 Eu-Anti-GST Kinase Tracer 236  
TBK1 NP_037386
Eu-Anti-His Kinase Tracer 236  
TEC NP_003206 Eu-Anti-His Kinase Tracer 178  
TEK (TIE2) NP_000450 Eu-Anti-GST Kinase Tracer 236  
TEK (TIE2) R849W NP_000450.2 Eu-Anti-GST Kinase Tracer 236  
TEK (TIE2) Y1108F NP_000450.2 Eu-Anti-GST Kinase Tracer 236  
TESK2 NP_009101.2 Eu-Anti-GST Kinase Tracer 178
TGFBR1 (ALK5) NP_004603.1 Eu-Anti-GST Kinase Tracer 178  
TGFBR2 NP_003233
Eu-Anti-GST Kinase Tracer 178  
TLK1 NP_036422.3 Eu-Anti-GST Kinase Tracer 236  
TLK2 NP_006843.2
Eu-Anti-GST Kinase Tracer 236  
TNIK NP_055843.1 Eu-Anti-GST Kinase Tracer 236  
TNK2 (ACK) NP_005772.3 Eu-Anti-GST Kinase Tracer 236  
TTK NP_003309 Eu-Anti-GST Kinase Tracer 236  
TYK2 NP_003322.2
Eu-Anti-GST Kinase Tracer 236  
TYRO3 (RSE) NP_006284
Eu-Anti-GST Kinase Tracer 236  
ULK1
NP_003556.1
Eu-Anti-GST Kinase Tracer 236  
ULK2
NP_055498.3
Contains V370M SNP
Eu-Anti-GST Kinase Tracer 236  
ULK3 NP_001271293.1 Eu-Anti-His Kinase Tracer 236  
WEE1 NP_003381.1
Eu-Anti-GST Kinase Tracer 178  
WNK2 NP_006639.3 Eu-Anti-GST Kinase Tracer 178  
WNK3 NP_065973 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 1710
YES1 NP_005424
Eu-Anti-GST Kinase Tracer 236 Inquire Here
ZAP70
NP_001070 Biotin-Anti-His | Eu-Streptavidin Kinase Tracer 236