NanoDrop Products Guide
Pipette. Measure. Know.
Quantify DNA, RNA and protein samples in seconds with only 1–2µL using Thermo Scientific™ NanoDrop™ instruments. Whether you choose a NanoDrop UV-Vis spectrophotometer or fluorospectrometer, you can rely on patented* NanoDrop technology for accurate, fast results to assess sample concentration and purity. 
Request a quote ›  Free trial program ›
Choose the NanoDrop instrument that's right for you
Simplify DNA, RNA, and protein quantitation with direct sample measurement.
  NanoDrop One/Oneᶜ
UV-Vis
NanoDrop 8000
UV-Vis
NanoDrop Lite
UV-Vis
NanoDrop 3300
Fluorescence
NanoDrop 2000/2000c
UV-Vis
Full-spectrum microvolume UV-Vis measurements with a cuvette option in a single instrument
Higher throughput, full-spectrum microvolume UV-Vis measurements
Basic microvolume UV-Vis measurements
Full-spectrum microvolume fluorescence measurements
Full-spectrum microvolume UV-Vis measurements; cuvette option available
Measures 1-2 μL sample
Dynamic range (ng/μL dsDNA) 0.227,500
ng/μL for dsDNA
2.5–3,700
ng/μL for dsDNA
4–1,500
ng/μL for dsDNA
< 1 fmol
(FITC)
2–15,000
ng/µL for dsDNA
Full-spectral data
Pre-programmed methods for nucleic acids and proteins
Measures nucleic acid A260/A280 ratio
Measures nucleic acid A260/A230 ratio
Measures purified protein A280
Measures protein A260/A280 ratio
Pre-programmed methods for colorimetric assays (BCA, Bradford, Lowry, Pierce 660)
Measures protein and peptides at A205
Custom methods
Built-in cuvette option
Standalone local control
High-resolution touchscreen interface
Measures fluorescently-labeled nucleic acid and protein samples using absorbance
Measures a wide variety of fluorescence assays: RiboGreen™, PicoGreen™, GFP, BFP mutants, Hoechst, 4-MU, Quantum Dots, OPA, DyLite™, Alexa Fluor™, etc.      
Measures multiple fluorophores from a single sample      
Auto-Measure capability
Contaminant identification
Embedded technical support
Password protected access
Enhanced connectivity (Wi-Fi and Bluetooth™)
Thermo Fisher Connect (Cloud enabled)
Move one step closer with next-generation microvolume UV-Vis
Be more successful in downstream applications when you qualify your sample before use with the NanoDrop One UV-Vis spectrophotometer built with novel Thermo Scientific™ Acclaro™ Sample Intelligence Technology.
NanoDrop Free Trial Program
Test a NanoDrop instrument in your laboratory with your own samples and convince yourself.
Request an instrument ›
For research use only. Not for diagnostic purposes. Bluetooth is a registered trademark of Bluetooth SIG, Inc. All other trademarks are the property of Thermo Fisher Scientific and its subsidiaries. All NanoDrop instruments are approved to CE and UL/CSA.
* Patents US6628382 and US6809826.