A primary antibody is an immunoglobulin that specifically binds to a particular protein or other biomolecule of research interest for the purpose of purifying or detecting and measuring it. Primary antibodies are developed as polyclonal or monoclonal antibodies using mouse, rat, rabbit, goat, and other animal species as hosts. They are produced and supplied in various forms, ranging from crude antiserum to antigen-purified preparations. Primary antibodies for frequently researched targets are also available conjugated to fluorescent dyes or biotin.
Search for specific primary antibodies
Find antibodies of interest using the search tool below. Then filter the results by target or host species, monoclonal or polyclonal antibody type, and other criteria.
Featured primary antibodies categories
Monoclonal Antibodies
Learn about our thousands of mouse, rat, hamster, rabbit, and other species of monoclonal primary antibodies.
Polyclonal Antibodies
Learn about our thousands of rabbit, goat, sheep, chicken, and other species of monoclonal primary antibodies.
Recombinant Rabbit Monoclonal Antibodies
Learn about our Invitrogen™ ABfinity™ rabbit monoclonal antibodies, developed by immunizing animals, screening for functionality, and then cloning the genes into high-level mammalian expression vectors.
Conjugated Primary Antibodies
Discover and select primary antibodies that are directly labeled or conjugated with biotin affinity tags, fluorescent dyes (Invitrogen™ Alexa Fluor™ dyes), or reporter enzymes (HRP, AP).
Research Area Antibodies
Find and select primary antibodies for key markers and targets relevant to cancer research, stem cell research, neurobiology research, epigenetic studies, and more.
Control Antibodies
Find loading and isotype control antibodies for standardizing, normalizing and validating semi-quantitative western blotting and other immunoassay results.
Signal Pathway Antibodies
View menus and lists of primary antibodies specific to proteins of key signaling pathways, transcription pathways, and various phosphorylation or activation states.
Epitope Tag Antibodies, etc.
Find information on primary antibodies specific to fluorescent proteins, fluorescent dyes, fusion and reporter proteins, biotin, and epitope tags such as GST, His, HA, and Myc.
Cell Marker Antibodies
Select a cell marker, organelle marker, or subcellular component antibody to identify and outline cellular structures and organelles by detecting proteins that are specific to them.
Flow Cytometry Antibodies
Invitrogen eBioscience Super Bright polymer dyes join the Alexa Fluor dye and eFluor dye antibody conjugate portfolios to help answer complex cell biology questions—in less time and with less sample than other approaches.
Fluorescent antibody-drug conjugates- a new potential for antibodies
This video demonstrates the dual labeling of a primary antibody. Watch how  to create a fluorescent antibody-drug conjugate and learn about its  application, beyond the basic function as protein detection.