MA1-012_IF_NCI-H460-230X195
GTP hydrolases (GTPases) are capable of binding and cleaving GTP. The GTPase family contains sub families such as the RAS super family which is critical to cell signaling. We offer high quality antibodies against a number of GTPase targets for use in applications including western blotting, immunoprecipitation (IP), immunofluorescence (IF), immunohistochemistry (IHC), and ELISA.
Key GTPase targets include
16121-002-Arf1-Pull-Down-475px
We offer high-quality Arf1 antibodies for applications including Western blotting, IHC, IF, and IP. Arf1 is one of the ADP ribosylation factor proteins 1-6, which are members of the Ras superfamily of small GTPases. Although structurally similar, the cellular roles of Arf1-6 are different from the other Arf family members; their endogenous roles are not ADP ribosylation, but rather regulation of heterotrimeric G proteins. The Arf proteins can be divided into three classes: Class I – Arf 1-3; Class II – Arf 4,5; Class III – Arf6. Class I and II Arfs are associated with trans-Golgi network (TGN) and are involved in recruiting effector proteins to the Golgi membrane and forming vesicles. Unlike other GTPases, Arf GTPases are modified by myristoylation at the amino-terminus to allow insertion into the membrane. The Class III protein Arf6 is associated with the plasma membrane and is involved in vesicle formation at the plasma membrane, vesicle recycling and remodeling of the actin cytoskeleton.
Although the Arf GTPases are expressed in all cells, Arf1 is the most abundant, active and best-characterized. Arf1 recruits its effectors, including coatomer and clathrin adaptor complex (AP), to the Golgi for vesicle formation and budding. After recruitment, Arf1 maintains the coat proteins on the membrane until the vesicle is formed. Arf1 then interacts with its effector, resulting in the release of Arf1 from the membrane and vesicle budding. The golgi-localized gamma-ear-containing Arf (GGA) effector binding proteins contain an amino-terminal VHS domain, a GGA homology domain (GGAH), a proline-rich linker region and a carboxy-terminal gamma-ear adaptin homology domain (AGEH). The VHS domain interacts with cytoplasmic domains for receptor sorting, the GGAH domain binds activated Arf, the proline-rich region interacts with clathrin and the AGEH domain interacts with cytosolic proteins. Through mutational studies, the GGAH domain has been shown to be sufficient to bind GTP-bound Arf, and Arf1 binds all three GGA effector proteins. The association of the GGA proteins with the TGN and with the plasma membrane is regulated by the Arf. Arf 1 also functions as an allosteric activator of the cholera toxin catalytic subunit, an ADP-ribosyltransferas
16119-Cdc42-Pull-Down-250
Arf6 is one of the ADP ribosylation factor proteins 1-6, which are members of the Ras superfamily of small GTPases. Although structurally similar, the cellular roles of Arf1-6 are different from the other Arf family members; their endogenous roles are not ADP ribosylation, but rather regulation of heterotrimeric G proteins. The Arf proteins can be divided into three classes: Class I – Arf 1-3; Class II – Arf 4,5; Class III – Arf6. Class I and II Arfs are associated with trans-Golgi network (TGN) and are involved in recruiting effector proteins to the Golgi membrane and forming vesicles. Unlike other GTPases, Arf GTPases are modified by myristoylation at the amino-terminus to allow insertion into the membrane. The Class III protein Arf6 is associated with the plasma membrane and is involved in vesicle formation at the plasma membrane, vesicle recycling and remodeling of the actin cytoskeleton.
Although the Arf GTPases are expressed ubiquitously in all cells, Arf6 is less abundant and its cellular localization is cell type-dependent. Arf6 is differentiated from the other Arf proteins due to its plasma membrane association and its involvement in the regulation of receptor mediated endocytosis and remodeling of the actin cytoskeleton. Arf6 interacts with the Arf GAP effectors ACAP1, ACAP2, Git1, Git2, ASAP1 and ASAP2 for plasma membrane rearrangement. Arf6 is also involved in phospholipid metabolism. The Golgi-localized gamma-ear-containing Arf (GGA) binding proteins make up another class of effector proteins. The GGA proteins contain an amino-terminal VHS domain, a GGA homology domain (GGAH), a proline-rich linker region and a carboxy-terminal gamma-ear adaptin homology domain (AGEH). The VHS domain interacts with cytoplasmic domains for receptor sorting, the GGAH domain binds activated Arf, the proline-rich region interacts with clathrin and the AGEH domain interacts with cytosolic proteins. Through mutational studies, the GGAH domain is sufficient to bind GTP-bound Arf, and in vitro Arf6 binds GGA1 with the highest affinity. Arf6 also functions as an allosteric activator of the cholera toxin catalytic subunit, an ADP-ribosyltransferase.
Cdc42-GTPase-250
Antibodies to Cdc42 are important for GTPase research. The Cdc42 GTPase belongs to the Rho subfamily and Ras superfamily of GTPases. ThermoFisher offers quality Cdc42 antibodies for a variety of applications including Western blotting, IP, IHC, and IF. Cdc42 activation results in the polymerization of actin filaments and filopodia formation. Like other small GTPases, Cdc42 acts as a molecular switch activated by GEFs, which catalyze the exchange of bound GDP for GTP, and inhibited by GAPs, which catalyze the hydrolysis of GTP to GDP. Another level of regulation comes through the binding of Rho GDP dissociation inhibitor (RhoGDI), which retains Rho family GTPases in their inactive GDP-bound state.
Both Cdc42 and Rac1 are activated through tyrosine receptor kinase signaling leading to SAPK and p38 stress kinase pathway activation. Cdc42 is also activated by the chemoattractant fMLP in neutrophils. Fibronectin activates Cdc42 and Rac1 to induce cell spreading, and stimulation with TNF-alpha and IL-1 results in changes in the actin cytoskeleton. There is significant cross-talk between Cdc42 and Rac1, as they act in overlapping pathways, and in some cases, Cdc42 may act upstream of Rac1 during signal transduction. Some of the main effector proteins of Cdc42 are Pak1, N-WASP and IQGAP. Pak1 is a kinase involved in the activation of JNK in the SAPK stress pathway. N-WASP is an effector that induces filopodia formation, and IQGAP interacts with F-actin filaments. In differentiating neurons, Cdc42 plays an active role in neurite outgrowth. However, the Rho family of GTPases can work agonistically during cell signaling and antagonistically during differentiation.
http-www.piercenet.commedia16118-002-Rac1-GTPase-250
Rac1 Antibodies are important in GTPase research. The Rac1 GTPase is part of the Rho subfamily and Ras superfamily of GTPases. Thermo Fisher offers robust Rac1 antibodies for a variety of applications including IHC, IF, Western blotting and IP. Activation of Rac1 results in actin polymerization and appears as membrane ruffling at the cellular periphery. Rac1 activation also results in lamelipodia formation. The Rac1 GTPase transduces signals through tyrosine kinases, adhesion molecules or cytokine/chemokine receptors after stimulation with growth factors (EGF, insulin, PDGF, NGF), integrins (fibronectin) or chemoattractants (fMLP). For example, stimulation with EGF results in PI3 kinase activation, resulting in cell growth and reorganization at the cell periphery (membrane ruffling).
Alternatively, tyrosine receptor kinase signaling through Rac1 leads to activation of the MAPK stress response pathways SAPK (JNK) and p38. Stimulation of cells with fibronectin results in integrin-mediated cell spreading. Two of the main effector proteins of Rac1 are Pak1 and phosphoinositol 4-phosphate 5-kinase. Pak1 (p65 Pak) is a kinase that activates the JNK pathway, while phosphoinositol 4-phosphate 5-kinase promotes actin filament assembly. Rac is critical for T-cell development and for promoting differentiating cells. However, the Rho family of GTPases can work agonistically during cell signaling and and antagonistically during differentiation.
Rap1-GTPase-250
The Rap GTPases are part of the Ras superfamily of GTPases and are encoded by Rap1a, Rap1b and Rap2. Thermo Fisher offers many Rap1 antibodies for a number of applications including Western blotting.  Rap GTPases are structurally similar to Ras GTPases and have similar effector and activator proteins, although Rap GTPases have different functional activities than Ras. While Ras is involved in cell proliferation and survival, Rap1 regulates cytoskeletal rearrangements, cell adhesion and cell junction formation.
The specificity of Rap1 and Ras is mediated by their respective upstream regulators and downstream effectors. The GEFs for Rap contain a CDC25 homology domain that mediates the GDP/GTP exchange reaction and a REM (Ras exchange motif) domain. Some Rap GEFs include C3G, Epac1 and 2, RasGRP2, PDZ-GEF1 and 2 and PLC epsilon. The binding domain used in the active Rap1 Pull-Down and Detection kit is RalGDS, a GEF that contains an RA domain to which Rap1 has a higher binding affinity than Ras. The RapGAPs, including Rap1GAP and the Spa-1 family, insert an asparagine side chain into the nucleotide-binding pocket to catalyze the GTP hydrolysis reaction. Rap effector proteins, including RAPL, Riam, AF-6, Krit1, RacGEFs, Tiam1, Vav2, Rho GAPs and ARAP3, are involved in cell-cell junctions and adhesion and are often localized to the membrane or at cell-cell junctions. Besides Ras, there is also cross-talk between Rap and Rho GTPases.
ras-250px
The Ras subfamily of GTPases is part of the Ras superfamily of GTPases, named after the rat sarcoma viral oncogene. Thermo Fisher offers high-quality Ras antibodies for many applications including Western blotting, IHC, IF, and IP. The Ras subfamily contains many Ras isoforms, including K-Ras, H-Ras, and N-Ras. While the three isoforms are expressed at different levels in different types of cells, in general, activating mutations of at least one of these isoforms are present in approximately 15 percent of all cancers. The Ras proteins serve as initiators of intracellular signal transduction from extracellular molecules and associate with the plasma membrane via lipid modification and prenylation of its carboxy terminus. These modifications at the carboxy terminus determine the localization of Ras to distinct membrane microdomains, which dictates subsequent downstream signaling. Ras signaling pathways affect many cellular processes including proliferation, survival, vesicular trafficking and gene expression.
Signaling through Ras is central to many cellular responses, and activation of Ras is regulated by several GEF and GAP proteins. The GEF proteins mediate GTPase activation by dissociating GDP from the inactive Ras to allow binding of Ras to GTP. Conversely, GAP proteins inactivate GTPases by hydrolyzing GTP to GDP. Ras mediates downstream signaling by interacting with effector proteins including Raf and PI3 kinase. Raf1 is a serine/threonine protein kinase that is part of the MAP kinase kinase signaling pathway that leads to the activation of ERK and p38, which influences proliferation and survival. PI3 kinase signaling results in activation of AKT and mTOR, which are central for cell growth and survival. Ras is also integral for cellular differentiation and development, including immune cell development and function. Mutations in the Ras genes (H-Ras, N-Ras, and K-Ras) are found in a variety of cancers, with the highest incidences in lung adenocarcinoma, mucinous adenoma, ductal carcinoma of the pancreas, colorectal carcinoma, thyroid tumors, and myeloid leukemia.
bulk-pod-showcase-270x195
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
Please click on the button below to fill out our bulk and custom ordering form. 
Complete request form
Resources
Product selection guides
Learning and support
Cell analysis resources