V5 Tag Antibodies, MA5-15253-D680
The V5 tag is derived from a small epitope (Pk) found on the P and V proteins of the paramyxovirus of simian virus 5 (SV5). The V5 tag is generally used with all 14 amino acids (GKPIPNPLLGLDST), but it may also be used with a shorter 9-amino acid (IPNPLLGLD) sequence.
V5 tag antibodies provide a dependable method for the detection and purification of tagged target proteins without a protein-specific antibody or probe. Invitrogen V5 tag antibodies are designed to reliably detect recombinant V5-tagged proteins, and each antibody is validated for use in many applications.
See all V5 tag antibodies
Featured product data
Immunofluorescence analysis of V5 tag was done on HEK-293 cells
Immunofluorescence analysis of V5 tag was done on HEK-293 cells transiently overexpressing V5-His -LacZ. The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with V5 Tag Mouse Monoclonal Antibody (Cat. No. 377500) at a dilution of 1:500 in 1% BSA and incubated for 3 hours at room temperature and then labeled with Alexa Fluor 488 Rabbit Anti-Mouse IgG Secondary Antibody (Cat. No. A11059) at a dilution of 1:400 for 30 minutes at room temperature (Panel a: green). Nuclei (Panel b: blue) were stained with SlowFadeGold Antifade Mountant with DAPI (S36938). F-actin (Panel c: red) was stained with Alexa Fluor 594 Phalloidin (Cat. No. A12381). Panel d is a merged image showing cytoplasmic localization. Panel e is untransfected HEK-293 cells.
Flow cytometry analysis of V5 tag
Flow cytometry analysis of V5 tag was done on HEK-293 cells transiently overexpressing V5-His-LacZ. Cells were fixed with 70% ethanol for 10 minutes, permeabilized with 0.25% Triton™ X-100 for 20 minutes, and blocked with 5% BSA for 1 hour at room temperature. Cells were labeled with V5 Tag Mouse Monoclonal Antibody (Cat. No. 377500, red histogram) or with mouse isotype control (pink histogram) at a dilution of 1:250 in 2.5% BSA. After incubation at room temperature for 3 hours, the cells were labeled with Alexa Fluor 488 Rabbit Anti-Mouse Secondary Antibody (Cat. No. A11059) at a dilution of 1:400 for 30 minutes at room temperature. The representative 10,000 cells were acquired and analyzed for each sample using an Attune Acoustic Focusing Cytometer. The purple histogram represents unstained control cells and the green histogram represents no-primary-antibody control. 
Western blot analysis of V5. Analysis
Western blot analysis of V5. Analysis was performed by loading 25 µg of 293T cells transfected with 12Tag onto an SDS polyacrylamide gel. Proteins were transferred to a PVDF membrane and blocked at 4°C overnight. The membrane was probed with a V5 epitope tag polyclonal antibody (Cat. No. PA1-993) at a dilution of 1:2,000 overnight at 4°C, washed in TBST, and probed with an HRP-conjugated secondary antibody for 1 hour at room temperature in the dark. Chemiluminescent detection was performed using Thermo Scientific Pierce ECL Plus Western Blotting Substrate (Cat. No. 32132). Results show a band at ~45 kDa.
Annotated product references
Cat. No. MA5-15253 was used in western blot to identify the genetic basis of congenital sodium diarrhea Heinz-Erian P, Müller T, Krabichler B et al. (2009) Mutations in SPINT2 cause a syndromic form of congenital sodium diarrhea. Am J Hum Genet 84:188–196.
Resources
Product selection guides
Learning and support
Cell analysis resources