1413927588139
Our anti-dog secondary antibodies are affinity-purified polyclonal and monoclonal antibodies with well-characterized specificity for mouse immunoglobulin classes. They are available in host species including goat, mouse and sheep, and in the following formats:
  • Unlabeled antibodies
  • Enzyme conjugates (alkaline phosphatase (AP) or horseradish peroxidase (HRP))
  • Fluorescent conjugates (Fluorescein (FITC), Tetramethylrhodamine (TRITC), etc.)
  • Biotin
View all anti-dog secondaries
Our secondary antibodies are available as whole IgG. Each of these antibodies is affinity purified using its target antigen (the primary antibody class) coupled to an agarose support. This affinity purification eliminates nonspecific antibodies, resulting in high specificity and low background signal. Conjugated antibodies are affinity purified before labeling.
How to find a secondary antibody
Step 1: Search for the target species of interest (e.g., “anti-mouse” or “goat anti-rabbit” or “alexa fluor 488”).
Step 2: Narrow results by host species, conjugate, application, and other criteria using side filters.
bulk-pod-showcase-270x195
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
Please click on the button below to fill out our bulk and custom ordering form. 
Complete request form
Resources
Product selection guides
Learning and support