anti-sheep-showcase
Our anti-sheep secondary antibodies are affinity-purified polyclonal or monoclonal antibodies with well-characterized specificity for chicken immunoglobulin classes. They are available in host species including donkey, rabbit, and mouse, and in the following formats:
  • Unlabeled antibodies
  • Enzyme conjugates (alkaline phosphatase (AP) or horseradish peroxidase (HRP))
  • Fluorescent conjugates (Alexa Fluor, DyLight, Fluorescein (FITC), etc.)
  • Biotin
View all anti-sheep secondaries
Our secondary antibodies are available as whole IgG or the F(ab')2 fragment. Each of these antibodies is affinity purified using its target antigen (the primary antibody class and fragment) coupled to an agarose support. This affinity purification eliminates nonspecific antibodies, resulting in high specificity and low background signal. Conjugated antibodies are affinity purified before labeling.
Selected secondary antibodies have been further purified by adsorbing the antigen-purified antibody to an affinity column containing immobilized serum proteins or IgG of selected species. This additional processing minimizes antibody cross-reactivity to the serum proteins of those selected species. These antibodies are designated as Cross Adsorbed or Highly Cross Adsorbed, based on the amount of processing which has occurred.
How to find a secondary antibody
Step 1: Search for the target species of interest (e.g., “anti-mouse” or “goat anti-rabbit” or “alexa fluor 488”).
Step 2: Narrow results by host species, conjugate, application, and other criteria using side filters.
bulk-pod-showcase-270x195
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
Please click on the button below to fill out our bulk and custom ordering form. 
Complete request form
Resources
Product selection guides
Learning and support