expect-more-liq
For over 50 years, production facilities have turned to our trusted Gibco media, feeds, buffers, and bioprocessing liquids to enhance product performance. Our products are designed to meet your specifications including flexible BioProcess Containers (BPCs) in the size of your choice from 1 L to 1,000 L. By combining our standard and custom products with our worldwide supply chain distribution centers we can offer you superior solutions for your process with the continuity of having one consistent supply partner.
If you want more information or have questions regarding our catalog or custom offerings for bulk liquids and/or bioprocess containers (BPCs), then please complete this short form and a BioProduction Account Manager or Field Application Specialist will contact you shortly:
Contact us                               View the newest facility video
Overview of our bioprocessing liquid capabilities for custom
and catalog products
  Catalog Custom
Validated shipping configurations from bottles to bioprocess containers PET BOXY bottles: 100 mL, 500 mL, 1,000 mL 50 mL to 1,000 L
  Bioprocess containers (BPCs): 1 L, 5 L, 10 L, 20 L, 50 L, 100 L, 200 L, 500 L, 1,000 L  
Catalog formulation in
the film of your choice
Thermo Scientific  Aegis5-14 film Aegis5-14 film
  Thermo Scientific 
CX5-14 film
CX5-14 film
  ASI 26/77 film ASI 26/77 film
Batch/lot sizes Stainless steel tank sizes of 200 L to 10,000 L Best-in-class liquid formulation with capacity and processes to handle small- and large-volume liquids
  Portable Nalgene  tanks 10–400 L Assurance of supply via two-continent redundant manufacturing
  AOF 5,000L and 10,000 L tanks and suites A range of liquid formats to meet customer needs (concentrates, 1x liquids, bottles and bags)
  One formulation per room  
  Validated CIP/COP processes  
Gibco media, feeds, and supplements CD OptiCHO  
  Dynamis  
  CD CHO  
  EfficientFeed A+/B+/C+  
  FunctionMAX Titer Enhancer  
Bioprocess buffer Water for Injection (WFI)  
Media, feeds, and supplements   Your custom formulation or Gibco brand solutions
Bioprocess buffers   Your custom formulation for upstream (PBS, glucose, HBSS) and downstream buffers (NaOH, HCl, titration, equilibration, binding, elution buffers) as well as cleaning and sanitizing process liquids (WFI, EDTA, HCl)
Value-added customer service   Dedicated Client Project Managers will oversee each custom project
High standard quality processes   Detailed documentation
Unique global footprint with our manufacturing
and warehousing capabilities
With continual investment in multiple global plant expansions, Thermo Fisher Scientific aims to provide seamless customer experiences. Since 2011, we have invested over $38M (USD) to enhance supply security and risk mitigation, offering you peace of mind.
Select any of our integrated global locations and receive long-term supply with minimal production interruptions. 
Grand Island, New York
2015–2016 = $18.6M investment
  • Expansion: increase manufacturing footprint
    and add tank capacity (5M liters)
  • GMP 21 CFR 820
  • ISO 13485 additional finished good warehouse space
  • Ability to manage hazardous solutions
Paisley, Scotland 
2014–2015 = $20M investment
  • Expansion: 10,000 L AOF tank
  • GMP 21 CFR 820
  • Built to class 100,000 standard (ISO 8)
  • Water for Injection (WFI) systems—same quality of water
Harmonization of our manufacturing facilities in the US and Europe is in progress.
  • Raw materials—expanding our multi-compendial library for common chemical raw materials and standard bag offerings
  • Equipment—continually expanding our liquid capabilities for both AO and AOF requests
  • Finished product testing—USP/EP testing available; sterility, endotoxin, conductivity, osmolality, pH, etc.
Access warehousing and logistical services in addition to quality manufacturing. Whether your needs are short-term or long-term, we have storage to suit your production and distribution requirements. Manage your shipping by choosing from temperature-controlled, cold, and ambient-temperature warehouses located all over the globe.
global-supply-chart
Take control of your product delivery
Thermo Fisher Scientific’s supplier qualification program is designed to provide customers with more control over product delivery. We’ve implemented a robust supply risk mitigation strategy to help ensure that our suppliers meet the rigorous standards required to avoid stock-outs and shortages.
Thermo Fisher Scientific enables you to focus on delivering biotherapeutics and vaccines without worrying about costly production interruptions. With an expanded global reach and dual sourcing of raw materials, we’re able to leverage economies of scale and access high-quality resources at the best prices—so you can expect more savings on more options.   
To meet current and future customer requirements for insourcing large-volume liquids or bulk liquids, our Grand Island manufacturing facility will adhere to the following standards, supporting consistent performance for each batch:
  • ISO 13485
  • 210/211 CFR related to sterile filtration
  • Annex 1 standard
  • Animal Origin Free (AOF) 
  • Controlled spaces
  • Grade C & Grade D
  • Single material flow
  • Proximity to Finished Good warehouse
  • Proximity to Raw Materials warehouse
  • 2nd Floor formulation
Our robust supplier management program helps ensure sourcing from qualified and audited raw material suppliers. We have qualified multiple raw-material sources for risk mitigation and contingency planning purposes. We also offer Animal Origin Free (AOF) manufacturing segregation with single-use liquid management systems. These single-use advantages alleviate cleaning, validation, and cross-contamination concerns.
Custom media, buffers, and process liquid formulations
and their packaging
We offer expanded expertise in BPC film for bulk liquids. Get multiple proprietary BPC films in customizable types and sizes—only from Thermo Fisher Scientific. 
  • CX5-14 and Aegis5-14 films are validated for liquid fills and shipments from Grand Island and Paisley
  • Extractables and leachables data package available for each film
  • Stability data in progress for common media and buffer liquid in CX5-14 and Aegis films 
  • Customer-specified stability available as a fee for service
  • Continued supply of liquids in existing orders in 9101 and BF1400 BPCs
Liquid container options can be customized by size and configuration. Our Open Architecture component strategy features a library of more than 1,000 qualified components. This allows you to optimize several aspects of the BPC, including support container, chamber size and porting, tubing size and style, and fittings and end treatments. 
Global quality system and technical services
Get the support you need with Thermo Fisher Scientific’s Global Quality System and Gibco BioProduction Services. Our experienced professionals are ready to help you take control of any product development or process optimization challenges you may have. 
Stability testing
  • State-of-the-art analytical laboratory supports your stability testing requirements
  • Stability Data Sheet provides a summary of stability test data for model formulations 
Technical support 
  • Extractables and leachables data package available for each type of film, with customized testing available upon request
  • Packaging engineers to optimize packaging and shipping configurations for enhanced pre- and post-shipment product integrity 
Overview of our bioprocessing liquid capabilities for custom
and catalog products
  Catalog Custom
Validated shipping configurations from bottles to bioprocess containers PET BOXY bottles: 100 mL, 500 mL, 1,000 mL 50 mL to 1,000 L
  Bioprocess containers (BPCs): 1 L, 5 L, 10 L, 20 L, 50 L, 100 L, 200 L, 500 L, 1,000 L  
Catalog formulation in
the film of your choice
Thermo Scientific  Aegis5-14 film Aegis5-14 film
  Thermo Scientific 
CX5-14 film
CX5-14 film
  ASI 26/77 film ASI 26/77 film
Batch/lot sizes Stainless steel tank sizes of 200 L to 10,000 L Best-in-class liquid formulation with capacity and processes to handle small- and large-volume liquids
  Portable Nalgene  tanks 10–400 L Assurance of supply via two-continent redundant manufacturing
  AOF 5,000L and 10,000 L tanks and suites A range of liquid formats to meet customer needs (concentrates, 1x liquids, bottles and bags)
  One formulation per room  
  Validated CIP/COP processes  
Gibco media, feeds, and supplements CD OptiCHO  
  Dynamis  
  CD CHO  
  EfficientFeed A+/B+/C+  
  FunctionMAX Titer Enhancer  
Bioprocess buffer Water for Injection (WFI)  
Media, feeds, and supplements   Your custom formulation or Gibco brand solutions
Bioprocess buffers   Your custom formulation for upstream (PBS, glucose, HBSS) and downstream buffers (NaOH, HCl, titration, equilibration, binding, elution buffers) as well as cleaning and sanitizing process liquids (WFI, EDTA, HCl)
Value-added customer service   Dedicated Client Project Managers will oversee each custom project
High standard quality processes   Detailed documentation
Unique global footprint with our manufacturing
and warehousing capabilities
With continual investment in multiple global plant expansions, Thermo Fisher Scientific aims to provide seamless customer experiences. Since 2011, we have invested over $38M (USD) to enhance supply security and risk mitigation, offering you peace of mind.
Select any of our integrated global locations and receive long-term supply with minimal production interruptions. 
Grand Island, New York
2015–2016 = $18.6M investment
  • Expansion: increase manufacturing footprint
    and add tank capacity (5M liters)
  • GMP 21 CFR 820
  • ISO 13485 additional finished good warehouse space
  • Ability to manage hazardous solutions
Paisley, Scotland 
2014–2015 = $20M investment
  • Expansion: 10,000 L AOF tank
  • GMP 21 CFR 820
  • Built to class 100,000 standard (ISO 8)
  • Water for Injection (WFI) systems—same quality of water
Harmonization of our manufacturing facilities in the US and Europe is in progress.
  • Raw materials—expanding our multi-compendial library for common chemical raw materials and standard bag offerings
  • Equipment—continually expanding our liquid capabilities for both AO and AOF requests
  • Finished product testing—USP/EP testing available; sterility, endotoxin, conductivity, osmolality, pH, etc.
Access warehousing and logistical services in addition to quality manufacturing. Whether your needs are short-term or long-term, we have storage to suit your production and distribution requirements. Manage your shipping by choosing from temperature-controlled, cold, and ambient-temperature warehouses located all over the globe.
global-supply-chart
Take control of your product delivery
Thermo Fisher Scientific’s supplier qualification program is designed to provide customers with more control over product delivery. We’ve implemented a robust supply risk mitigation strategy to help ensure that our suppliers meet the rigorous standards required to avoid stock-outs and shortages.
Thermo Fisher Scientific enables you to focus on delivering biotherapeutics and vaccines without worrying about costly production interruptions. With an expanded global reach and dual sourcing of raw materials, we’re able to leverage economies of scale and access high-quality resources at the best prices—so you can expect more savings on more options.   
To meet current and future customer requirements for insourcing large-volume liquids or bulk liquids, our Grand Island manufacturing facility will adhere to the following standards, supporting consistent performance for each batch:
  • ISO 13485
  • 210/211 CFR related to sterile filtration
  • Annex 1 standard
  • Animal Origin Free (AOF) 
  • Controlled spaces
  • Grade C & Grade D
  • Single material flow
  • Proximity to Finished Good warehouse
  • Proximity to Raw Materials warehouse
  • 2nd Floor formulation
Our robust supplier management program helps ensure sourcing from qualified and audited raw material suppliers. We have qualified multiple raw-material sources for risk mitigation and contingency planning purposes. We also offer Animal Origin Free (AOF) manufacturing segregation with single-use liquid management systems. These single-use advantages alleviate cleaning, validation, and cross-contamination concerns.
Custom media, buffers, and process liquid formulations
and their packaging
We offer expanded expertise in BPC film for bulk liquids. Get multiple proprietary BPC films in customizable types and sizes—only from Thermo Fisher Scientific. 
  • CX5-14 and Aegis5-14 films are validated for liquid fills and shipments from Grand Island and Paisley
  • Extractables and leachables data package available for each film
  • Stability data in progress for common media and buffer liquid in CX5-14 and Aegis films 
  • Customer-specified stability available as a fee for service
  • Continued supply of liquids in existing orders in 9101 and BF1400 BPCs
Liquid container options can be customized by size and configuration. Our Open Architecture component strategy features a library of more than 1,000 qualified components. This allows you to optimize several aspects of the BPC, including support container, chamber size and porting, tubing size and style, and fittings and end treatments. 
Global quality system and technical services
Get the support you need with Thermo Fisher Scientific’s Global Quality System and Gibco BioProduction Services. Our experienced professionals are ready to help you take control of any product development or process optimization challenges you may have. 
Stability testing
  • State-of-the-art analytical laboratory supports your stability testing requirements
  • Stability Data Sheet provides a summary of stability test data for model formulations 
Technical support 
  • Extractables and leachables data package available for each type of film, with customized testing available upon request
  • Packaging engineers to optimize packaging and shipping configurations for enhanced pre- and post-shipment product integrity 
Custom Media Request
Custom MR Form
Download your media request form.
Easy Reordering
Quick Order Page
Use the Quick Order page for reorders of the same quantity and specifications.
Contact Customer Care
Customer Care Page
3175 Staley Road
Grand Island, NY 14072
USA
Phone: 800.955.6288
Fax: 800.331.2286
Email: customerservice@lifetech.com
BioProduction resources
Learn more about our custom liquid products for downstream bioprocessing 
Download your copy ›
Learn more about our offerings from media, feeds, buffers, and bioprocessing liquids to bioprocessing
containers (BPCs) 
Download your copy ›