AAV
POROS CaptureSelect AAV resins are the first affinity chromatography resins for the purification of viral vectors that are in use for gene therapy applications. The resins are specifically intended for the purification of adeno-associated virus (AAV) particles of different subtypes.
The unique CaptureSelect ligand technology is combined with the large pore structure of POROS resins to enable the development of large biomolecules such as viral vectors and vaccines. The POROS CaptureSelect AAV affinity resins are designed to significantly improve downstream processing by reducing the number of purification steps and offering scalability, thus maximizing processing consistency and productivity.
Benefit from:
  • High selectivity and capacity in the purification of viral vectors
  • A fast, robust, high yield and purity, from crude material in one step, purification process, offering scalability and processing consistency
  • Significant improvement to downstream processing, by reducing the purification steps and maximizing productivity
POROS CaptureSelect resins and reagents for viral vector purification
The large pore structure of POROS CaptureSelect AAV affinity resins makes them suited for purifying large, complex AAV vectors, supporting chromatographic separations at considerably faster flow rates than conventional liquid chromatography separations while maintaining high dynamic binding capacity. In addition, the 50 μm particle size provides superior resolution for excellent single-step impurity clearance that is independent of scale and flow rate. These affinity resins address the high-selectivity and high-capacity requirements for purifying viral vectors, exhibiting capacities of >1014 viral genomes per milliliter (vg)/mL (Table 1).
  Binding Capacity (vg/mL) Serotype Affinity
POROS CaptureSelect AAV8 >1013 AAV8
POROS CaptureSelect AAV9 >1014 AAV9
POROS CaptureSelect AAVX >1013 AAV1, AAV2, AAV3, AAV4, AAV5. AAV6, AAV7, AAV8, AAV9, AAVrh10**
Table 1. Summary of POROS CaptureSelect AAV affinity resin properties
* Binding capacity will vary based on serotype, feed stream, additives, and mutations to parent serotypes
** The AAVX Affinity Matrix has demonstrated binding reactivity towards a large set of AAV serotypes (including chimeric vectors), suggesting that an even broader range of AAV serotypes might be purified by this resin.
POROS CaptureSelect resins and reagents for purification of viral vectors
Additional reading and webinars
Application note
  POROS CaptureSelect AAV Affinity Resins: Enabling technologies for efficient downstream processing of AAV viral vectors
Article
Innovative Downstream Purification Solutions for Viral Vectors: Enabling Platform Approaches to Advance Gene Therapies
Webinar
1. Terova O, Parra S, Clasen R et al. (2016) Innovative Downstream Purification Solutions for Viral Vectors: Enabling Platform Approaches to Advance Gene Therapies. BioProcess†International.
Resources
Bioproduction resources- Have questions about Gibco media, POROS chromatography resins, Applied Biosystems SEQ rapid contaminant tests, or cell therapy products? Click below to access product brochures, training videos, mobile apps, and more.
  • NEW Bioproduction Learning Center
  • Bioproduction resources
  • Tools by product type
  • Tools by media type
  • Support
    For research use or further manufacturing, not for diagnostic use or direct administration in humans or animals.