Large-volume rigid plastic support containers
Our portfolio of plastic outer support containers include rigid plastic vessels to support our BioProcess Container (BPC) products to help meet your large-volume liquid storage, handling, and shipping needs. These containers also give you the flexibility to add custom-configured options without extensive lead times or associated costs.
Request information  Download product brochure  
ALLpaQ rectangular support containers
ALLpaQ rectangular support containers are qualified for storage and shipment of sterile process liquids in bioprocess containers.
Key features:
  • Available in 50, 100, 200, 250, 500, 600, and 1,000 L
  • Cleanable—Polypropylene construction with smooth, easy-to-clean surfaces
  • Reusable—Robust design and sturdy construction allows for multiple uses
  • Stackable—easy storage both empty and full
  • Easy-to-use—bottom drain options are available
  • Validated systems for transporting sterile process liquids in volumes up to 1,000 L
Applications:
  • Media and buffer storage and transport within a facility
  • Feeding media into bioreactors and buffers into purification systems
  • Receiving and storing of harvest from bioreactors
  • Shipping bioprocess liquids to another site when proper packing and dunnage is utilized
Accent plastic outer container
The Accent plastic outer container systems are designed for in-process fluid handling and storage. 
Key features:
  • Standard sizes: 100 L, 200 L, 500 L, and 1,000 L.
  • Wide configuration option available in 1,000 L
  • Easy-of-use and operation
  • Clean room grade swivel casters (available up to 500 L)
  • Smooth, clean room grade plastic
  • Integrated handles for ease of use
  • Custom-configured options: container lid, filter rack, and pump shelving
Applications:
  • Buffers and media storage and distribution
  • Bulk feed and harvest
  • Bulk intermediate hold
  • Bulk drug substance
  • Final product transport
ALLpaQ rectangular support containers
ALLpaQ rectangular support containers are qualified for storage and shipment of sterile process liquids in bioprocess containers.
Key features:
  • Available in 50, 100, 200, 250, 500, 600, and 1,000 L
  • Cleanable—Polypropylene construction with smooth, easy-to-clean surfaces
  • Reusable—Robust design and sturdy construction allows for multiple uses
  • Stackable—easy storage both empty and full
  • Easy-to-use—bottom drain options are available
  • Validated systems for transporting sterile process liquids in volumes up to 1,000 L
Applications:
  • Media and buffer storage and transport within a facility
  • Feeding media into bioreactors and buffers into purification systems
  • Receiving and storing of harvest from bioreactors
  • Shipping bioprocess liquids to another site when proper packing and dunnage is utilized
Accent plastic outer container
The Accent plastic outer container systems are designed for in-process fluid handling and storage. 
Key features:
  • Standard sizes: 100 L, 200 L, 500 L, and 1,000 L.
  • Wide configuration option available in 1,000 L
  • Easy-of-use and operation
  • Clean room grade swivel casters (available up to 500 L)
  • Smooth, clean room grade plastic
  • Integrated handles for ease of use
  • Custom-configured options: container lid, filter rack, and pump shelving
Applications:
  • Buffers and media storage and distribution
  • Bulk feed and harvest
  • Bulk intermediate hold
  • Bulk drug substance
  • Final product transport
Related products
Available in standard or custom configurations in a wide range of sizes for a number of applications
Bring your bioproduction processes together seamlessly with an extensive range of standard and customized fluid transfer products
Ordering information
Resources
Storage and transport solutions
BioProduction resources
Click below to access products brochures, training videos, mobile apps, and more.
Support
 Request more info on BioProduction