Welcome to the EVOS® FL Auto Cell Imaging System image and video gallery.  All images and videos here were generated on the EVOS® FL Auto Cell Imaging System using a variety of vessel types.
Learn more about EVOS® FL Auto
EVOS® FL Auto Videos
HeLa 
Reagents: CellLight® H2B GFP and  transmitted light
Objective: 20x Objective
Time lapse: 14 hours with images taken every 5 minutes
HeLa treated with LatrunculinA
Reagents: CellLight® Actin GFP, MitoTracker® Red, NucBlue® Live
Time Lapse: 14 hours with images taken every 5 minutes
BPAEC treated with Menadione
Reagents: CellRox® Green and Transmitted Light
Time Lapse: 2 hours with images taken every 5 minutes
USOS treated with Staurosporine
Reagents: LIVE/DEAD® Cell Imaging Kit
Time Lapse: 14 hours with images taken every 5 minutes
USOS treated with Staurosporine
Reagents: CellEvent® Green
Time Lapse: 14 hours with images taken every 5 minutes
HeLa
Reagents: CellLight® H2B GFP, CellLight® Mito RFP
Time Lapse: 24 hours with images taken every 12 minutes
Wound Healing in HDFn cells 
Reagents: CellTracker™ Red CMTPX
Time Lapse: 70 hours with images taken every 2 hours
Cell Migration during angiogenesis 
Reagents: CellTracker™ Red CMTPX
Time Lapse: 40 hours with images taken every 2 hours
EVOS® FL Auto Images