HCS Oxidative Stress Assay
Optimized, automated assays for HCS
Generation of reactive oxygen species (ROS) is inevitable for aerobic organisms and, in healthy cells, occurs at a controlled rate. Increased production of ROS by a cell is an early hallmark of cellular stress. Intracellular ROS cause damage to proteins, nucleic acids, and membrane lipids. Oxidative damage of these biomolecules is associated with aging as well as a variety of pathological events, including atherosclerosis, carcinogenesis, ischemia reperfusion injury, and neurodegenerative disorders. Using fluorogenic reporters such as CellROX dyes allows for the real-time analysis of ROS generation in living cells and in fixed-cell preparations.
Request a demo  Access HCA assay builder  Request a poster
Typical oxidative stress assay
Live or fixed-cell assay protocol.
Imaging mode
  • Widefield or confocal
  • 10x or 20x magnification
Automatically measured properties
  • Fluorescence intensity, morphology, and count values for each object
  • Fluorescence intensity, morphology, and count values of spots in different cell regions
  • Average fluorescence intensity difference and ratios between different regions for each cell
  • Intensity and count ratio between channels within different regions for each cell
Oxidative stress assay images
HCS oxidate stress assay
Hap1 cells were treated with menadione and labeled with CellROX Green and Hoechst 33342. The cells were imaged using a Thermo Scientific CellInsight CX5 high-content platform (row A), and Thermo Scientific HCS Studio Software, which were used to automatically identify (row B) and quantify the intensity of CellROX green fluorescence from each cell.
Sample experimental data
HCS Oxidative Stress Assay sample data
Dose-dependent increase in ROS production in Hap 1 cells loaded with CellROX Green and treated with menadione for one hour.
Resources
Instrument systems
Learn more
HCS reagents optimized for use with your HCS platform