ViewRNA Cell Plus Assay
Simultaneously visualize RNA, with single-molecule sensitivity, and protein in single cells
The complex interactions between transcription, translation, and post-translational modifications are hidden from view in typical endpoint assays that measure either RNA or protein levels. While in situ hybridization (ISH) provides a method for examining the levels of specific RNA transcripts in individual cells, and immunocytochemistry (ICC) utilizes antibodies to visualize the localization of specific proteins (and protein modifications), the ability to simultaneously observe RNA and protein in a single cell has been thwarted by the incompatibility of ICC and ISH protocols.
Further complicating this analysis, traditional ISH methods lack sensitivity for detecting low-abundance RNA species and have limited multiplexing capability that further constrain its compatibility with other cell assays.
Invitrogen ViewRNA Cell Plus Assay is a novel method that combines ICC with the Invitrogen ViewRNA technology, a proprietary fluorescence ISH (FISH) and sequential branched-DNA (bDNA) amplification technique, to visualize RNA with single-molecule sensitivity along with protein in individual cells. This assay enables simultaneous detection of up to three RNA targets in combination with immunophenotyping for cell surface and intracellular proteins using both indirect and direct ICC, which allows for in-depth characterization of specific cell subpopulations. The combined visualization of RNA and protein expression at the single-cell level is a valuable tool that has broad implications for advancing research across multiple fields of biology.
Technology overview
FISH is a powerful technique that enables the specific detection of target RNA localization in fixed cells. Traditional FISH techniques using large oligonucleotide sequences labeled with one to five fluorophores are generally limited by high background and low sensitivity due to non-specific binding and insufficient signal amplification.
ViewRNA Cell Assays incorporate a proprietary probe set design and bDNA signal amplification technology. A target-specific Probe Set of approximately 20 oligonucleotide pairs hybridizes to the target RNA of interest. Signal amplification is achieved through specific hybridization of adjacent oligonucleotide pairs to bDNA structures, which are formed by Pre-amplifiers, Amplifiers, and fluorochrome-conjugated Label Probes. This methodology results in greater specificity, lower background, and higher signal-to-noise ratios. ViewRNA Cell Assays are the most sensitive and specific RNA ISH methods on the market.
In comparison, to create images with similar discernible spots, traditional FISH techniques can require a 600-times longer exposure and a 100-times greater camera gain than ViewRNA ISH Cell Assays. Furthermore, under equivalent imaging conditions, ViewRNA ISH Cell Assay is 100-times brighter, creating a minimum two- to three-times higher signal-to-noise ratio (Nat Methods 10:1127 (2013) ).
Search for available (in stock) probe sets
Assay kits and supporting products
The ViewRNA Cell Plus Assay contains all the reagents needed to conduct the assay. Positive control probes and target probes for genes of interest are sold separately.
Antibody compatibility
To date, we have evaluated the performance of more than 100 ICC-compatible antibodies (comprised of a mix of unconjugated/purified, biotinylated, and fluorophore-conjugated format; see table below) for use in ViewRNA Cell Plus Assay. To accomplish this, antibody performance was tested either in the context of the full assay (antibody staining and RNA detection) or by using Invitrogen ViewRNA Cell Plus Fixation/Permeabilization Buffer Set (Cat. No. 00-19001) according to the antibody staining portion of the protocol. We found that 95% of the antibodies tested that perform optimally with formaldehyde-fixed cells also perform well with the ViewRNA Cell Plus Assay. Antibodies that require methanol fixation were generally found to be unsuitable for this assay.
In this evaluation, we have assessed a broad panel of antibodies with specificities against structural proteins, transcription factors, organelles, and surface markers. Our results show that signal intensity and resolution of detail is maintained with ViewRNA Cell Plus Assay compared to our standard immunocytochemistry protocol. Based on our studies, we conclude that antibodies which are suitable for ICC or immunohistochemistry (IHC) (i.e., optimized for formaldehyde fixation) are likely to be compatible with ViewRNA Cell Plus Assay. Nevertheless, we recommend using ViewRNA Cell Plus Fixation/Permeabilization Buffer Set (Cat. No. 00-19001) to establish acceptable performance criteria prior to running the complete ViewRNA Cell Plus Assay.
Species
Specificity
Clone
Cat. No.
Conjugate
Concentration tested
Recommended
Human, Mouse, Rat, Canine, Bovine, Guinea Pig, Pig, Horse
Actin
HHF35
14-6496
Purified
10 µg/mL
Human, Mouse, Rat, Canine, Bovine, Guinea Pig, Pig, Horse
Actin
HHF35
53-6496
Invitrogen Alexa Fluor 488
30 µg/mL
Human
AFP (alpha fetoprotein)
1E8
14-9499
Purified
5 µg/mL
Human
AHR
FF3399
14-9854
Purified
20 µg/mL
Human
ALDH1L1
7G8
14-9595
Purified
1 µg/mL
Human
alpha-Catenin
1G5
14-9120
Purified
5 µg/mL
Human
Arginase-1
sl6arg
53-9779
Alexa Fluor 488
20 µg/mL
Human
Arginase-1
sl6arg
50-9779
Invitrogen eFluor 660
5 µg/mL
Human
Arginase-1
sl6arg
14-9779
Purified
5 µg/mL
Human, Mouse
B220
RA3-6B2
53-0452
Alexa Fluor 488
20 µg/mL
Human, Mouse, Rat, Bovine
Beta tubulin class III
2G10-TB3
14-4510
Purified
5 µg/mL
Human, Mouse, Rat, Bovine
Beta tubulin class III
2G10-TB3
53-4510
Alexa Fluor 488
10 µg/mL
Human, Mouse, Rat, Bovine
Beta tubulin class III
2G10-TB3
50-4510
eFluor 660
10 µg/mL
Human, Mouse, Rat, Bovine
Beta tubulin class III
2G10-TB3
41-4510
Invitrogen eFluor 570
5 µg/mL
Human
Brachyury
X1AO2
14-9770
Purified
5 µg/mL
Mouse
CD3
17A2
50-0032
eFluor 660
10 µg/mL
Human, Mouse and Canine
CD324, E-Cadherin
DECMA-1
53-3249
Alexa Fluor 488
20 µg/mL
Mouse
CD4
RM4-5
13-0042
Biotin
10 µg/mL
Mouse
CD4
RM4-5
41-0042
eFluor 570
5 µg/mL
Mouse
CD8
53-6.7
50-0081
eFluor 660
10 µg/mL
Mouse
CD8a
4SM15
13-0808
Biotin
10 µg/mL
Mouse
CD11b
M1/70
50-0112
eFluor 660
5 µg/mL
Human, Mouse, Rat, Chicken
Connexin 32
2A
14-9759
Purified
5 µg/mL
Human, Mouse, Rat
Connexin 43
1A
50-4759
eFluor 660
10 µg/mL
Human
Cox IV
IB52C31H10
14-9775
Purified
1 µg/mL
Human
Cox IV
IB52C31H10
41-9775
eFluor 570
1 µg/mL
Human
CoxIV
IB52C31H10
53-9775
Alexa Fluor 488
10 µg/mL
Human
Cytokeratin 7
LP5K
14-9005
Purified
10 µg/mL
Human, Mouse, Rat
Early endosome antigen 1
1G11
14-9114
Purified
1 µg/mL
Human
CD326 EpCAM
1B7
14-9326
Purified
10 µg/mL
Human
ER alpha
tbykt
14-9740
Purified
1 µg/mL
Human
Fibronectin
FN-3
14-9869
Purified
10 µg/mL
Human, Mouse, Rat, Pig
GFAP
GA5
53-9892
Alexa Fluor 488
10 µg/mL
Human
Golgin-97
CDF4
53-9767
Alexa Fluor 488
10 µg/mL
Human
Golgin-97
CDF4
50-9767
eFluor 660
5 µg/mL
Human
Golgin-97
CDF4
14-9767
Purified
1 µg/mL
Human, Mouse, Rat
Grim-19
1A8
14-9937
Purified
5 µg/mL
Human
Her2
MJD2
14-9757
Purified
5 µg/mL
Human
Her2
MJD2
41-9757
eFluor 570
5 µg/mL
Human, Mouse
Her4
HFR1
14-9687
Purified
10 µg/mL
Human
Ki-67
20Raj1
42-5699
Invitrogen eFluor 615
1 µg/mL
Human
Ki-67
20Raj1
50-5699
eFluor 660
20 µg/mL
Human
Ki-67
20Raj1
11-5699
FITC
10 µg/mL
Human
Ki-67
20Raj1
14-5699
Purified
1 µg/mL
Human
Ki-67
20Raj1
13-5699
Biotin
5 µg/mL
Human
Ki-67
20Raj1
41-5699
eFluor 570
5 µg/mL
Mouse, Rat
Ki-67
SolA15
41-5698
eFluor 570
0.5 µg/mL
Mouse, Rat
Ki-67
SolA15
50-5698
eFluor 660
10 µg/mL
Hu, m, Rt
MAP2
AP20
41-9763
eFluor 570
5 µg/mL
Mouse, Rat
MASH1
24B72D11
14-5794
Purified
20 µg/mL
Human, Mouse, Rat, Dog
Myogenin
F5D
53-5643
Alexa Fluor 488
5 µg/mL
Human, Mouse, Rat, Dog
Myogenin
F5D
14-5643
Purified
1 µg/mL
Human, Mouse, Rat, etc.
Myosin
MF20
53-6503
Alexa Fluor 488
5 µg/mL
Human, Mouse, Rat, etc.
Myosin
MF20
50-6503
eFluor 660
10 µg/mL
Mouse
Nanog
eBioMLC-51
50-5761
eFluor 660
10 µg/mL
Mouse
Nanog
eBioMLC-51
53-5761
Alexa Fluor 488
10 µg/mL
Human
Nestin
10C2
53-9843
Alexa Fluor 488
10 µg/mL
Human
Nestin
10C2
14-9843
Purified
5 µg/mL
Human, Mouse
OCT3/4
EM92
41-5841
eFluor 570
5 µg/mL
Human, Mouse, Rat
PCNA
PC10
14-9910
Purified
1 µg/mL
Human, Mouse, Rat
Phospho-ATM
10H11.E12
14-9046
Purified
10 µg/mL
Human, Mouse
Phospho-ERK1/2
MILAN8R
14-9109
Purified
5 µg/mL
Human, Mouse
Phospho-H2AX
CR55T33
14-9865
Purified
5 µg/mL
Human
Progesterone receptor
KMC912
13-9764
Biotin
2.5 µg/mL
Human
Progesterone receptor
KMC912
14-9764
Purified
1 µg/mL
Human
Progesterone receptor
KMC912
50-9764
eFluor 660
20 µg/mL
Human
Snail1
20C8
53-9859
Alexa Fluor 488
20 µg/mL
Human
Snail1
20C8
14-9859
Purified
5 µg/mL
Human, Mouse
Sox2
Btjce
41-9811
eFluor 570
5 µg/mL
Human, Mouse
Sox2
Btjce
50-9811
eFluor 660
10 µg/mL
Human, Mouse
Sox2
Btjce
14-9811
Purified
5 µg/mL
Human, Mouse
Sox9
GMPR9
53-9765
Alexa Fluor 488
10 µg/mL
Human, Mouse
Sox9
GMPR9
14-9765
Purified
1 µg/mL
Human
Survivin
STLALYV
53-9176
Alexa Fluor 488
10 µg/mL
Human, Mouse, Rat
Thrombospondin-1
A6.1
14-9756
Purified
1 µg/mL
Human, Mouse, Rat, etc.
Tubulin
DM1A
42-4502
eFluor 615
1 µg/mL
Human, Mouse, Rat, etc.
Tubulin
DM1A
53-4502
Alexa Fluor 488
5 µg/mL
Human, Mouse, Rat, etc.
Tubulin
DM1A
14-4502
Purified
5 µg/mL
Human, Mouse, Rat, etc.
Tubulin
DM1A
13-4502
Biotin
10 µg/mL
Human, Rat, Chicken, Canine
Vimentin
V9
41-9897
eFluor 570
1 µg/mL
Human, Rat, Chicken, Canine
Vimentin
V9
50-9897
eFluor 660
5 µg/mL
Human, Mouse, Rat, Avian
Vinculin
7F9
41-9777
eFluor 570
10 µg/mL
Human, Mouse, Rat, Avian
Vinculin
7F9
53-9777
Alexa Fluor 488
20 µg/mL
Human, Mouse, Rat, Avian
Vinculin
7F9
14-9777
Purified
5 µg/mL
Mouse, Rat, Canine
ZO-1
R26.4C
41-9776
eFluor 570
1 µg/mL
Human
CD3
OKT3
50-0037
eFluor 660
0.12 µg/ml
Human
CD4
N1UG0
41-2444
eFluor 570
20 µg/mL
Mouse
FoxP3
FJK-16s
53-5773
Alexa Fluor 488
10 µg/mL
Not recommended 
Human, Mouse
B220
RA3-6B2
50-0452
eFluor 660
10 µg/mL
Mouse
CD4
GK1.5
53-0041
Alexa Fluor 488
10 µg/mL
Mouse
CD4
GK1.5
50-0041
eFluor 660
5 µg/mL
Mouse, Rat
Nestin
Rat-401
14-5843
Purified
10 µg/mL
Mouse, Human, Rat, etc.
Cytokeratin
AE1/AE3
42-9003
eFluor 615
5 µg/mL
Mouse, Human, Rat, etc.
Cytokeratin
AE1/AE3
41-9003
eFluor 570
1 µg/mL
Mouse, Human, Rat, etc.
Cytokeratin
AE1/AE3
53-9003
Alexa Fluor 488
1 µg/mL
Human, Mouse, Rat, Chicken
Connexin 32
2A
53-9759
Alexa Fluor 488
1 µg/mL
Human, Mouse, Rat
Connexin 43
1A
14-4759
Purified
5 µg/mL
Human
Heat shock protein 27
STRSN
14-9112
Purified
1 µg/mL
Mouse
MHC Class II
M5/114.15.2
11-5321
FITC
20 µg/mL
Human, Mouse, Rat
PCNA
PC10
42-9910
eFluor 615
0.5 µg/mL
Human
CD8
AMC908
50-0008
eFluor 660
5 µg/mL