Alexa Fluor 546 dyeInvitrogen Alexa Fluor 546 dye is a bright, yellow-fluorescent dye that can be excited using the 488 nm or 532 nm laser lines. For stable signal generation in imaging and flow cytometry, Alexa Fluor 546 dye is pH-insensitive over a wide molar range.
Probes with high fluorescence quantum yield and high photostability allow detection of low-abundance biological structures with great sensitivity. Alexa Fluor 546 dye molecules can be attached to proteins at high molar ratios without significant self-quenching, enabling brighter conjugates and more sensitive detection.
We offer Alexa Fluor 546 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection. In addition, reactive dye forms and protein labeling kits are available to allow you to generate your own antibody conjugates or probes.
Alexa Fluor 546 dye dashboard
Initial brightness
An alternative for tetramethylrhodamine or Cy3 dye, with higher initial intensity than Alexa Fluor 555 dye, but lower photostability in buffer or antifade.  
Photostability in buffer
Photostability in antifade
  488/532 TRITC 556 573  
Laser line
Common filter set
Excitation max
Emission max
Alexa Fluor 546 products
We offer a broad range of Alexa Fluor  546 products including labeling kits, secondary antibodies, bioconjugates, and more.
Learn more here:
Alexa Fluor® 546 dye
The intermediate filaments in bovine pulmonary artery endothelial cells were visualized using our anti-desmin antibody, followed by Alexa Fluor 647 goat anti–mouse IgG antibody (purple). Endogenous biotin in the mitochondria was labeled with Alexa Fluor 546 streptavidin (red), and DNA in the cell was stained with DAPI (white).
Alexa Fluor 546 reactive dyes
Select dye-labeling chemistries for conjugation with the following reactive groups: 
Amine Thiol
Alexa Fluor 546 protein/antibody labeling kits
Select an optimized labeling kit for your target antibody or protein:
Target protein Kit Cat. No.
Site-specific antibody labeling
<1–20 µg mouse IgG1 Zenon Alexa Fluor 546 Mouse IgG1 Labeling Kit Z25004
<1–20 µg rabbit IgG Zenon Alexa Fluor 546 Rabbit IgG Labeling Kit Z25304
Conventional antibody/protein labeling
100 µg antibody/protein Alexa Fluor 546 Antibody/Protein Labeling Kit A20183
1 mg antibody/protein Alexa Fluor 546 Antibody/Protein Labeling Kit A10237
Alexa Fluor dye family
Illuminating research for over 15 years
Alexa Fluor® dyesAlexa Fluor dyes are among the most trusted fluorescent dyes available today. In addition to superior performance, you’ll get a technical support team experienced in problem-solving, >50,000 published references, application and experimental tips, and protocols to help with experimental planning. For an outstanding research experience, choose Alexa Fluor dyes.
Resources
Molecular Probes Handbook
Cell stain tool
The significantly revised 11th Edition is the most complete fluorescence labeling and detection reference available. The Molecular Probes Handbook can help you achieve the best experimental results for your fluorescence labeling and detection experiments. View the handbook online or order your own copy.
Go to the Handbook online
Order your copy of the Handbook
Antifade reagents
Cell stain tool
Antifade reagents suppress photobleaching and preserve the signals of your fluorescently labeled target molecules. They offer excellent compatibility with a multitude of dyes across the spectrum, making them especially valuable tools for multicolor applications. For added convenience, some formulations include the nuclear counterstain DAPI, eliminating the need for a separate counterstaining step.
See the performance of this dye in SlowFade Antifade reagents
See the performance of this dye in ProLong Antifade reagents
Cell stain tool
Cell stain tool
Stain your own cell by using our simulation tool to develop reproducible results with many of our signature fluorescent dyes. Develop your multiplexing strategy for antibody probes, reactive dyes, and counterstains to create your perfectly labeled fluorescent cell.
Go to the cell stain tool
Fluorescence SpectraViewer
thumbnail50x50.par.42353.image.50.50.1.thu-spectraviewer-jpg
Plot and compare spectra and check the spectral compatibility for common fluorophores including all of the fluorescent probes from Thermo Fisher Scientific. The Fluorescence SpectraViewer tool is formatted for both microscope imaging and flow cytometry applications.
Go to the Fluorescence SpectraViewer