Cell Type
Source
Cell Definition
Cell Source
Hepatocytes: pure culture
Rat
Primary, adult
Isolation
Hepatocytes: co-culture with Kupffer cells or non-parenchymal cells
Rat
Primary, adult Isolation
Mixed heptaoblasts and oval (liver stem) cells
Rat
Primary, neonatal
Isolation
Hepatocytes - C3A
Human
Cell line, clone derivative of HepG2
ATCC (CRL-10741)
Endothelial cells dermal microvascular (HDMEC)
Human
Primary, frozen
Clonetics (CC-2543)
Endothelial cells umbilical vein (HUVEC)
Human
Primary, fresh
Isolation
Endothelial cells dermal microvascular-transfected (HMEC-1)
Human
Cell line (transfected with encoding SV-40 large T antigen)
Gift*
Endothelial cells bovine aortic (BAEC)
Bovine
Primary, frozen
Isolation
Dermal fibroblasts (NHDF)
Human
Primary, frozen
Clonetics (CC-2511)
Cardiac fibroblasts
Rat
Primary
Isolation
Cardiac myocytes
Rat
Primary, embryonic
Isolation
Cardiac myocytes
Rat
Primary, neonatal
Isolation
Embryonic stem cells
Human
(H-9-Wisconsin lines, Israeli lines)
Technion
Mesenchymal stem cells, bone marrow
Human
Primary, bone marrow
Isolation based on plate adherence
Mesenchymal stem cells, cord blood (CD133+ enriched)
Human
Primary, cord blood
Isolation based on plate adherence
Hematopoeitic stem cells, cord blood (CD133+ enriched)
Human
Primary, cord blood
Isolation
*Gift courtesy of Smadar Cohen and Tsiona Elkayam Ben-Gurion, University of Negev