Thermo Scientific™ Nunc™ Lab-Tek™ and Lab-Tek II™ Chambered Coverglass products provide optimal optical characteristics required for high magnification microscopy and confocal image analysis.
Which Chambered Coverglass is right for you?
Nunc Lab-Tek Chambered Coverglass
Nunc Lab-Tek II Chambered Coverglass
Formats 1, 2, 4, 8 well 1, 2, 4, 8 well
Chamber material Clear polystyrene lid with no handle to facilitate stacking. Clear polystyrene lid with handle to facilitate aseptic manipulations.
Coverglass material
#1.0 borosilicate coverglass
#1.5 borosilicate coverglass
Sealant Non-toxic medical grade silicone Biocompatible acrylic adhesive
Removable chamber No
Imaging application
Inverted microscopes Inverted microscopes
  Order now Order now