BglII nucleotide recognition sequence
Figure color legend
Name derivation: Bacillus globigii
Recognition and Cutting site: A^GATC T
T CTAG^A
Resulting DNA Ends: 5′ protruding ends
Prototype: Bglii
Isoschizomers: None
Sensitivity to methylation:  EcoBI impaired if overlapped
Neoschizomers None
Anza RE generating compatible ends Anza™ 5 BamHI, Anza™ 55 MboI, Anza™ 73 BclI, Anza™ 97 Bsp143I
References