Name derivation: Desulfovibrio desulfuricans
Recognition and Cutting site: C^TNA G
G ANT^C
Resulting DNA Ends: 5′ protruding ends
Prototype: DdeI
Isoschizomers: HpyF3I, BstDEI
Sensitivity to methylation:  None
Neoschizomers None
Anza RE generating compatible ends Cannot be predicted as the recognition sequence contains variable nucleotides
DdeI nucleotide recognition sequence Figure color legend
References