SnaBI nucleotide recognition sequence
Figure color legend
Name derivation: Sphaerotilus natans
Recognition and Cutting site: TAC^GTA
ATG^CAT
Resulting DNA Ends: blunt
Prototype: SnaBI
Isoschizomers: Eco105I, BstSNI
Sensitivity to methylation:  CpG blocked
Neoschizomers None
Anza RE generating compatible ends Any enzyme generating blunt ends
References