BindIt
KingFisher automated purification systems are designed to adapt to your changing research needs. That’s why we include BindIt PC software with all KingFisher instruments so that you can create, modify and store your protocols. You can also search and download automated protocols for our MagMAX nucleic acid purification kits, and Dynabead and Pierce protein purification beads and kits.
Create and store protocols using Thermo Scientific BindIt Software for Thermo Scientific KingFisher instrument systems. Once a protocol has been created, the protocol can either be transferred to your KingFisher instrument memory or executed directly from the software. Based on a step list, the parameters for the active step are shown on the screen. All steps have default parameters which can be changed according to the demands of the application. Alternatively, download and execute protocols from our extensive library.
Features of BindIt Software
  • Compatible with open-platform KingFisher instrument systems
  • Allows KIngFisher plates and reagents to be defined in the plate layout
  • Generates status reports that include run log, plate layout, and step parameters
 Instructions for downloading and installing protocols:
  1. Make sure you have BindIt 4.0 Software installed in your computer
  2. Click the protocol in the table (DNA/RNA protocols, Protein protocols). Protocol opens directly in BindIt Software
  3. Save the protocol in your computer in the preferred folder.
  4. Connect your PC with USB cable to KingFisher instrument and select Home> Connect in BindIt Software.
  5. You can start the protocol from the BindIt or transfer it into instrument. If you transfer the protocol select Home > Transfer in BindIt Software.
  6. Select Upload and find the saved protocol. Click open.
  7. Select the folder (category) you want to store the protocol in the instrument.
  8. Your protocol can be now found in the instrument memory.
  9. Disconnect your instrument from BindIt and check using keypad that you protocol is in the desired folder.
Already have a KingFisher Instrument and BindIt Software?
Download BindIt Software Upgrade
No records were found matching your criteria
Nucleic acid Sample type Kit name SKU Format Software protocol
DNA Blood, Buccal, Swabs MagMAX DNA Multi-Sample Ultra A25597
A25598
500 preps
2,500 preps
MagMAX DNA Blood Buccal
 Duo Prime Flex
DNA Blood, Buccal, Swabs MagMAX DNA Multi-Sample Ultra w/ extra PK and beads A25919
A25920
500 preps
2,500 preps
MagMAX DNA Blood Buccal
Duo PrimeFlex
DNA Blood, Buccal, Swabs MagMAX DNA Multi-Sample 4413020 50 preps MagMAX DNA Blood Buccal
Duo PrimeFlex
DNA Blood, Buccal, Swabs MagMAX-96 DNA Multi-Sample 4413021
4413022
96 preps
5 x 96 preps
MagMAX DNA Blood Buccal
 Duo PrimeFlex
DNA Tissue, Cells MagMAX DNA Multi-Sample Ultra A25597
A25598
500 preps
2,500 preps
MagMAX DNA tissue cells
Duo PrimeFlex
DNA Tissue, Cells MagMAX DNA Multi-Sample Ultra w/ extra PK and beads A25919
A25920
500 preps
2,500 preps
MagMAX DNA tissue cells
Duo PrimeFlex
DNA Tissue, Cells MagMAX DNA Multi-Sample 4413021 50 preps MagMAX DNA tissue cells
Duo PrimeFlex
DNA Tissue, Cells MagMAX-96 DNA Multi-Sample 4413021
4413022
96 preps
5 x 96 preps
MagMAX DNA tissue cells
Duo PrimeFlex
RNA/DNA Viral MagMAX Pathogen RNA/DNA 4462359 480 preps MagMAX Pathogen Std Volume
Duo PrimeFlex
RNA/DNA Viral MagMAX Viral RNA AM1836 96 preps MagMAX Pathogen Std Volume
Duo PrimeFlex
RNA/DNA Viral MagMAX Total Nucleic Acid AM1840 100 preps MagMAX Pathogen Std Volume
Duo PrimeFlex
RNA/DNA Viral MagMAX Pathogen RNA/DNA 4462359 480 preps MagMAX Pathogen High Volume
Duo PrimeFlex
RNA/DNA Viral MagMAX Viral RNA AM1836 96 preps MagMAX Pathogen High Volume
Duo PrimeFlex
RNA/DNA Viral MagMAX Total Nucleic Acid AM1840 100 preps MagMAX Pathogen High Volume
Duo PrimeFlex
RNA Viral MagMAX-96 AI/ND Viral RNA Isolation Kit AM1835 384 preps MagMAX AI/ND Viral RNA
Duo PrimeFlex
RNA/
microRNA
Tissue, Cells MagMAX mirVana Total RNA A27828
A27828
96 preps
96 preps
MagMAX mirVana tissue cells
Duo PrimeFlex
MagMAX mirVana biofluids
Duo PrimeFlex
RNA/
microRNA
Tissue, Cells
Blood, Plasma, Urine
MagMAX mirVana Total RNA A27828
A27828
96 preps
96 preps
MagMAX mirVana tissue cells
Duo PrimeFlex
MagMAX mirVana biofluids
Duo PrimeFlex
DNA Plasma MagMAX Cell-Free DNA A29319
A29319
96 preps
96 preps
MagMAX Cell-Free DNA 2mL plasma
Duo PrimeFlex
MagMAX Cell-Free DNA 4mL plasma
Duo PrimeFlex
RNA Tissue, Cells, Plants MagMAX-96 Total RNA AM1830 96 preps MagMAX Total RNA tissue cell plant
Duo PrimeFlex
RNA Blood MagMAX for Stabilized Blood (Paxgene tubes) 4451894 96 preps MagMAX for Paxgene tubes
Duo PrimeFlex
RNA Blood MagMAX for Stabilized Blood (Tempus tubes) 4451893 96 preps MagMAX for Tempus tubes
Duo PrimeFlex
DNA Leukocytes MagMAX DNA Multi-Sample Ultra A25597 500 preps MagMAX DNA leukocytes
Duo PrimeFlex
mRNA   Dynabeads mRNA Direct 61011
61011
5 mL
10 mL
Dynabeads mRNA from crude samples
Duo PrimeFlex
RNA   MagMAX for Microarrays Total RNA Isolation AM1839 96 preps MagMAX for Microarrays with 100uL aqueous phase
Duo PrimeFlex
DNA FFPE MagMAX FFPE DNA/RNA Ultra Kit A31881 1 kit MagMAX FFPE Ultra DNA only
Duo PrimeFlex
RNA FFPE MagMAX FFPE DNA/RNA Ultra Kit A31881 1 kit MagMAX FFPE Ultra RNA only
Duo PrimeFlex
RNA/DNA FFPE MagMAX FFPE DNA/RNA Ultra Kit A31881 1 kit MagMAX FFPE Ultra DNA and RNA
Duo PrimeFlex
 Instructions for downloading and installing protocols:
  1. Make sure you have BindIt 4.0 Software installed in your computer
  2. Click the protocol in the table. Protocol opens directly in BindIt Software
  3. Save the protocol in your computer in the preferred folder.
  4. Connect your PC with USB cable to KingFisher instrument and select Home> Connect in BindIt Software.
  5. You can start the protocol from the BindIt or transfer it into instrument. If you transfer the protocol select Home > Transfer in BindIt Software.
  6. Select Upload and find the saved protocol. Click open.
  7. Select the folder (category) you want to store the protocol in the instrument.
  8. Your protocol can be now found in the instrument memory.
  9. Disconnect your instrument from BindIt and check using keypad that you protocol is in the desired folder.
† Strategy (Indirect or Direct). This refers to the order of binding steps, and it primarily pertains to immunoprecipitation protocols. Indirect protocols involve incubating the target-containing sample and antibody first, and then incubating with the beads second. Direct protocols involve incubating beads and antibody first (either to bind or covalently couple the IP antibody), and then incubating the bead-antibody complex with the sample.
No records were found matching your criteria
Ligand Product Protocol name: description Strategy† Download file
Protein A Dynabeads Protein A IP Dynabeads M-280 Protein A G Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Protein A Dynabeads Protein A IP Dynabeads M-280 Protein A G Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Duo Prime
Protein A Dynabeads Protein A Immunoprecipitation Kit IP Dynabeads M-280 Protein A G Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Protein A Dynabeads Protein A Immunoprecipitation Kit IP Dynabeads M-280 Protein A G Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Duo Prime
Protein G Dynabeads Protein G IP Dynabeads M-280 Protein A G Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Protein G Dynabeads Protein G IP Dynabeads M-280 Protein A G Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Duo Prime
Protein G Dynabeads Protein G Immunoprecipitation Kit IP Dynabeads M-280 Protein A G Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Protein G Dynabeads Protein G Immunoprecipitation Kit IP Dynabeads M-280 Protein A G Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Duo Prime
Sheep anti-Mouse IgG Dynabeads Sheep anti-Mouse IgG IP Dynabeads M-280 SAM SAR Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Sheep anti-Rabbit IgG Dynabeads Sheep anti-Rabbit IgG IP Dynabeads M-280 SAM SAR Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Streptavidin Dynabeads M-280 Streptavidin IP Dynabeads M-280 SA Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Glutathione Pierce Glutathione Magnetic Agarose Beads Purify GST tag Magnetic Agarose GSH Direct:
GST fusion protein purification
Direct Flex
Duo Prime
Ni-NTA Pierce Ni-NTA Magnetic Agarose Beads Purify His tag Magnetic Agarose Ni-NTA Direct:
His-tagged fusion protein purification
Direct Flex
Duo Prime
Protein A/G Pierce Protein A/G Magnetic Agarose Beads Purify Ab Magnetic Agarose Protein AG Direct:
Antibody purification of IgG containing Fc region
Direct Flex
Duo Prime
Anti-HA Antibody Pierce Anti-HA Magnetic Beads IP HA tag Pierce anti-HA Ab Direct:
HA-tagged fusion protein purification or immunoprecipitation (IP or Co-IP)
Direct Flex
Anti-c-Myc Antibody Pierce Anti-c-Myc Magnetic Beads IP Myc tag Pierce anti-Myc Ab Direct:
Myc-tagged fusion protein purification or immunoprecipitation (IP or Co-IP)
Direct Flex
Glutathione Pierce Glutathione Magnetic Beads Purify GST tag Pierce GSH Direct:
GST fusion protein purification or pull-down
Direct Flex
Ni-NTA HisPur Ni-NTA Magnetic Beads Purify His tag Pierce Ni-NTA Direct:
His-tagged fusion protein purification or pull-down
Direct Flex
NHS Ester Pierce NHS-Activated Magnetic Beads Couple Pierce NHS ester Direct:
Coupling (immobilization) of primary amine molecule (protein, peptide)
Direct Flex
NHS Ester Pierce NHS-Activated Magnetic Beads IP Pierce NHS ester Direct:
Affinity purification with coupled (immobilized) primary amine molecule (protein, peptide)
Direct Flex
Protein A/G Pierce Protein A/G Magnetic Beads Purify Ab Pierce Protein AGL Direct:
Antibody purification of IgG containing Fc region
Direct Flex
Protein A Pierce Protein A Magnetic Beads Purify Ab Pierce Protein AGL Direct:
Antibody purification of IgG containing Fc region
Direct Flex
Protein G Pierce Protein G Magnetic Beads Purify Ab Pierce Protein AGL Direct:
Antibody purification of IgG containing Fc region
Direct Flex
Protein L Pierce Protein L Magnetic Beads Purify Ab Pierce Protein AGL Direct:
Antibody purification of immunoglobulins containing kappa light chains
Direct Flex
Protein A/G Pierce Protein A/G Magnetic Beads IP Pierce Protein AGL Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Protein A Pierce Protein A Magnetic Beads IP Pierce Protein AGL Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Protein G Pierce Protein G Magnetic Beads IP Pierce Protein AGL Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Protein A/G Pierce Classic Magnetic IP/Co-IP Kit IP Pierce Protein AGL Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
NHS Ester Pierce Direct Magnetic IP/Co-IP Kit IP Pierce Direct IP Kit Direct:
Immunoprecipitation (IP or Co-IP) or pull-down after coupling (immobilization) of IgG, protein or peptide to beads
Direct Flex
Protein A/G Pierce Crosslink Magnetic IP/Co-IP Kit IP Pierce Crosslink IP Kit Direct:
Immunoprecipitation (IP or Co-IP) after crosslinking IgG to beads
Direct Flex
Protein A/G Pierce Magnetic ChIP Kit ChIP Pierce ChIP Kit Indirect:
Chromatin immunoprecipition (ChIP) of protein-DNA complexes
Indirect Flex
Streptavidin Pierce Streptavidin Magnetic Beads IP Pierce SA heated elution Indirect:
Purification or enrichment of biotinylated or desthiobiotinylated molecules using heated elution
Indirect Flex
Streptavidin Pierce Streptavidin Magnetic Beads IP Pierce SA low pH elution Indirect:
Purification or enrichment of biotinylated or desthiobiotinylated molecules using low-pH elution
Indirect Flex
Titanium Dioxide (TiO2) Pierce Magnetic Titanium Dioxide Phosphopeptide Enrichment Kit Enrich pPep Pierce pPep Kit Direct deepwell plate:
Phosphopeptide enrichment in deep-well plates (peptides containing phosphorylated amino acids, e.g., phosphoserine, phosphotyrosine or phosphotyrosine)
Direct Flex
Titanium Dioxide (TiO2) Pierce Magnetic Titanium Dioxide Phosphopeptide Enrichment Kit Enrich pPep Pierce pPep Kit Direct PCR plate:
Phosphopeptide enrichment in PCR plates (peptides containing phosphorylated amino acids, e.g., phosphoserine, phosphotyrosine or phosphotyrosine)
Direct Flex
Instructions for downloading and installing protocols:
  1. Make sure you have BindIt 4.0 Software installed in your computer
  2. Click the protocol in the table. Protocol opens directly in BindIt Software
  3. Save the protocol in your computer in the preferred folder.
  4. Connect your PC with USB cable to KingFisher instrument and select Home> Connect in BindIt Software.
  5. You can start the protocol from the BindIt or transfer it into instrument. If you transfer the protocol select Home > Transfer in BindIt Software.
  6. Select Upload and find the saved protocol. Click open.
  7. Select the folder (category) you want to store the protocol in the instrument.
  8. Your protocol can be now found in the instrument memory.
  9. Disconnect your instrument from BindIt and check using keypad that you protocol is in the desired folder.
Create and store protocols using Thermo Scientific BindIt Software for Thermo Scientific KingFisher instrument systems. Once a protocol has been created, the protocol can either be transferred to your KingFisher instrument memory or executed directly from the software. Based on a step list, the parameters for the active step are shown on the screen. All steps have default parameters which can be changed according to the demands of the application. Alternatively, download and execute protocols from our extensive library.
Features of BindIt Software
  • Compatible with open-platform KingFisher instrument systems
  • Allows KIngFisher plates and reagents to be defined in the plate layout
  • Generates status reports that include run log, plate layout, and step parameters
 Instructions for downloading and installing protocols:
  1. Make sure you have BindIt 4.0 Software installed in your computer
  2. Click the protocol in the table (DNA/RNA protocols, Protein protocols). Protocol opens directly in BindIt Software
  3. Save the protocol in your computer in the preferred folder.
  4. Connect your PC with USB cable to KingFisher instrument and select Home> Connect in BindIt Software.
  5. You can start the protocol from the BindIt or transfer it into instrument. If you transfer the protocol select Home > Transfer in BindIt Software.
  6. Select Upload and find the saved protocol. Click open.
  7. Select the folder (category) you want to store the protocol in the instrument.
  8. Your protocol can be now found in the instrument memory.
  9. Disconnect your instrument from BindIt and check using keypad that you protocol is in the desired folder.
Already have a KingFisher Instrument and BindIt Software?
Download BindIt Software Upgrade
No records were found matching your criteria
Nucleic acid Sample type Kit name SKU Format Software protocol
DNA Blood, Buccal, Swabs MagMAX DNA Multi-Sample Ultra A25597
A25598
500 preps
2,500 preps
MagMAX DNA Blood Buccal
 Duo Prime Flex
DNA Blood, Buccal, Swabs MagMAX DNA Multi-Sample Ultra w/ extra PK and beads A25919
A25920
500 preps
2,500 preps
MagMAX DNA Blood Buccal
Duo PrimeFlex
DNA Blood, Buccal, Swabs MagMAX DNA Multi-Sample 4413020 50 preps MagMAX DNA Blood Buccal
Duo PrimeFlex
DNA Blood, Buccal, Swabs MagMAX-96 DNA Multi-Sample 4413021
4413022
96 preps
5 x 96 preps
MagMAX DNA Blood Buccal
 Duo PrimeFlex
DNA Tissue, Cells MagMAX DNA Multi-Sample Ultra A25597
A25598
500 preps
2,500 preps
MagMAX DNA tissue cells
Duo PrimeFlex
DNA Tissue, Cells MagMAX DNA Multi-Sample Ultra w/ extra PK and beads A25919
A25920
500 preps
2,500 preps
MagMAX DNA tissue cells
Duo PrimeFlex
DNA Tissue, Cells MagMAX DNA Multi-Sample 4413021 50 preps MagMAX DNA tissue cells
Duo PrimeFlex
DNA Tissue, Cells MagMAX-96 DNA Multi-Sample 4413021
4413022
96 preps
5 x 96 preps
MagMAX DNA tissue cells
Duo PrimeFlex
RNA/DNA Viral MagMAX Pathogen RNA/DNA 4462359 480 preps MagMAX Pathogen Std Volume
Duo PrimeFlex
RNA/DNA Viral MagMAX Viral RNA AM1836 96 preps MagMAX Pathogen Std Volume
Duo PrimeFlex
RNA/DNA Viral MagMAX Total Nucleic Acid AM1840 100 preps MagMAX Pathogen Std Volume
Duo PrimeFlex
RNA/DNA Viral MagMAX Pathogen RNA/DNA 4462359 480 preps MagMAX Pathogen High Volume
Duo PrimeFlex
RNA/DNA Viral MagMAX Viral RNA AM1836 96 preps MagMAX Pathogen High Volume
Duo PrimeFlex
RNA/DNA Viral MagMAX Total Nucleic Acid AM1840 100 preps MagMAX Pathogen High Volume
Duo PrimeFlex
RNA Viral MagMAX-96 AI/ND Viral RNA Isolation Kit AM1835 384 preps MagMAX AI/ND Viral RNA
Duo PrimeFlex
RNA/
microRNA
Tissue, Cells MagMAX mirVana Total RNA A27828
A27828
96 preps
96 preps
MagMAX mirVana tissue cells
Duo PrimeFlex
MagMAX mirVana biofluids
Duo PrimeFlex
RNA/
microRNA
Tissue, Cells
Blood, Plasma, Urine
MagMAX mirVana Total RNA A27828
A27828
96 preps
96 preps
MagMAX mirVana tissue cells
Duo PrimeFlex
MagMAX mirVana biofluids
Duo PrimeFlex
DNA Plasma MagMAX Cell-Free DNA A29319
A29319
96 preps
96 preps
MagMAX Cell-Free DNA 2mL plasma
Duo PrimeFlex
MagMAX Cell-Free DNA 4mL plasma
Duo PrimeFlex
RNA Tissue, Cells, Plants MagMAX-96 Total RNA AM1830 96 preps MagMAX Total RNA tissue cell plant
Duo PrimeFlex
RNA Blood MagMAX for Stabilized Blood (Paxgene tubes) 4451894 96 preps MagMAX for Paxgene tubes
Duo PrimeFlex
RNA Blood MagMAX for Stabilized Blood (Tempus tubes) 4451893 96 preps MagMAX for Tempus tubes
Duo PrimeFlex
DNA Leukocytes MagMAX DNA Multi-Sample Ultra A25597 500 preps MagMAX DNA leukocytes
Duo PrimeFlex
mRNA   Dynabeads mRNA Direct 61011
61011
5 mL
10 mL
Dynabeads mRNA from crude samples
Duo PrimeFlex
RNA   MagMAX for Microarrays Total RNA Isolation AM1839 96 preps MagMAX for Microarrays with 100uL aqueous phase
Duo PrimeFlex
DNA FFPE MagMAX FFPE DNA/RNA Ultra Kit A31881 1 kit MagMAX FFPE Ultra DNA only
Duo PrimeFlex
RNA FFPE MagMAX FFPE DNA/RNA Ultra Kit A31881 1 kit MagMAX FFPE Ultra RNA only
Duo PrimeFlex
RNA/DNA FFPE MagMAX FFPE DNA/RNA Ultra Kit A31881 1 kit MagMAX FFPE Ultra DNA and RNA
Duo PrimeFlex
 Instructions for downloading and installing protocols:
  1. Make sure you have BindIt 4.0 Software installed in your computer
  2. Click the protocol in the table. Protocol opens directly in BindIt Software
  3. Save the protocol in your computer in the preferred folder.
  4. Connect your PC with USB cable to KingFisher instrument and select Home> Connect in BindIt Software.
  5. You can start the protocol from the BindIt or transfer it into instrument. If you transfer the protocol select Home > Transfer in BindIt Software.
  6. Select Upload and find the saved protocol. Click open.
  7. Select the folder (category) you want to store the protocol in the instrument.
  8. Your protocol can be now found in the instrument memory.
  9. Disconnect your instrument from BindIt and check using keypad that you protocol is in the desired folder.
† Strategy (Indirect or Direct). This refers to the order of binding steps, and it primarily pertains to immunoprecipitation protocols. Indirect protocols involve incubating the target-containing sample and antibody first, and then incubating with the beads second. Direct protocols involve incubating beads and antibody first (either to bind or covalently couple the IP antibody), and then incubating the bead-antibody complex with the sample.
No records were found matching your criteria
Ligand Product Protocol name: description Strategy† Download file
Protein A Dynabeads Protein A IP Dynabeads M-280 Protein A G Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Protein A Dynabeads Protein A IP Dynabeads M-280 Protein A G Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Duo Prime
Protein A Dynabeads Protein A Immunoprecipitation Kit IP Dynabeads M-280 Protein A G Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Protein A Dynabeads Protein A Immunoprecipitation Kit IP Dynabeads M-280 Protein A G Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Duo Prime
Protein G Dynabeads Protein G IP Dynabeads M-280 Protein A G Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Protein G Dynabeads Protein G IP Dynabeads M-280 Protein A G Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Duo Prime
Protein G Dynabeads Protein G Immunoprecipitation Kit IP Dynabeads M-280 Protein A G Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Protein G Dynabeads Protein G Immunoprecipitation Kit IP Dynabeads M-280 Protein A G Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Duo Prime
Sheep anti-Mouse IgG Dynabeads Sheep anti-Mouse IgG IP Dynabeads M-280 SAM SAR Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Sheep anti-Rabbit IgG Dynabeads Sheep anti-Rabbit IgG IP Dynabeads M-280 SAM SAR Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Streptavidin Dynabeads M-280 Streptavidin IP Dynabeads M-280 SA Direct:
Immunoprecipitation (IP or Co-IP)
Direct Flex
Duo Prime
Glutathione Pierce Glutathione Magnetic Agarose Beads Purify GST tag Magnetic Agarose GSH Direct:
GST fusion protein purification
Direct Flex
Duo Prime
Ni-NTA Pierce Ni-NTA Magnetic Agarose Beads Purify His tag Magnetic Agarose Ni-NTA Direct:
His-tagged fusion protein purification
Direct Flex
Duo Prime
Protein A/G Pierce Protein A/G Magnetic Agarose Beads Purify Ab Magnetic Agarose Protein AG Direct:
Antibody purification of IgG containing Fc region
Direct Flex
Duo Prime
Anti-HA Antibody Pierce Anti-HA Magnetic Beads IP HA tag Pierce anti-HA Ab Direct:
HA-tagged fusion protein purification or immunoprecipitation (IP or Co-IP)
Direct Flex
Anti-c-Myc Antibody Pierce Anti-c-Myc Magnetic Beads IP Myc tag Pierce anti-Myc Ab Direct:
Myc-tagged fusion protein purification or immunoprecipitation (IP or Co-IP)
Direct Flex
Glutathione Pierce Glutathione Magnetic Beads Purify GST tag Pierce GSH Direct:
GST fusion protein purification or pull-down
Direct Flex
Ni-NTA HisPur Ni-NTA Magnetic Beads Purify His tag Pierce Ni-NTA Direct:
His-tagged fusion protein purification or pull-down
Direct Flex
NHS Ester Pierce NHS-Activated Magnetic Beads Couple Pierce NHS ester Direct:
Coupling (immobilization) of primary amine molecule (protein, peptide)
Direct Flex
NHS Ester Pierce NHS-Activated Magnetic Beads IP Pierce NHS ester Direct:
Affinity purification with coupled (immobilized) primary amine molecule (protein, peptide)
Direct Flex
Protein A/G Pierce Protein A/G Magnetic Beads Purify Ab Pierce Protein AGL Direct:
Antibody purification of IgG containing Fc region
Direct Flex
Protein A Pierce Protein A Magnetic Beads Purify Ab Pierce Protein AGL Direct:
Antibody purification of IgG containing Fc region
Direct Flex
Protein G Pierce Protein G Magnetic Beads Purify Ab Pierce Protein AGL Direct:
Antibody purification of IgG containing Fc region
Direct Flex
Protein L Pierce Protein L Magnetic Beads Purify Ab Pierce Protein AGL Direct:
Antibody purification of immunoglobulins containing kappa light chains
Direct Flex
Protein A/G Pierce Protein A/G Magnetic Beads IP Pierce Protein AGL Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Protein A Pierce Protein A Magnetic Beads IP Pierce Protein AGL Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Protein G Pierce Protein G Magnetic Beads IP Pierce Protein AGL Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
Protein A/G Pierce Classic Magnetic IP/Co-IP Kit IP Pierce Protein AGL Indirect:
Immunoprecipitation (IP or Co-IP)
Indirect Flex
NHS Ester Pierce Direct Magnetic IP/Co-IP Kit IP Pierce Direct IP Kit Direct:
Immunoprecipitation (IP or Co-IP) or pull-down after coupling (immobilization) of IgG, protein or peptide to beads
Direct Flex
Protein A/G Pierce Crosslink Magnetic IP/Co-IP Kit IP Pierce Crosslink IP Kit Direct:
Immunoprecipitation (IP or Co-IP) after crosslinking IgG to beads
Direct Flex
Protein A/G Pierce Magnetic ChIP Kit ChIP Pierce ChIP Kit Indirect:
Chromatin immunoprecipition (ChIP) of protein-DNA complexes
Indirect Flex
Streptavidin Pierce Streptavidin Magnetic Beads IP Pierce SA heated elution Indirect:
Purification or enrichment of biotinylated or desthiobiotinylated molecules using heated elution
Indirect Flex
Streptavidin Pierce Streptavidin Magnetic Beads IP Pierce SA low pH elution Indirect:
Purification or enrichment of biotinylated or desthiobiotinylated molecules using low-pH elution
Indirect Flex
Titanium Dioxide (TiO2) Pierce Magnetic Titanium Dioxide Phosphopeptide Enrichment Kit Enrich pPep Pierce pPep Kit Direct deepwell plate:
Phosphopeptide enrichment in deep-well plates (peptides containing phosphorylated amino acids, e.g., phosphoserine, phosphotyrosine or phosphotyrosine)
Direct Flex
Titanium Dioxide (TiO2) Pierce Magnetic Titanium Dioxide Phosphopeptide Enrichment Kit Enrich pPep Pierce pPep Kit Direct PCR plate:
Phosphopeptide enrichment in PCR plates (peptides containing phosphorylated amino acids, e.g., phosphoserine, phosphotyrosine or phosphotyrosine)
Direct Flex
Instructions for downloading and installing protocols:
  1. Make sure you have BindIt 4.0 Software installed in your computer
  2. Click the protocol in the table. Protocol opens directly in BindIt Software
  3. Save the protocol in your computer in the preferred folder.
  4. Connect your PC with USB cable to KingFisher instrument and select Home> Connect in BindIt Software.
  5. You can start the protocol from the BindIt or transfer it into instrument. If you transfer the protocol select Home > Transfer in BindIt Software.
  6. Select Upload and find the saved protocol. Click open.
  7. Select the folder (category) you want to store the protocol in the instrument.
  8. Your protocol can be now found in the instrument memory.
  9. Disconnect your instrument from BindIt and check using keypad that you protocol is in the desired folder.