PCR Clean-Up From Complex Mixtures of DNA
PCR clean-up is a routine but time-consuming laboratory procedure. Now you can use an easier, faster, safer method and get superior results. Purify DNA using a simple and rapid PCR clean-up method that efficiently removes short primers, unincorporated dNTPs, enzymes, short-failed PCR products, and salts from PCR reactions.
Isolated DNA is ready for sequencing, PCR, transcription, mapping, cloning, and labeling. We offer a wide range of Invitrogen™ PCR clean-up kits, plus the support you may need to get high yields of pure DNA.
Which PCR product clean-up kit is right for you?
Quick and Efficient removal of by-products Ideal for concentrating low yield PCR products Simple, reliable, rapid method, 96-well format Rapid, scalable, magnetic bead format
Order now Order now Order now Order now
  PureLink® PCR Purification Kit PureLink® PCR Micro Purification Kit PureLink® 96 PCR Purification Kit ChargeSwitch® PCR Clean-Up Kit
Format Spin/vacuum column Spin/vacuum column 96-well plate (vacuum/centrifuge) Magnetic beads
Protocol time 15 min 15 min 15 min 5 min
Sample volume 50–100 µL 50–100 µL 50–100 µL 25–50 µL
Yield plasmid DNA Up to 40 µg Up to 20 µg Up to 20 µg Up to 30 µg
DNA fragment size 100 bp–15 kb 100 bp–15 kb 100 bp–15 kb 90 bp–40 kb
Binding capacity Up to 40 µg Up to 40 µg Up to 40 µg ~25 µg per mg beads
Downstream Application Sequencing, Sequencing (Next Generation), Nucleic Acid Labeling, PCR, Cloning Sequencing, Nucleic Acid Labeling, PCR, Cloning Sequencing, Cloning Sequencing, Sequencing (Next Generation), Microarray Analysis, PCR, Cloning
High Throughput Compatibility No No Yes Yes
Pack size 50 preps 250 preps 50 preps 250 preps 4 plates 100 preps 960 preps
Cat. No. K310001
K310002
K310050
K310250
K3100-96A CS12000
CS12000-10
Perform both PCR purification and gel extraction?
If so, consider our PureLink® combo kit that allows you to do either process with a single kit.
  PureLink® Quick Gel Extraction and PCR Purification Combo Kit  
For Research Use Only. Not for use in diagnostic procedures.