Plasmid Isolation
Successfully isolate quality plasmid DNA. Choose from our wide range of high-performing, cost-effective plasmid purification technologies designed to help you overcome common plasmid prep issues, such as low recovery and the presence of impurities.
Prep sizes and purity grades are available to suit every application.
Which plasmid DNA isolation technology is right for you?
  Rapid isolation of molecular biology quality Gold standard for ultrapure, high quality Enhanced purity for sensitive applications
Plasmid  DNA purification kit GeneJET PureLink HiPure PureLink Expi Endotoxin-Free
Purity grade Molecular Transfection Advanced transfection
Prep size Mini–Maxi Mini–Giga Maxi, Mega, Giga
Total protocol time 15–60 min 30–120 min 90–120 min
Yield up to 20 µg–1 mg 20 µg–15 mg 1.5–5 mg
Endotoxin Standard (>10 EU/µg) Low endotoxin (0.1–1 EU/µg) Endotoxin-free (< 0.1 EU/µg)
Technology Silica membrane Anion exchange resin Anion exchange membrane
Applications Not sensitive
  • PCR
  • Cloning (digestion, ligation)
  • Sequencing
Sensitive
  • Transfection of standard, robust cell lines
  • All molecular biology applications
Very sensitive
  • Primary and stem cell transfection
  • Gene therapy and vaccine (in vivo) research
  • All standard transfection applications
  • All molecular biology applications
  Order now Order now Order now
Prep sizes
  Miniprep Midiprep Maxiprep Megaprep Gigaprep
Overnight bacterial culture volume 1–5 mL 25–100 mL 100–500 mL 500 mL–2.5 L 2.5–5 L
Approximate Yield Up to 40 µg Up to 200 µg 750–800 µg Up to 4 mg Up to 12 mg
Resources
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
DNA Purification Brochure
RNA Isolation & Purification Brochure