Starting with high-quality plasmid DNA is critical to your workflow.
The table below will help you choose the right plasmid DNA miniprep kit.
Which plasmid DNA miniprep kit is right for you?
Quick & cost effective Most widely accepted protocol for transfection Rapid purification with integrated lysate clarification filter Rapid, high-throughput transfection-grade purification
Order now Order now Order now Order now
  GeneJET Plasmid Miniprep Kit PureLink™ HiPure Plasmid Miniprep Kit PureLink™ Pro Quick96 Plasmid Kit PureLink™ 96 HQ Mini Plasmid DNA Purification Kit
Purity grade Molecular Transfection Molecular Transfection
Total protocol time < 14 min < 120 min 45 min 40 min
Yield Up to 20 µg Up to 30 µg Up to 15 µg Up to 60 µg
Throughput No No No Yes
Technology Silica membrane Anion-exchange gravity-flow column Silica-based spin-vacuum plates Silica-based spin-vacuum plates
Prep size 50, 250 preps 25, 100 preps 4 x 96, 24 x 96 preps 4 x 96 preps
Resources
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
DNA Purification Brochure
RNA Isolation & Purification Brochure