The GelQuant™ Express Analysis Software and GelCapture™ Acquisition Software applications integrate seamlessly with the E-Gel® Imager system to provide the researchers with all the applications, functions and tools for analyzing gel captured images and data. Both software applications are included with each E-Gel® Imager System.
E-Gel® GelQuant™ Express Analysis Software—Simplifying 1D Gel Analysis
E-Gel® GelQuant™ Express analysis software is a Microsoft® Windows® application for the analysis of 1D gels. This easy-to-use, high-precision software enables accurate, consistent, and fast analysis of all gel images (Figure 1).
  • Easy and fast quantitation: automatic detection and analysis of bands with just one click. Band boundaries are automatically and accurately detected using an innovative boundary detection algorithm. Even distorted gel images can be analyzed, and molecular weight calculated, based on marker lane standards.
  • Accurate analysis: automatic deduction of background noise allows accurate scientific quantitation and normalization of bands.
  • Flexible analysis tools: dynamic graphic intensity analysis tools allow easy adjustment to band and lane boundaries for complex results (e.g., degraded, doublet bands).
  • One-click data export: customize results with one-click export to a Microsoft® Excel® worksheet or as a jpeg image.
E-Gel® GelQuant™ Express software tutorial
E-Gel® GelQuant® Express data
Figure 1. Easy and fast quantitation. Analyses of gel images with E-Gel® GelQuant™ Express include (A) Multitiered function analysis, (B) Molecular weight analysis, and (C) Lane profile histogram.
GelCapture™ Acquisition Software
E-Gel® GelCapture™ software is the industry standard image acquisition software, designed for effective user-friendly gel imaging. GelCapture™ software enables real-time sample imaging and direct camera-to-PC image transfer to prevent loss of resolution, all while providing the best image quality in the least time.
  • User-friendly interface ensures all necessary commands are easily accessible
  • Up to 10 standard presets for common applications, all with click-of-a-button simplicity
  • Real-time image capturing
  • Preview application option confirms image focus for sharp, clear results
E-Gel® GelCapture™ software tutorial
Real-Time Imaging and Analysis
In combination with E-Gel® iBase™ Power System, the multiple image acquisition option is an innovative image capture function that allows real-time visualization and image capture of E-Gel® agarose gels.
E-Gel® Real-Time Imaging & Analysis
Registration and Software Updates
Register your E-Gel® Imager to download the latest software updates for the E-Gel® Imager Compact Agarose Gel Imaging System.
GelCapture™ Software
Note: Please make sure that you download the correct version of the GelCapture software based on the serial number of your instrument.
Version 1.7 is the latest version of the E-Gel® Imager acquisition software for camera hoods with serial numbers that begin with the number 1 (e.g. 1502382)
Version 2.2 is the latest version of the E-Gel® Imager acquisition software for camera hoods with serial numbers that begin with the letter N (e.g. N1500065)
GelQuant Express™ Software
Version 1.14.6 is the latest version of E-Gel® Imager Analysis software. Requires E-Gel® Imager Quantitation Software USB Dongle.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.