Owl Vertical Gel Electrophoresis Systems
Perform homogeneous or gradient polyacrylamide gel electrophoresis of nucleic acids or proteins using rigs, casters, plates, spacers, combs, blotters, power supplies, and other accessories of Thermo Scientific™ Owl™ Vertical Gel Electrophoresis Systems. Choose among the following format series: sequencing (S3S-S4S, SGC), midi-to-mini (P7-P10, JGC, P-CST), blotters (HEP, VEP).
Owl Sequencing Gel Systems, Casters, Accessories, and Parts
Thermo Scientific™ Owl™ S3S (wide) and S4S (narrow) sequencing gel rigs, SGC gel casting system, and their accessories enable polyacrylamide gel electrophoresis for the purpose of DNA sequencing. Each sequencer includes an aluminum heat sink, and well-designed upper and lower buffer chambers to minimize buffer volume and facilitate easy clean-up.
Owl P-Series Vertical Gel Systems, Casters, Accessories, and Parts
Thermo Scientific™ Owl™ P-Series gel systems, JGC casters and related accessories enable casting and running of single-percent and gradient polyacrylamide gels for nucleic acid or protein electrophoresis. Choose single or double gel (DS) systems for mini (10 x 10 cm) to midi (20 x 20 cm) gel sizes.
Owl Blotting, Staining, and Gel Drying Systems
Select Thermo Scientific™ Owl™ electrophoresis devices, accessories, and supplies for wet or semi-dry blotting, staining, or drying and storing of polyacrylamide gels.
Owl Electrophoresis Power Supplies
Select Thermo Scientific™ Owl™ power supplies for horizontal (agarose) or vertical (polyacrylamide) gel electrophoresis systems of various kinds, whether tanks, chambers, sequencers, or blotters.
Owl online warranty card registration
Thermo Scientific Owl products provide optimal laboratory throughput and efficiency for gel electrophoresis.
Register your product online for the Owl Warranty.
Complete form
Custom Ordering
Request custom sequencing combs, protein combs, plates, and spacers to meet your electrophoresis and radiation safety requirements.
Download our custom order forms