Related Hepatic Cell Culture Media
Dulbecco’s Modified Eagle Medium (DMEM), low glucose, without phenol red, without glutamine for short-term maintenance of primary hepatocyte cultures. 
Hepatozyme-SFM is a serum-free, phenol red-free medium for the long-term maintenance of hepatocyte phenotypic expression, including active and inducible forms of cytochromes P450 and active phase II enzymes.
Related Hepatic Specialty Reagents
Liver Digest Medium is a collagenase-dispase medium for dissociation of viable liver cells.
Liver Perfusion Medium is a buffered, balanced salt solution formulated to cleanse the liver of blood, prevent clotting, and initiate loosening of cell-to-cell contact.
Related Matricies
AlgiMatrix™ 3D Culture System is an animal-free bioscaffold for the development of cell culture models that are more predictive of disease states and drug responses.