See how Jump-In™ and BacMam technologies can be used to save time in your cell engineering projects.
The task of generating cell lines, parental cell lines, high throughput-ready cells and other tools for your cell-based discovery projects can be greatly simplified with custom cell engineering tools from Life Technologies.
Complete your cell engineering projects in far less time by collaborating with our Custom Biology Services team.  Whether your research requires engineered or Jump-In™ parental cell lines; BacMam virus particles or validated, high-throughput screening-ready cells in large-scale, Life Technologies has the in-house expertise to support and accelerate your cellular projects.
Send us an inquiry to start your project
Jump-In™ Parental Cell Lines
Access stable, isogenic mammalian cell lines faster with the Jump-In™ cell engineering platform.
Whether you are an experienced cell engineer, or need engineered cells for your projects, the Jump-In™ cell engineering platform can accelerate your projects by enabling you to generate engineered cell lines in much less time than traditional methods. Jump-In™ technology helps accelerate stable cell line development due to the targeted integration of genetic material into specific integration sites using R4 integrase.
Depending on your needs and experience, we offer both do-it-yourself and outsourced options for accessing Jump-In™ technology, allowing you to choose the method that best utilizes your available resources and fits your desired timelines.
For more information on our stable cell line generation custom services, submit an online inquiry.
BacMam Virus Production
BacMam-enabled Cellular Assays are an efficient, easy-to-optimize, and robust method for interrogating specific signal transduction events in a cell background of choice.  BacMam-enabled cellular assays offer several key advantages.
  • Efficient transduction of mammalian cell lines, including primary cells and stem cells
  • Safety (non-replicating in mammalian cells) and lack of observable cytopathic effect
  • Frozen storage of pre-transduced cells generates assay-ready cells
Interested in custom BacMam virus generation or building an assay for your target of interest? Custom project scopes can range from creating a BacMam virus from your gene of interest to full assay development and batch transduction of your BacMam virus into your cell line of choice.
For more information on custom BacMam projects, submit an online inquiry or email us.
Cell Provisioning Services
Obtain validated, high-throughput screening–ready cells in scale more efficiently with our cryopreservation service.
Our standardized, high-quality process yields validated cell lines typically within two to four weeks.
The process is simple:
  1. Provide us your cell line or purchase one of our cell lines
  2. Our scientists scale-up and produce cryopreserved cells
  3. Cell lines are quality control-tested and validated
For more information on our cell provisioning services, submit an online inquiry or email us.
Custom Biology Services Team
  With many labs facing resources restraints, our Custom Biology team offers institutions a powerful resource to extend their research teams.  Wholly dedicated to developing custom tools and assays, the team has the expertise to select an assay technology from the extensive Life Technologies toolbox and use these technologies to craft tools that meet your unique requirements.
Dedicated Project Management
From your initial inquiry through every milestone of your project, you can expect exceptional service and frequent, proactive communication from a dedicated project manager.  Your project manager will help ensure all parties involved in the project are aligned on project requirements, timelines and budgets.   
For Research Use Only. Not for use in diagnostic procedures.