• Citations (5)
Description
miRNA mimics are small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity.
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
We have now released the second generation of miRNA mimics & inhibitors:
  • mirVana™ miRNA Mimics, which are more specific than their predecessors due to inactivation of the star strand by proprietary chemical modifications.
  • mirVana™ miRNA Inhibitors, which are more potent than their predecessors.
For Research Use Only. Not for use in diagnostic procedures.
Learn more about mirVana™ miRNA Mimics and Inhibitors
Note: Our first generation mimics & inhibitors, Ambion® Anti-mir™ miRNA Inhibitors & Ambion® Pre-miR™ miRNA Precursors, are still available for completing your ongoing in vitro miRNA studies, but for new studies or in vivo studies, we highly recommend our new mirVana™ miRNA Mimics & Inhibitors.
Catalog # Product Size
4464066mirVana™ miRNA Mimic5 nmol
4464067mirVana™ miRNA Mimic20 nmol
4464070mirVana™ miRNA Mimic250 nmol (In Vivo Ready)
4464084mirVana™ miRNA Inhibitor5 nmol
4464085mirVana™ miRNA Inhibitor20 nmol
4464088mirVana™ miRNA Inhibitor250 nmol (In Vivo Ready)
Search more than 2000 Ambion® miRNA Mimics and Inhibitors
Watch the video and learn how >
Meet the Inventor: Silencer® Select siRNA technology
Citations & References
Authors
Wang W., Corrigan-Cummins M., Hudson J., Maric I., Simakova O., Neelapu S.S., Kwak L.W., Janik J.E., Gause B., Jaffe E.S., Calvo K.R.
Journal
Haematologica. 2012 Apr; 97(4):586-94
Authors
Schaeffer V., Hansen K.M., Morris D.R., LeBoeuf R.C., Abrass C.K.
Journal
Am J Physiol Renal Physiol. 2012 Jul 1;303(1):F75-82
Authors
Barnes N.A., Stephenson S., Cocco M., Tooze R.M., Doody G.M.
Journal
J Immunol. 2012 Jul 1;189(1):253-60
Authors
Migliore C., Martin V., Leoni V.P., Restivo A., Atzori L., Petrelli A., Isella C., Zorcolo L., Sarotto I., Casula G., Comoglio P.M., Columbano A., Giordano S.
Journal
Clin Cancer Res. 2012 Feb 1;18(3):737-47.
Authors
Sokolov M.V., Panyutin I.V., Neumann R.D.
Journal
PLoS One. 2012;7(2):e31028
Help Center
Order Support
Contact Order & Web Support, Search FAQs, View Order History and Track Your Order
Technical Support
Contact a Technical Application Scientist, Search Product Documents & FAQs and Instrument Maintenance & Repair Requests