Dependable solutions are critical when time and expense mean everything. This tool has been designed to provide the information you need when selecting a biological safety cabinet (BSC) or when you are in the critical planning stages of designing a new lab. Use the tabs below to access resources that will aid in your decision-making process, including brochures, technical studies, certifications, and site preparation forms.
Information within does not include products and certifications for China.
Start planning now ›
Our Thermo Scientific™ biological safety cabinets are designed to maximize sample protection and operator safety. Discover why laboratories around the globe rely on our BSCs every day.
Learn how our team can assist you through the lab-planning process with technical and application guidance.
BSC Overview Guide (ENESDEFR)
Containment
  • SmartFlow™ dual motor design automates airflow compensation and balancing
  • Digital airflow verification (DAVe) alerts user to compromised containment
  • H14 HEPA filters remove airborne contaminants
Comfort
  • Reduced noise level enhances user focus 
  • 10° sloped front promotes exceptional comfort and ergonomics 
  • Spacious work area includes comfortable armrest
Convenience
  • SmartClean™ Plus hinged front window design for easy cleaning 
  • Intuitive control panel for at-a-glance monitoring 
  • Motorized frameless window sash ensures safe sash height at the push of a button 
  • Programmable cross beam UV light extends bulb life and saves energy
Efficiency
  • Dual DC motors provide operation cost savings and low heat emission 
  • Night set-back mode reduces airflow to 30% when window is closed
To obtain any of the below information, please contact us and get the downloadable links at your finger tips immediately.
Available site prep forms: 0.9 Site Prep Forms |1.2 Site Prep Forms | 1.5 Site Prep Forms | 1.8 Site Prep Forms
EN 12469 Certified Products 
  • Herasafe KSP Class II, Biological Safety Cabinets
  • Maxisafe 2020, Class II Biological Safety Cabinets
  • Herasafe KS Class II, Biological Safety Cabinets
  • Safe 2020, Class II Biological Safety Cabinets
  • MSC-Advantage, Class II Biological Safety Cabinets
  • 1300 Series Class II, Type A2 Biological Safety Cabinets 
  • 1300 Series Class II, Type B2 Biological Safety Cabinets
Start planning now ›
To obtain any of the below information, please fill out the form above and get the downloadable links at your finger tips immediately.
1300 Series A2, Class II Type A2 Biological Safety Cabinets
  • 3 foot drawings
  • 4 foot drawings
  • 5 foot drawings
  • 6 foot drawings
Herasafe KS Class II Biological Safety Cabinets
  • 9R drawings
  • 12R drawings
  • 15R drawings
  • 18R drawings
1300 Series Class II Type B2 Biological Safety Cabinets
  • 4 foot drawings
  • 6 foot drawings
Stat planning now ›
Not all BSCs are manufactured to the same safety certification standards. Our design philosophy is grounded in compliance with current worldwide safety standards. Thermo Scientific biological safety cabinets are subjected to the following certifications:
biological-safety-cabinets-certifications
Click the links below to obtain a copy of your desired certificate.
CE certificates
  • 1300 Series A2, Class II Biological Safety Cabinet
  • 1300 Series B2, Class II Biological Safety Cabinet
  • Herasafe KSP, Class II Biological Safety Cabinet
  • Maxisafe 2020, Class II Biological Safety Cabinet
  • Herasafe KS, Class II Biological Safety Cabinet
  • Safe 2020, Class II Biological Safety Cabinet 
  • MSC-Advantage, Class II Biological Safety Cabinet
TUV certificates
Herasafe KSP Class II, Biological Safety Cabinets
  • 9R TUV Certificate
  • 12R TUV Certificate
  • 15R TUV Certificate
  • 18R TUV Certificate
Maxisafe 2020, Class II Biological Safety Cabinets
  • 1.2 TUV Certificate
  • 1.8 TUV Certificate
Herasafe KS Class II, Biological Safety Cabinets
  • 9R TUV Certificate
  • 12R TUV Certificate
  • 15R TUV Certificate
  • 18R TUV Certificate
Safe 2020, Class II Biological Safety Cabinets
  • 1.2 TUV Certificate
  • 1.8 TUV Certificate
MSC-Advantage, Class II Biological Safety Cabinets
  • 0.9 TUV Certificate
  • 1.2 TUV Certificate
  • 1.5 TUV Certificate
  • 1.8 TUV Certificate
NSF certified models
NSF International certifies that the products appearing on this listing conform to the requirements of NSF/ANSI Standard 49—Biosafety Cabinetry: Design, Construction, Performance, and Field Certification.
Resources