flammable materials bottles caps
Designed for storage of volatiles, compounds, and solvents where a spark-free internal cabinet is required, or in hazardous environments, Thermo Scientific flammable material and explosion-proof refrigerator-freezer combination units help you meet your safety requirements. Plus, our refrigerator-freezer combo units will satisfy your temperature, reliability, performance, and compliance needs. All flammable material refrigerator-freezers and explosion-proof refrigerator-freezers meet NFPA and OSHA standards. Availability may be limited in certain regions.
Quick links
Flammable material storage (FMS) refrigerator-freezers
These refrigerator-freezer combo units are designed to be used for the storage of volatiles, compounds, and solvents where a spark-free internal cabinet environment is required. Household/commercial refrigerators and freezers are not suitable; they have components in their electrical and refrigeration systems that can trigger explosions of flammable air-vapor mixtures inside the unit.
  10 cu. ft. FMS Refrigerator-Freezer
  TS FMS freezer fridge combo
  Order online
  10FCEETSA;
115V/60Hz
Availability North America only
Compliance UL, NFPA1, OSHA1
Temperature range Refrigerator: 1° to 12°C
freezer: –12° to –20°C
Capacity 10.1 cu. ft. (286L)
Plug type NEMA 5-15P
User interface Exterior thermostat
Defrost Refrigerator: automatic; freezer: manual
Security None
Interior cabinet
ABS plastic
Interior refrigerator (DxWxH) 18 x 20 x 38 in.
(46 x 50 x 97cm)
Interior freezer (DxWxH) 11 x 18 x 18 in.
(29 x 46 x 46cm)
Exterior (LxWxH) 28 x 24 x 60 in.
(76 x 60 x 152cm)
Shipping weight 150 lb. (68kg)
1Products complying with the applicable requirements of NFPA 70, "National Electric Code", NPFA 45, "Fire Protection for Laboratories Using Chemicals," and NFPA 99 "Health Care Facilities" are considered to meet the OSHA guidelines for these types of facilities. These units have no internal electrical components that could trigger an explosion or fire of flammable materials inside the unit. Flammable material storage units are not for use in classified hazardous locations which require the use of explosion-proof units.
Top
Explosion-proof refrigerator-freezers
Spark-free interiors and exteriors reduce the risk of explosion. These lab refrigerator-freezer combination units are designed to be used in Class 1, Groups C and D, and Class 1, Zone 1, Group IIB areas where ignitable materials are normally present. Thermo Scientific explosion-proof laboratory refrigerator-freezers are particularly suited for safe storage in hazardous environments.
  10.1 cu. ft. Explosion-Proof Refrigerator-Freezer
  TS explosion-proof freezer fridge combo
  Order online
  10ECEETSA;
115V/60Hz
Availability North America only
Compliance UL, NFPA1, OSHA1
Temperature range Refrigerator: 1° to 12°C
freezer: –12° to –20°C
Capacity 10.1 cu. ft. (286L)
Plug type Hardwired to prevent sparking and accidental disconnection
User interface Thermostat and compressor components in vapor-proof enclosure
Defrost Refrigerator: automatic; freezer: manual
Security Built-in key lock
Interior cabinet
ABS plastic
Interior refrigerator (DxWxH) 18 x 20 x 38 in.
(46 x 50 x 97cm)
Interior freezer (DxWxH) 11 x 18 x 18 in.
(29 x 46 x 46cm)
Exterior (LxWxH) 28 x 24 x 60 in.
(76 x 60 x 152cm)
Shipping weight 150 lbs. (68kg)
Top
Is there a safer way to store potentially flammable, pharmaceutical-grade materials that require refrigerated or frozen temperatures?
Thermo Scientific FMS high-performance refrigerators and freezers meet industry protocol for the storage of critical, potentially flammable, pharmaceutical-grade materials or samples containing trace amounts of flammables that could cause a spark.
Learn more
Top
Resources
Flammable material storage
Explosion proof storage
Support