floor-model-centrifuges
Performance simplified at every turn
Set the standard for quality and productivity in the lab with our floor model centrifuges. For high-speed or high-capacity sample processing, our advanced superspeed, ultracentrifuges, micro-ultracentrifuges and large capacity centrifuges meet your critical research needs.  
Quick links
Which type of floor model centrifuge fits your needs best?
Large Capacity Centrifuges
High Speed Centrifuges
Ultracentrifuges
Especially ideal for high-throughput blood banking and bioprocessing applications.
  • Enhanced capacity plus an ergonomic design
  • Flexibility to select higher capacity rotors or match existing workflows
High speed superspeed centrifuges with maximized capacity up to 6 liters for application flexibility.
  • High speed separations ideal for shared lab settings
  • Fiberlite carbon fiber rotors plus Auto-Lock rotor exchange and Auto-ID
Specially designed for high-performance applications in cellular biology, drug discovery, nanotechnology and others.
  • Small footprint ideal for space-challenged labs
  • Fiberlite carbon fiber rotors for unequalled durability and performance
Max capacity
16 x 500mL blood bags or
8 x 2000mL bottles
6 x 1000mL bottles
24 x 1.5mL microtubes to 6 x 250mL bottles
Max RCF 8,500 x g 100,605 x g 1,048,680 x g
Max speed 5,400rpm 29,000rpm 150,000rpm
Rotors 11 optional rotors 19 optional rotors 37 optional rotors
Dimensions (H x W x D) Lid closed:
39.9 x 32.1 x 35.4 in.
(101.5 x 81.6 x 90.0cm)
Lid closed:
36.6 x 27.6 x 31.7 in.
(93 x 70 x 80.5cm)
Lid closed:
34.6 x 31.1 x 27.2 in.
(88 x 79 x 69cm)
Display type Centri-Touch touchscreen interface Touchscreen interface Touch screen LCD panel
Performance range Speed 300-5,400rpm
RCF 29-8,500 x G
Speed 500-29,000rpm
RCF 7,068-100,605 x G
1,000 to 150,0000rpm
RCF 201,046 to 1,048,680 x G
Weight 230V: 1047.2 lbs. (475kg)
400V: 1091.3 lbs. (495kg)
644 lbs. (293kg) 859.8 lb. (390kg)
  Most popular:
Sorvall BP 8 and 16 Blood
Banking Centrifuge ›
Most popular:
Sorvall LYNX Superspeed
Centrifuge Series ›
Most popular:
Sorvall MTX 150
Micro-Ultracentrifuge ›
  View all View all View all
Need a smaller format floor model?
We also have portable, floor-standing units in our benchtop centrifuges category.
Compact floor-standing benchtop centrifuges ›
  • 6 x 50mL up to 4 x 145mL bottles
  • Speeds up to 17,850rpm
  • RCF up to 30,279 x g
High performance floor-standing benchtop centrifuges ›
  • 6 x 250mL up to 4 x 1000mL bottles
  • Speeds up to 15,200rpm
  • RCF up to 25,830 x g
Top
Centrifuges for Bioprocessing
Our centrifuges for bioprocessing are built to help ensure processing safety, quality control and traceability for GMP compliance. They can be used specifically for large-volume processing—up to 8 x 2 L bioprocessing bottles per run at speeds up to 8,500 x g—and can be easily used in a variety of applications, including density gradient separations, cell harvesting and vaccine production.
Learn more
Top
Sorvall LYNX Superspeed Centrifuge Series Brochure
Experience powerful technologies with breakthrough simplicity. Multiple users. A variety of processing requirements. A range of user experience. To overcome these everyday challenges of a shared laboratory setting choose the Thermo Scientific Sorvall LYNX superspeed centrifuge. The result? Productivity, safety, and reliability simplified.
  • Cellular biology
  • Diagnostic and clinical biology
  • Genomics and molecular biology
  • Proteomics
  • Virus and nanotechnology
  • Research and drug discovery
  • Bioprocessing
Learn more
Top
Featured floor model centrifuge videos
Top
Related products for floor model centrifuges
Benchtop centrifuges
Microcentrifuges
Centrifuge rotors
Related-centrifuge-rotors-175x170
Centrifuge ware
Resources
Support
Unity Lab Services 
Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.