Why settle for high speed when you can have the powerful technology and beakthrough simplicity of a Superspeed?
Our superspeed centrifuges simplify performance at every turn with up to 100,000 x g, maximized capacity up to 6 liters, and an extensive rotor portfolio, including Fiberlite LEX rotor designs, supporting labware from microplates, to microtubes, to 1 Liter bottles. Overcome everyday challenges of a shared laboratory setting with the productivity, safety, and reliability of a Sorvall LYNX Superspeed Centrifuge.
Compare our Sorvall LYNX Superspeed Centrifuges
  Sorvall LYNX 4000 Superspeed Centrifuge Sorvall LYNX 6000 Superspeed Centrifuge
  For a wide range of cellular and subcellular separations, or any other application requiring up to 68,905 x g, in a powerful, versatile, highly reliable unit that can improve your productivity.
For large capacity and high performance, up to 100,605 x g, in a powerful unit that can perform a wide range of research applications, providing a highly reliable platform for downstream BioProcess.
Request a quote
Max capacity 4 x 1000mL 6 x 1000mL
Max RCF 68,905 x g 100,605 x g
Max speed 24,000rpm 29,000rpm
Rotors 16 optional rotors 19 optional rotors
Dimensions
(H x W x D)
Lid closed:
36.6 x 27.6 x 31.7 in.
93.0 x 70.0 x 80.5cm
Lid closed:
36.6 x 27.6 x 31.7 in.
93.0 x 70.0 x 80.5cm
Display type Touchscreen interface Touchscreen interface
Performance range Speed 500-24,000rpm
RCF 7,068-68,905 x g
Speed 500-29,000rpm
RCF 7,068-100,605 x g
Weight 622 lbs. (283kg) 644 lbs. (293kg)
Top
Rotors for Sorvall LYNX superspeed centrifuges
Superspeed rotors support a variety of labware and are available for a wide range of applications. Find the ideal high speed rotor to fit your application, from microplates, to microtubes, to 1L bottles.
  • Cellular biology
  • Diagnostic and clinical biology
  • Genomics and molecular biology
  • Proteomics
  • Virology and nanotechnology
  • Research and drug discovery
  • Bioprocessing
Auto-Lock rotor exchange
Push-button security and application flexibility.
Auto-ID instant rotor identification
Immediate rotor detection and programming.
Fiberlite carbon fiber rotors
Improved durability, ergonomics and performance.
ClickSeal biocontainment lids
One-handed certified* sample protection.
Biocontainment certification by the Public Health England, Porton Down, UK.
Top
Sorvall LYNX Superspeed Centrifuge Series Brochure
Experience powerful technologies with breakthrough simplicity. Multiple users. A variety of processing requirements. A range of user experience. To overcome these everyday challenges of a shared laboratory setting choose the Thermo Scientific Sorvall LYNX superspeed centrifuge. The result? Productivity, safety, and reliability simplified.
  • Cellular biology
  • Diagnostic and clinical biology
  • Genomics and molecular biology
  • Proteomics
  • Virus and nanotechnology
  • Research and drug discovery
  • Bioprocessing
Learn more
Superspeed centrifuge videos
Top
Related products for high speed centrifuges