Sorvall Superspeed Centrifuges and Ultracentrifuges
Thermo Scientific™ Sorvall™ superspeed centrifuges and ultracentrifuges
Perform high-speed separations simply in shared lab settings with the Thermo Scientific Sorvall LYNX™ Superspeed Centrifuge Series, equipped with advanced rotor innovations that shorten run set-up time and increase rotor security. 
Thermo Scientific Sorvall ultracentrifuges allow you to maximize productivity with impressive acceleration rates that reach up to 150,000rpm in just 80 seconds. These ultracentrifuges combine cutting-edge technology, high-speed performance, versatile rotor capacities, and a compact footprint.
Thermo Scientific rotors are available for a wide range of processing needs, supporting labware from microplates, to microtubes, to 1 Liter bottles to address separation needs across:
  • cellular biology
  • diagnostic and clinical biology
  • genomics and molecular biology
  • proteomics
  • virology and nanotechnology
  • research and drug discovery
  • bioprocessing
Featured Sorvall superspeed centrifuge and ultracentrifuge products
Perform high-speed separations simply in shared lab settings with the Thermo Scientific Sorvall LYNX Superspeed Centrifuge Series. Advanced rotor innovations including Thermo Scientific Auto-Lock™ rotor exchange, Thermo Scientific Auto-ID instant rotor identification and lightweight and durable Thermo Scientific Fiberlite™ carbon fiber rotors shorten run set-up time and increase rotor security. For real-time run monitoring and control via a smart device, download the Thermo Scientific Centri-Vue™ application.
Select from our extensive rotor portfolio of Thermo Scientific Sorvall LYNX superspeed centrifuges which offer up to 100,605 x g performance and 6 L capacity along with Thermo Scientific Fiberlite LEX rotor designs. The series supports labware—from microplates to microtubes and 1 L bottles—all with our push-button Auto-Lock rotor exchange.
Combining outstanding speed, safety and ergonomics in a compact design, our ultracentrifuges and micro-ultracentrifuges are designed to deliver exceptional performance and versatility for a variety of applications.
Our versatile ultracentrifuge rotor portfolio addresses your high-performance processing applications, accommodating tubes from 0.2 mL to 250 mL—and provides application flexibility as your lab's research focus evolves.
Featured superspeed centrifugation videos
Support
  • Unity Lab Services:
    Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.