Ultimate protection, optimum capacities
Whether you are planning for one or several ultra low–temperature (ULT) freezers in a new lab, it’s important to know how much footprint to allocate as well as electrical requirements and additional specifications. Thermo Scientific™ ultra-low freezers offer a wide range of options for labs with different footprint requirements as well as new models that are designed to meet energy-savings and sustainability preferences.
Our Thermo Scientific upright ultra-low freezers are designed to protect your samples and maximize sample capacity within the freezer footprint. Choose from 5-inner door models (Revco™ freezers) or 4-inner door (TSX Series, Forma™ freezers).
  • Our new TSX Series –86°C freezers offer outstanding energy savings and sustainable design features. Models hold up to 600 2-inch cryo boxes.
  • Revco UxF and Forma 88000 Series –86°C models feature a touch-screen user interface and capacities of 300, 400, 500, 600, and 700 boxes.
  • Revco ExF and Forma 900 Series –86°C models feature microprocessor controls and capacities of 240, 320, 400, and 600 boxes.
  • Revco DxF and Forma 7000 models offer –40°C temperatures, microprocessor controls, and capacities of 240, 320, and 400 boxes.
Combined technical data sheets and drawings are available in PDF form for download. Choose from 5-inner door upright freezers (Revco) or 4-inner door upright (TSX, Forma). Model numbers ending in A are 120 V, 60 Hz; D models are 208–230V, 60 Hz; and V models are 230 V, 50 Hz. Models with a WC at the end include water-cooled condensers. CAD and Revit format drawings will be available on this site in the near future; consult your sales representative for immediate needs for CAD and Revit.
Access drawings now ›
  • Thermo Scientific Forma 995
  • Thermo Scientific Forma 7240A
  • Thermo Scientific Forma 7240D
  • Thermo Scientific Forma 7240V
  • Thermo Scientific Forma 7320A
  • Thermo Scientific Forma 7320D
  • Thermo Scientific Forma 7320V
  • Thermo Scientific Forma 7400A
  • Thermo Scientific Forma 7400D
  • Thermo Scientific Forma 7400V
  • Thermo Scientific Revco UxF30086A
  • Thermo Scientific Revco UxF30086AWC
  • Thermo Scientific Revco UxF30086D
  • Thermo Scientific Revco UxF30086DWC
  • Thermo Scientific Revco UxF30086V
  • Thermo Scientific Revco UxF30086VWC
  • Thermo Scientific Revco UxF40086A
  • Thermo Scientific Revco UxF40086AWC
  • Thermo Scientific Revco UxF40086D
  • Thermo Scientific Revco UxF40086DWC
  • Thermo Scientific Revco UxF40086V
  • Thermo Scientific Revco UxF40086VWC
  • Thermo Scientific Revco UxF50086A
  • Thermo Scientific Revco UxF50086AWC
  • Thermo Scientific Revco UxF50086D
  • Thermo Scientific Revco UxF50086DWC
  • Thermo Scientific Revco UxF50086V
  • Thermo Scientific Revco UxF50086VWC
  • Thermo Scientific Revco UxF60086A
  • Thermo Scientific Revco UxF60086AWC
  • Thermo Scientific Revco UxF60086D
  • Thermo Scientific Revco UxF60086DWC
  • Thermo Scientific Revco UxF60086V
  • Thermo Scientific Revco UxF60086VWC
  • Thermo Scientific Revco UxF70086D
  • Thermo Scientific Revco UxF70086DWC
  • Thermo Scientific Revco UxF70086V
  • Thermo Scientific Revco UxF70086VWC
  • Thermo Scientific Revco ExF24086A
  • Thermo Scientific Revco ExF24086D
  • Thermo Scientific Revco ExF24086V
  • Thermo Scientific Revco ExF32086A
  • Thermo Scientific Revco ExF32086D
  • Thermo Scientific Revco ExF32086V
  • Thermo Scientific Revco ExF40086A
  • Thermo Scientific Revco ExF40086D
  • Thermo Scientific Revco ExF40086V
  • Thermo Scientific Revco ExF60086D
  • Thermo Scientific Revco ExF60086V
  • Thermo Scientific Forma 989
  • Thermo Scientific TSX600D and TSX600V
  • Thermo Scientific Forma 88300A
  • Thermo Scientific Forma 88300AWC
  • Thermo Scientific Forma 88300D
  • Thermo Scientific Forma 88300DWC
  • Thermo Scientific Forma 88300V
  • Thermo Scientific Forma 88300VWC
  • Thermo Scientific Forma 88400A
  • Thermo Scientific Forma 88400AWC
  • Thermo Scientific Forma 88400D
  • Thermo Scientific Forma 88400DWC
  • Thermo Scientific Forma 88400V
  • Thermo Scientific Forma 88400VWC
  • Thermo Scientific Forma 88500A
  • Thermo Scientific Forma 88500AWC
  • Thermo Scientific Forma 88500D
  • Thermo Scientific Forma 88500DWC
  • Thermo Scientific Forma 88500V
  • Thermo Scientific Forma 88500VWC
  • Thermo Scientific Forma 88600A
  • Thermo Scientific Forma 88600AWC
  • Thermo Scientific Forma 88600D
  • Thermo Scientific Forma 88600DWC
  • Thermo Scientific Forma 88600V
  • Thermo Scientific Forma 88600VWC
  • Thermo Scientific Forma 88700D
  • Thermo Scientific Forma 88700DWC
  • Thermo Scientific Forma 88700V
  • Thermo Scientific Forma 88700VWC
  • Thermo Scientific Forma 903
  • Thermo Scientific Forma 909
  • Thermo Scientific Forma 902
  • Thermo Scientific Forma 904
  • Thermo Scientific Forma 910
  • Thermo Scientific Forma 905
  • Thermo Scientific Forma 905 WC
  • Thermo Scientific Forma 956
  • Thermo Scientific Forma 957
  • Thermo Scientific Forma 906
  • Thermo Scientific Forma 908
  • Thermo Scientific Forma 907
  • Thermo Scientific Forma 907 WC
  • Thermo Scientific Forma 992
  • Thermo Scientific Forma 986
  • Thermo Scientific Forma 991
  • Thermo Scientific Forma 993
  • Thermo Scientific Forma 988
  • Thermo Scientific Forma 994
  • Thermo Scientific Forma 990
Access Revit files ›
All 120 V, 60 Hz ultra-low freezers are UL, and 230 V, 50 Hz are CE marked. Contact your Thermo Fisher Scientific representative for more details.
We can support your new construction or remodeling project in all stages of the process, including:
  • Drawings, equipment specifications, technical data, or white papers
  • New construction managers and local technical specialists to provide support and training
  • On-site trainings and seminars to meet your schedule
  • Dedicated service support for installation, training, and product certification
  • AIA presentations (qualify for CEU credits)
  • Local distributor network