Meters from economical and simple to robust and modular
High-quality meters are essential to the critical, routine tasks you perform every day. Our portfolio of benchtop and portable dissolved oxygen meters are designed to help you meet your sample analysis challenges with speed and confidence.
Meet your regulatory needs with technology that is ideal for a wide range of applications and advanced dissolved oxygen analysis in the lab. Orion advanced and premium benchtop meters offer the flexibility to measure both polargraphic and optical technology. A dedicated kit, complete with BOD stirrer probe needed to fit in a DO bottle for airtight incubation, is available with the Orion Star A213 meter.
Calibrate using water-saturated air, air-saturated water, or custom value, using Winkler titration or zero point options. Log up to 2,000 data point sets with time/date stamp, and easily transfer calibration and data logs via USB or RS232 to a printer or computer when using an Orion advanced or premium dissolved oxygen meter. 
Premium
Advanced
Basic
  Orion Versa Star Pro RDO/DO
(VSTAR30 series)
Orion Star A213 RDO/DO
(STARA2130 series)
Orion Star A113 DO
(STARA1130 series)
  Premium Orion benchtop  dissolved oxygen meters Advanced Orion benchtop dissolved oxygen meters Basic Orion benchtop dissolved oxygen meters
  Versatile, high-performance laboratory measurement system  Easy-to-use, dependable laboratory meters Economical, simplified
laboratory meters
Ideal for
applications in:
Pharmaceuticals, biotechnology, food and beverage, industrial Water, 
wastewater,
chemical, contract testing
Academic, 
environmental, 
power
Compatible probes  RDO, polarographic RDO, polarographic  Polarographic 
DO range (polarographic)  0 to 90 mg/L, 0 to 600%  0 to 90 mg/L, 0 to 600%  0 to 20 mg/L, 0 to 200% 
DO range (RDO optical)  0 to 50 mg/L, 0 to 500%  0 to 50 mg/L, 0 to 500%  --- 
DO calibration modes  Water-saturated air, air-saturated water, manual (Winkler), zero point  Water-saturated air, air-saturated water, manual (Winkler), zero point  Water-saturated air, manual (Winkler) 
Barometric pressure compensation  Automatic with built-in barometer  Automatic with built-in barometer  Automatic with manual value input 
Salinity correction  Automatic with conductivity module and probe or manual input  Automatic with manual factor input  Automatic with manual factor input 
Display type Adjustable color graphic LCD Graphic LCD Fixed segment LCD
Backlit display   Yes  Yes  --- 
Onscreen text prompts Step-by-step calibration and guided setup instructions Step-by-step calibration and guided setup instructions Basic calibration and setup prompts
Keypad Numeric with menu-specific function keys Comprehensive with menu-specific function keys Simple with highlighted measure key
Number of channels  4, customizable with modules 1 1
Included modules 1 x VSTAR-RD --- ---
Stirrer probe inputs 2 1 ---
Measurement read modes Auto-read, continuous, timed, single-shot Auto-read, continuous, timed Auto-read, continuous
Methods 10 per channel 10 ---
Data log 2,000 with date/time 2,000 with date/time 50
Calibration log 30 per mode 10 1
Identification options Electrode ID, sample ID, user ID Sample ID, user ID ---
Alarms Set point, high/low limit, calibration due High/low limit, calibration due ---
External device output USB (2), RS232 USB, RS232 ---
Certifications IP-54, CE, TUV 3-in-1, FCC Class A IP-54, CE, TUV 3-in-1, FCC Class A IP-54, CE, TUV 3-in-1, FCC Class A
Power supply Universal, 1000150–240 VAC, 50–60 Hz (Cat. No. STARA-PWR) Universal, 100–240 VAC, 50–60 Hz (Cat. No. 1010003) or 4 AA batteries  Universal, 100–240 VAC, 50–60 Hz (Cat. No. 1010003) or 4 AA batteries 
  Enhance your productivity with accessories for your benchtop and portable meters. Meter Accessories ›
Complete your dissolved oxygen (DO) measurements faster
We offer a range of dissolved oxygen (DO) measurement instrumentation and accessories designed to meet your standards for performance and price. Our DO meters, DO probes, and standards are designed to offer maximum repeatability and accuracy in your DO readings.
Dissolved Oxygen (DO)
Portable Meters
Dissolved oxygen portable meters
Bring the lab outdoors with portable meters for dependable performance in the field. High-quality meters are essential to the critical, routine measurements you perform every day. We offer a wide range of Dissolved Oxygen (DO) meters for your measurement needs, performance specifications, and budgetary constraints.
Learn more  
Electrodes & Probes
dissolved oxygen electrodes and probes
Perform accurate and reproducible measurements in any application. Our wide selection of electrochemical probes, sensors, and electrodes are designed to make your complex measurements routine, reliable, and accurate—in the lab or in the field. Available in a variety of body materials, body styles, and sample requirements.
Learn more  
Buffers and Solutions
buffers and solutions
Using high-quality, fresh solutions to calibrate and maintain your electrodes, probes, and sensors is an essential part of good laboratory practices. Having fresh Thermo Scientific Orion solutions available for your routine calibration needs is the best way to ensure your data is both accurate and reproducible when accuracy is critical.
Learn more  
Resources
Support
Knowledge Base: Certificates of Analysis and SDS for Orion, ROSS, AQUAfast, AquaMate, and AquaSensors products.
Orion Meters Software and Upgrades
Thermo Scientific Orion benchtop and portable meters are designed to help deliver accurate, consistent results, day after day. Boost productivity by upgrading your software today.
Find your software