Orion buffers solutions and standards
Manufactured with quality in mind
Using high-quality solutions to calibrate and maintain your sensors is the best way to ensure your data is both accurate and reproducible. When choosing buffers and solutions for pH measurement, pay attention to the quality of the water and chemicals used. Use USP grade water whenever possible.
Thermo Scientific Orion buffers and solutions are manufactured with USP grade purified water and are traceable to NIST certified standards. They undergo rigorous quality testing throughout the process to prevent contamination before and after bottling. 
Avoid using low-quality, reconstituted, or expired buffers. These can lead to measurement errors, which require lengthy troubleshooting, or may go unnoticed. Our bottles of buffers have a safety seal to eliminate leaking during shipment and also as a tamper-proof measure.
Orion solutions, standards, and buffers brochure
Quick links:
Quality pH measurements
The quality of your measurement results depends upon the quality of your standards, buffers, and reagents. We offer a complete selection of quality-assured support solutions for virtually any electrochemistry analysis. Our pH buffers are designed to meet end-user requirements, including laboratory grade and DIN standards. Buffer temperature tables are programmed into the meter software for use with the Auto-Buffer-Recognition feature. 
Attention to quality ensures accurate and reproducible measurements:
  • USP-grade purified water
  • NIST traceability
  • Rigorous testing for accuracy and shelf life performance
  • Manufacturing in the USA under ISO 9001 standards and strict ecological controls
Electrochemistry expertise enables solution performance and longevity:
  • Solutions formulated exclusively for electrochemistry
  • Produced using ultra-pure water and chemicals
  • Strict control over manufacturing process
  • Special handling and packaging when applicable
Extensive application knowledge results in the right solution for your needs, when you need it:
  • All the solutions you will need from one high-quality source
  • Technical and applications phone support
  • Quick and timely shipments ensure measurement readiness
pH buffers and test kits
We offer pH buffers and test kits for a variety of applications. All Orion pH buffers and standards are traceable to NIST reference materials and are available in a variety of sizes ranging from 15 mL single-use pouches to large-volume 19 L cubitainers.
Orion pH Buffer
Individual Use Pouches
  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
  • Manufactured under ISO 9001 quality standards
  • Pouches are 15mL each and are available in 10 packs or 25 packs
  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Pouches are 15mL each and are available in 10 packs or 25 packs
  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Pouches are 15mL each and are available in 10 packs or 25 packs
  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Pouches are 15mL each and are available in 10 packs or 25 packs
  • Orion ORP Standards for Redox/ORP Electrodes
    • Stable shelf life without hazardous chemicals
    • Included in the new ASTM ORP method
    • Used as relative mV standard to calibrate the offset of sample
    • +220mV vs. Ag/AgCl using 4M KCl saturated with AgCl filling solution (Part No. 900011), +420mV vs. Standard Hydrogen Electrode (EH)
    Orion pH Buffer Bottles
    All Orion pH buffer bottles have pH/temperature tables printed on the labels for accurate calibrations and NIST traceability. Manufactured under ISO 9000 quality standards.
    Buffer color codes:
    • pH 4.01 buffer is pink
    • pH 5.00 buffer is orange
    • pH 7.00 buffer is yellow
    • pH 10.01 buffer is blue
    View all pH buffers test-kits ›
    Container sizes: pH buffers and ORP standards
    All Orion pH buffer bottles have pH/temperature tables printed on the labels for accurate calibrations and NIST traceability, manufactured under ISO 9000 quality standards. The buffers are available in pint (475 mL) bottles, portable five packs of 2 oz. (5 x 60 mL) bottles, convenient All-in-One buffer kits, 5 gallon (19 L) cubitainers, and single-use pouches. See the table below to identify which size buffers and standards are necessary for your applications.
      Laboratory Field Pharmaceutical Single use Food and
    beverage
    15 mL (0.5 oz.) individual use pouches > available in 10 pack or 25 pack boxes
    60 mL (2 oz.) bottle > available in 5 pack box
    475 mL (1 pint) bottle
    1 L (0.25 gallon) bottle
    4 L (1 gallon) bottle
    19 L (5 gallon) cubitainer
    810007 ROSS Filling Solutions
    pH electrode and reference electrode fill solutions
    Ensure the proper performance and function from your electrode by using our specially formulated fill solutions. Use Thermo Scientific Orion pH Electrode Filling Solution with pH electrodes and reference electrodes. Packaged as five 2 oz. (5 x 60 mL) bottles.
    Which fill solution should I use?
    Solution Description Use with electrodes
    810007 ROSS electrode fill solution, 3 M KCl, 5 x 60 mL ROSS Ultra, ROSS and Micro ROSS Electrodes
    800300800500U8102BN8102BNUWP, 8103BN, 8103BNUWP, 8104BN, 8104BNUWP, 8115BN, 8115BNUWP, 8135BN, 8135BNUWP, 815600, 8156BNUWP, 8157BNU, 8157BNUMD, 8157UWMMD, 8163BNWP, 8165BNWP, 8172BNWP, 8175BNWP, 8220BNWP, 8302BNUMD
    900011 Silver Chloride pH electrode fill solution, 4 M KCl with Ag/AgCl, 5 x 60 mL Standard, Specialty, Single Junction Refillable Green, Micro and ORP Electrodes
    9102BNWP, 9103BNWP, 9104BNWP, 9156BNWP, 9157BN, 9157BNMD, 9162BNWP, 9165BNWP, 9172BNWP, 9180BNMD, 9678BNWP, 9778BNWP, 9780SC, 9810BN, 9826BN, 9863BN, GS9156BNWP
    900004 Silver Chloride pH electrode (low level) fill solution, 2 M KCl with Ag/AgCl,
    5 x 60 mL
    Standard, Specialty, Single Junction Refillable Green, Micro and ORP Electrodes (low ionic strength samples)
    9102BNWP, 9103BNWP, 9104BNWP, 9156BNWP, 9157BN, 9157BNMD, 9162BNWP, 9165BNWP, 9172BNWP, 9180BN, 9180BNMD, 9678BNWP, 9778BNWP, 9780SC, 9810BN, 9826BN, 9863BN, GS9156BNWP
    910008-WA Double junction pH
    electrode fill solution, 3 M KCl, 5 x 60 mL
    Double Junction, Double Junction Refillable Green and KNIpHE Electrodes
    9102DJWP, 9110DJWP, 9120APWP, GD9156BNWP
    510011 No Cal pH electrode fill solution, 5 x 60 mL No Cal Electrodes
    5107BNMD, 5107NC
    610001 pHuture pH electrode fill solution, 5 x 60 mL pHuture Electrodes
    900001 Single junction reference electrode fill solution, equitransferent solution with Ag/AgCl, 5 x 60 mL Single Junction Reference Electrode
    900100
    900002 Double junction reference electrode inner fill solution, equitransferent solution with Ag/AgCl, 5 x 60 mL Double Junction Reference Electrode
    900200
    900003 Double junction reference electrode outer fill solution, 10% KNO3, 5 x 60 mL Double Junction Reference Electrode
    900200
    Perfectly designed to work together
    Orion solutions are an essential part of the entire Orion portfolio of electrochemistry measurement products. Whether you are in the lab or in the field, you can be confident that your Orion benchtop and portable meters, electrodes, accessories, and solutions will deliver the accuracy and repeatable measurements you need, every time. 
    Benchtop pH Meters
    Benchtop pH measurement
    The ideal choice for dedicated pH testing in water, process, research, and QA/QC samples. The Thermo Scientific portfolio of pH Meters offer a range from simple & economical models to powerful & sophisticated pH meters, to help you analyze your samples with confidence.
    Learn more
    Portable pH Meters
    Portable pH measurement
    Our portable pH meters are designed to offer pH testing in the field for applications including environmental, drinking water, and wastewater analysis.
    Learn more
    Electrodes and Probes 
    Electrodes for pH measurement
    An extensive assortment of quality pH electrodes and sensors are available in a variety of body materials, body styles, fill types, reference styles, and junctions to meet the needs of your sample measurements.
    Learn more
    Orion pH buffer values vs. temperature
    This useful guide illustrates how pH buffer values differ according to temperature. 
    pH buffer values vs. temperature
    Using high-quality solutions to calibrate and maintain your sensors is the best way to ensure your data is both accurate and reproducible. Learn more about why high-quality solutions deliver correct measurements every time.  
    Download
    Resources
    Support
    Knowledge Base: Certificates of Analysis and SDS for Orion, ROSS, AQUAfast, AquaMate, and AquaSensors products.
    Orion Meters Software and Upgrades
    Thermo Scientific Orion benchtop and portable meters are designed to help deliver accurate, consistent results, day after day. Boost productivity by upgrading your software today.
    Find your software