Clog free long lasting junction electrode
Clog free long lasting junction
  • The unique, free-flowing liquid-to-liquid junction offers a stable, drift-free measurement.
  • Easy-to-clean junction that avoids clogging by simply pressing the cap and flushing the junction area.  
  • Sure-Flow junction electrodes are compatible with samples containing organic compounds, proteins, heavy metals, and other compounds that react with silver such as bromides, iodides, cyanides and sulfides.
  • Silver chloride Sure-Flow electrodes can be used on tough samples that do not require a double junction electrode to prevent contact with silver.
  • ROSS Sure-Flows are also available as semi-micro pH and reference half cell electrodes.
Featured electrodes:
View all
  High performance in all samples Fast response –
even in difficult
samples
For all samples including solvents Fast response in difficult samples & soils
  ROSS Sure-Flow combo pH glass body electrodes ROSS Sure-Flow combo pH epoxy body electrodes Sure-Flow combo pH glass body electrodes Sure-Flow combo pH epoxy electrodes
  ROSS Sure-Flow combination pH electrode with glass body ROSS Sure-Flow combination pH electrode with epoxy body Sure-Flow combination pH electrode with glass body Sure-Flow combination pH electrode with epoxy body
  • Sure-Flow junction prevents clogging
  • For soil, sludge, colloids, viscous material and organic solvents
  • Ideal for soil samples
  • Durable epoxy design
  • For TRIS, sulfides and proteins
  • For soil, sludge, colloids, and viscous material
  • Chemical resistant glass body
  • Rugged, durable epoxy design
  • Has removable bulb guard
Product SKU number 8172BNWP 8165BNWP
9172BNWP 9165BNWP
pH Range 0-14 0-14 0-14 0-14
pH Precision 0.01 0.01 0.02 0.02
Temp. Range 0-100°C 0-100°C 0-100°C 0-100°C
Internal Reference ROSS ROSS Ag/AgCl Ag/AgCl
Junction Sure-Flow Sure-Flow Sure-Flow Sure-Flow
Fill Solution (Cat. No.) 3 M KCl (810007) 3 M KCl (810007) 4 M KCl w/ Ag/AgCl (900011) 4 M KCl w/ Ag/AgCl (900011)
Dimension D (Dia) L (Length) T (Tip)
L - 120 mm
D - 12 mm
L - 120 mm
D - 12 mm
L - 120 mm
D - 12 mm
L - 120 mm
D - 12 mm
Connector Type BNC Waterproof
BNC Waterproof
EDIN Waterproof
BNC Waterproof BNC Waterproof
  Increase your productivity by adding accessories to your benchtop or portable meter. Meter Accessories ›
The Sure-Flow technology has a lever that allows reference solution to be flushed from a spring-compressed junction. This enables easier clog-free maintenance when measuring difficult samples.
Sure-Flow technology junction press to release
Step 1: Press and hold top of the electrode
Sure-Flow technology spring-compressed junction to flush
Step 2: The spring-compressed junction opens to release fill solution through a ¼ inch gap
Sure-Flow technology spring-compressed junction release
Step 3: When pressure is released from the top of the electrode, the junction closes the gap to a nominal sleeve junction spacing
Sure-Flow electrodes for your parameter measurements
Ensure clog-free measurements in samples when using ion-selective electrodes (ISE) and other designs. Learn more about measuring other parameters using a Sure-Flow electrode.
Learn more
pH and ORP
benchtop meters
find the right benchtop meter for your pH and ORP measurement needs
The accuracy and reliability of your pH and ORP measurements depend on the instrumentation you use. Learn more about how you can identify the best match based on your required features, performance, specifications, and budget.
Learn more
pH and ORP
portable meters 
find the right portable meter for your pH and ORP measurement needs
Take your pH and ORP measurements to the field without sacrificing accuracy and reliability. Our portable meters are rugged and waterproof, and provide the dependable performance that your demanding applications require.
Learn more
Electrodes
and probes
view our selection of electrodes
The accuracy and reliability of your measurements depend on the instrumentation you use. Learn more about how you can identify the best match based on your required features, performance, specifications, and budget.
Learn more
Buffers
and solutions
view our buffer and solutions
We offer buffers, electrode storage solutions, electrode cleaning solutions, lab buffer kits, and electrode filling solutions for your measurement needs. Find the buffers and solutions that you need for your process.
Learn more
Support
Knowledge Base: Certificates of Analysis and SDS for Orion, ROSS, AQUAfast, AquaMate, and AquaSensors products.
Orion Meters Software and Upgrades
Thermo Scientific Orion benchtop and portable meters are designed to help deliver accurate, consistent results, day after day. Boost productivity by upgrading your software today.
Find your software