membranes-pes
The asymmetric PES (aPES) membrane provides superior flow rates, excellent downstream cleanliness, and low protein binding for cell culture applications, like media, sera, and growth. The Nalgene Rapid-Flow 0.2 micron aPES membranes are stem cell tested and recommended for fluids requiring sterilization, while the 0.1 micron membrane is ideal for mycoplasma removal, and the 0.45 micron can be used as a clarifying pre-filter.
Pore size quick reference chart
0.65/0.8μm 0.45μm 0.2μm 0.1μm
Analysis of mold, fungi and yeast, stabilization of fermented product, clarification and prefiltration Analysis of bacteria, sterility testing, clarification and prefiltration
Sterilization, sterility testing, “final filtration”
Reduction of mycoplasma, ultracleaning
aPES membrane materials reference
Application Material properties Flow rate Throughput Extractables Autoclave Protein binding
Cell culture media
Serum
Molecular biology solutions
Hydrophilic
High High Low Yes Low
Select the best aPES Membrane for your application
To find the membrane you need, select one of the column categories to sort the products. You can also use the search box. The "Select" button will direct you to the product specifications.
No records were found matching your criteria
Catalog # Pore Size Volume Format  
565-0010 0.1μm 150mL Filter Unit Select
566-0010 0.1μm 500mL Filter Unit Select
567-0010 0.1μm 1000mL Filter Unit Select
568-0010 0.1μm 250mL Filter Unit Select
298-9020 0.2μm 5000mL Bottle Top Filter Select
524-0020 0.2µm 115mL Filter Unit Select
564-0020 0.2µm 50mL Filter Unit Select
565-0020 0.2μm 150mL Filter Unit Select
566-0020 0.2μm 500mL Filter Unit Select
567-0020 0.2μm 1000mL Filter Unit Select
568-0020 0.2μm 250mL Filter Unit Select
569-0020 0.2μm 500mL Filter Unit Select
595-3320 0.2µm 500mL Bottle Top Filter Select
595-4520 0.2µm 500mL Bottle Top Filter Select
596-3320 0.2µm 150mL Bottle Top Filter Select
596-4520 0.2µm 150mL Bottle Top Filter Select
597-3320 0.2µm 1000mL Bottle Top Filter Select
597-4520 0.2µm 1000mL Bottle Top Filter Select
720-1320 0.2μm 2mL-10mL Syringe Filter Select
725-2520 0.2μm 10mL - 100mL Syringe Filter Select
124-0045 0.45μm 115mL Filter Unit Select
165-0045 0.45μm 150mL Filter Unit Select
166-0045 0.45μm 500mL Filter Unit Select
167-0045 0.45μm 1000mL Filter Unit Select
168-0045 0.45μm 250mL Filter Unit Select
169-0045 0.45μm 500mL Filter Unit Select
295-3345 0.45μm 500mL Bottle Top Filter Select
295-4545 0.45μm 500mL Bottle Top Filter Select
296-3345 0.45μm 150mL Bottle Top Filter Select
296-4545 0.45μm 150mL Bottle Top Filter Select
725-2545 0.45μm 10mL - 100mL Syringe Filter Select