Syringe filters
Thermo Scientific Nalgene syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.  
Sterile Syringe Filters
PES Membrane
SFCA Membrane
CA Membrane
Nylon Membrane
The PES membrane is the best choice for media with its lowest protein binding and very low extractables and fast flow rates.  The SFCA membrane is useful for cell culture fluids and cellular solutions with its low protein binding and lower extractables than standard CA.
The CA membrane is good for buffers, with low protein binding and extractables.
The Nylon membrane is recommended for alcohols, cryoprotectants, like DMSO glycerol and solvents, providing very low extractables and high protein binding capabilities.
Catalog #
Material
Volume
Pore Size
720-1320 PES 13mm 0.2μm Select
725-2520 PES 25mm 0.2μm Select
725-2545 PES 25mm 0.45μm Select
723-2520 SFCA 25mm 0.22μm Select
723-2545 SFCA 25mm 0.45μm Select
723-9920 SFCA 25mm 0.22μm Select
723-9945 SFCA 25mm 0.45μm Select
723-2580 CA 25mm 0.8μm Select
722-2520 GF/CA 25mm 0.22μm Select
726-2520 NYLON 25mm 0.22μm Select
726-2545 NYLON 25mm 0.45μm Select
0.2μm
724-2045
724-2045
122-0045
25mm Syringe
25mm Syringe
155-0020
155-0020
Non-Sterile Syringe Filters
SFCA Membrane
CA Membrane
NYLON Membrane
PTFE Membrane
The SFCA membrane is most commonly used for immunological samples and biological fluids The CA membrane is ideal for buffers, aqueous reagents and solutions, including EIA and ELISA samples.  The nylon membrane is good for filtration of aqueous biological solutions, alcoholic solutions and non-aqueous HPLC/GC solvents.  Ideal for filtration of solvents and aggressive acids (requires prewetting with ethanol).
Catalog #
Material
Volume
Pore Size
724-2020 SFCA 25mm 0.22μm Select
724-2045 SFCA 25mm 0.45μm Select
171-0020 CA 4mm 0.2μm Select
171-0045 CA 4mm 0.45μm Select
724-2080 CA 25mm 0.8μm Select
176-0020 Nylon 4MM  0.2μm Select
176-0045 Nylon 4MM  0.45μm Select
727-2020 Nylon 25mm 0.22μm Select
727-2045 Nylon 25mm 0.45μm Select
721-1320 PTFE 13mm 0.22μm Select
721-1345 PTFE 13mm 0.45μm Select
728-2020 PTFE 25mm 0.22μm Select
728-2045 PTFE 25mm 0.45μm Select
DS0222-0020 PTFE 50MM 0.2μm Select
DS0222-0045 PTFE 50MM 0.45μm Select
0.2μm
724-2045
724-2045
122-0045
25mm Syringe
25mm Syringe
155-0020
155-0020