Affymetrix Annotation Converter software tool enables you to create custom SQLite format annotations (annot.db) for use in Axiom™ Analysis Suite, GeneChip™ Genotyping Console (GTC) Software, and Affymetrix Power Tools (APT). You can customize annotations for Affymetrix catalog genotyping arrays or generate annotations for Axiom custom genotyping arrays such as the Axiom myDesign™ array plates. The Annotation Converter is available as a standalone application, within GTC Software, or is accessible from a Windows command-line prompt.
Download information and instructions
Technical documentation
Manuals
Other
Related products