Microarray Analysis Sample Data
Sample data is provided for evaluation and development purposes. Each demo data section will describe the contents and format of the data files.
GeneChip™ Human Transcriptome Array 2.0 data sets
The sample HTA 2.0 array data set includes four replicates of two tissue types (liver and muscle).
Exon 1.0 ST Array expression data sets
Sample data generated using the single strand amplified whole transcript protocol.
Exon 1.0 ST Array sample data from the Human, Mouse and Rat Exon 1.0 ST Arrays, including:
  • Tissue panel data sets on all three arrays
  • Tissue mixture data set on both the Human Exon 1.0 ST Array and the HG-U133 Plus 2.0 Array
  • Small colon cancer data set on the Human Exon 1.0 ST Array
Gene 1.0 ST Array sample data sets
Gene 1.0 ST Array sample data from the Human, Mouse and Rat Gene 1.0 ST Arrays, including:
  • Tissue panel data sets on all three arrays
  • Tissue mixture data set on the Human Gene 1.0 ST Array, the Human Exon 1.0 ST Array and HG-U133 Plus 2.0 Array
Gene 1.1 ST Array Strip sample data sets
Sample data generated using the Human Gene 1.1 ST Array Strips processed on the GeneAtlas™ system. The sample data set includes three replicates of four different tissue types (heart, brain, prostate and testes). Download Gene 1.1 ST Array Strip sample data sets ›
Genotyping mapping data sets
Axiom™ Exome sample data set
The Axiom Exome sample data set includes CEL files and observed genotypes for 1,249 individuals. These data are useful as reference data for GWAS or for ancestry determination, and a variety of other applications. Additional information about the Axiom Exome array can be found on the related product page. Download the sample data set ›
Axiom Genomic Database sample data
With the Axiom Genomic Database of validated SNPs, researchers can create custom arrays or array plates containing tens of thousands or several million SNPs. This broad spectrum of custom capabilities enables highly precise designs leveraging the Axiom Genomic Database of validated SNPs, including millions of variants from the 1000 Genomes Project as well as SNPs with low minor allele frequencies. Download the sample data set ›
DMET Plus sample data set
The DMET™ Plus Sample Data Set is a useful tool for software and workflow demonstrations, development of probe-level analysis methods for making genotype calls from probe intensity data, and a variety of other applications.
The data set consists of eight distinct HapMap  CEPH samples, three other genomic samples, and three synthetic plasmid samples. One CEPH sample is replicated two times.
Copy number and cytogenetics sample data on CytoScan™ Cytogenetics Suite
Several samples are available for analysis and visualization in Chromosome Analysis Suite (ChAS). These sample data sets depict typical aberrations found with CytoScan arrays, such as deletions and duplications, LOH/AOH and consanguinity, hyperploidy, mosaics, etc. Additionally, trio samples are included to be used with the Mendelian Consistency Check tool.
Tiling arrays for regulation & transcript mapping
These tiling array sample data sets leverage various tiling arrays with the CHiP-on-chip assay to identify genomic binding locations of DNA binding proteins of interest.
3' expression data sets
  • High-throughput plate data
    Zipped GCO EXP/CEL files used in performance whitepaper.
  • 3' expression sample data
    Zipped files with sample GeneChip probe array data (.exp, .dat, .cel., .dat, .rpt, files) for a variety of expression and genotyping arrays.
  • Latin square data
    Zipped collection of files of 3' expression data from Human U95 and U133 array containing spikes at known concentrations for the purpose of algorithm development.
3' expression GeneAtlas array strip data sets
Human Tiling 1.0 & Human U133 expression data
Data is from: Carroll J. S., et al. Genome-wide Analysis of Estrogen Receptor Binding Sites. Nature Genetics38(11):1289-97 (2006).
We thank the laboratories of Shirley Liu and Myles Brown for permission to redistribute these data.
Data is hosted on the Affymetrix Secure File Exchange (FTP) due to its size. For access to this data set, please email webmaster@affymetrix.com.
Resequencing data
This data set contains 14 Mitov2 array .cel files generated from the Human Mitochondrial Resequencing Array 2.0. The probe array files represent 3 sets of CEPH trios including 2 sets of technical replicates and 2 human mito standard samples. Human Mitochondrial Resequencing Array 2.0 ›