Choose the 1-Step Kit, if you... Choose the 2-Step Kit, if you...
  • Do not store cDNA
  • Need to store cDNA
  • Dispose samples after one or few uses
  • Have limited sample quantity
  • Have many samples with one or few targets
  • Have many targets per sample
  • Use liquid handling robotics
  • Require maximum performance of both RT and PCR steps
  • Choose to reduce chance of cross contamination during procedure
  • Need to reduce time to results
One-Step Kit Overview
Two-Step Kit Overview