real-time-pcr-testing-solution-vaginal-microbiota-sexually-transmitted-pathogens-showcase-image-230x195
Real-time PCR qualified solution for vaginal microbiota investigations
Urogenital infections, including bacterial vaginosis, urinary tract infections, and vaginitis, afflict over 1 billion women each year. Our qualified solution for vaginal microbiota investigations combines the sensitivity and specificity of Applied Biosystems TaqMan real-time PCR assays with the flexibility and scalability of the QuantStudio family of real-time PCR systems. Choose the content and format that you require to implement this flexible, scalable, and low-cost real-time PCR testing solution in your laboratory.
Get a quote
Features of this solution
  • Widest range of both commensal and pathogenic microbes compared to other currently available molecular tests
  • Qualified content including positive controls, validated user guide, and analytical validation
  • Higher specificity, accuracy, and precision compared to traditional culture and microscopy methods
  • Flexible formats—choose from four assay formats, including preloaded Applied Biosystems OpenArray plates
  • Lowest cost per sample, due in part to the low reagent costs of OpenArray nanofluidic technology compared to other commercially available solutions
 Get more information and a list of pathogen targets ›
What’s included as part of this qualified solution?
Qualified TaqMan Assays 
OpenArray format (flexible panel) 
Order now ›
OpenArray format (comprehensive panel) 
Order now ›
Single-tube assays 
Download list ›
Controls and master mixes
Amplification control  
Order now ›
Extraction control 
Order now ›
TaqMan OpenArray Master Mix  
Order now ›
Data and support
Application note 
Download ›
Workflow application guide 
Download ›
Compliance and training products/services 
Learn more ›
The role of real-time PCR in vaginal microbiota investigations
Traditional culture-based research methods for investigations into microbiota related to bacterial vaginosis can lack sensitivity and specificity, and can be subjective and inaccurate.
Quantitative PCR (qPCR) can detect slow-growing, difficult-to-cultivate, or uncultivable microorganisms, and can be used when traditional microbiological techniques are inadequate, ambiguous, time-consuming, difficult, or costly. Real-time PCR techniques provide better categorization and detection of these microorganisms. 
This flexible, scalable, and low-cost real-time PCR testing solution for vaginal microbiota offers researchers a low-cost solution for vaginal and urogenital microbiota investigations.
If you would like more information on our real-time PCR solution for vaginal microbiota, please complete the webform on this page and a sales representative will contact you directly.
Get a quote or additional information
To get a quote, or if you would like a sales representative to contact you, please complete the form below.
*
*
*
*
*
Resources
Support
© 2017 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. TaqMan is a registered trademark of Roche Molecular Systems, Inc., used under permission and license.