Gene Expression Assays
TaqMan Gene Expression Assays
Applied Biosystems TaqMan Gene Expression Assays consist of a pair of unlabeled PCR primers and a TaqMan probe with an Applied Biosystems FAM or VIC dye label on the 5’ end and minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3’ end. Over 1.8 million predesigned TaqMan Gene Expression Assays covering more than 30 species are available in a single-tube, 96-well plate, 384-well microfluidic card, and Applied Biosystems OpenArray formats.
Assay search tool
Applied Biosystems TaqMan formats and custom assays
Design your own custom TaqMan Gene Expression Assay to study any gene or splice variant in any organism.
TaqMan Gene Expression Assays are available in easy-to-use 96-well plates and higher-throughput 384-well microfluidic cards. Order from our collection of gene signature panels or customize your own plates or cards. 384-well plates can also be configured with any pre-designed or custom assay using our TaqMan™ Custom Plating Service.
Applied Biosystems TaqMan OpenArray technology is an affordable real-time PCR–based solution for studying hundreds of samples per day, using the Applied Biosystems QuantStudio 12K Flex Real-Time PCR System with OpenArray Block. Each OpenArray plate can hold as many samples as 8 traditional 384-well plates.
A convenient collection of pre-designed TaqMan Assays (probe and primer sets) enables you to normalize the amount of sample RNA or DNA in a reaction.
Watch this short video to see how easy it is to find the right TaqMan Gene Expression Assay using the Assay Search Tool above. 