Choosing the correct reverse transcriptase (RT) for cDNA synthesis is critical for obtaining high yields of quality, full-length cDNA that accurately represents the input RNA. In addition, good enzyme performance at high temperatures is important for efficient reverse transcription of RNA with secondary structure, and for specificity when using gene-specific RT primers.
Reverse transcriptase enzymes with reduced RNase H activity produce more full length cDNA, and provide greater gene product representation than enzymes with wild-type levels of RNase H activity.
SuperScript™ III Reverse Transcriptase is widely used for cDNA synthesis, due to its proven ability to enable consistently high yields of fully representative, full length cDNA.
Choose the Correct Reverse Transcriptase for cDNA Synthesis
Resources
Reverse transcription education
Review educational resources for reverse transcription basics, cDNA synthesis, enzyme selection, troubleshooting tips, and cDNA applications.
PCR & cDNA Synthesis Support Center
Find tips, troubleshooting help, and resources for your PCR & cDNA synthesis applications.